Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122404137-122404410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM162A
Entrez Gene ID
26355
Gene Name
family with sequence similarity 162 member A
Gene Aliases
C3orf28, E2IG5, HGTD-P
UniGene
Hs.584881
Gene Chromosome Location
Chr.3: 122384176-122410114 on Build GRCh38
Transcripts
NM_014367.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
160926 c.221C>T p.S74L Chr.3: 122404321-122404321 on Build GRCh38
479300 c.172G>A p.V58M Chr.3: 122404272-122404272 on Build GRCh38
4439556 c.255G>A p.E85E Chr.3: 122404355-122404355 on Build GRCh38
3845985 c.175C>T p.P59S Chr.3: 122404275-122404275 on Build GRCh38
4439555 c.255G>A p.E85E Chr.3: 122404355-122404355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376464403 Chr.3: 122404326-122404326 on Build GRCh38
rs759478287 Chr.3: 122404242-122404242 on Build GRCh38
rs746592402 Chr.3: 122404207-122404207 on Build GRCh38
rs369872384 Chr.3: 122404269-122404269 on Build GRCh38
rs1017235640 Chr.3: 122404188-122404188 on Build GRCh38
rs776066316 Chr.3: 122404231-122404231 on Build GRCh38
rs779234341 Chr.3: 122404324-122404324 on Build GRCh38
rs928355535 Chr.3: 122404199-122404199 on Build GRCh38
rs377168422 Chr.3: 122404362-122404362 on Build GRCh38
rs769819160 Chr.3: 122404363-122404363 on Build GRCh38
rs980236162 Chr.3: 122404198-122404198 on Build GRCh38
rs149572114 Chr.3: 122404327-122404327 on Build GRCh38
rs775378118 Chr.3: 122404265-122404265 on Build GRCh38
rs768322150 Chr.3: 122404374-122404374 on Build GRCh38
rs777463955 Chr.3: 122404331-122404331 on Build GRCh38
rs760893843 Chr.3: 122404292-122404292 on Build GRCh38
rs372433804 Chr.3: 122404221-122404221 on Build GRCh38
rs755537046 Chr.3: 122404314-122404314 on Build GRCh38
rs548915819 Chr.3: 122404195-122404195 on Build GRCh38
rs1020741914 Chr.3: 122404251-122404251 on Build GRCh38
rs200627653 Chr.3: 122404364-122404364 on Build GRCh38
rs747971677 Chr.3: 122404263-122404264 on Build GRCh38
rs1008226136 Chr.3: 122404187-122404187 on Build GRCh38
rs181358776 Chr.3: 122404254-122404254 on Build GRCh38
rs753902355 Chr.3: 122404304-122404304 on Build GRCh38
rs762636640 Chr.3: 122404268-122404268 on Build GRCh38
rs371721266 Chr.3: 122404298-122404298 on Build GRCh38
rs185586046 Chr.3: 122404295-122404295 on Build GRCh38
rs1045802485 Chr.3: 122404366-122404366 on Build GRCh38
rs947256688 Chr.3: 122404263-122404263 on Build GRCh38
rs372973157 Chr.3: 122404333-122404333 on Build GRCh38
rs938436079 Chr.3: 122404202-122404202 on Build GRCh38
rs138311983 Chr.3: 122404291-122404291 on Build GRCh38
rs749175952 Chr.3: 122404372-122404372 on Build GRCh38
rs199848888 Chr.3: 122404332-122404332 on Build GRCh38
rs1015707755 Chr.3: 122404353-122404353 on Build GRCh38
rs781184981 Chr.3: 122404320-122404320 on Build GRCh38
rs969753053 Chr.3: 122404164-122404164 on Build GRCh38
rs771099859 Chr.3: 122404337-122404337 on Build GRCh38
rs767822587 Chr.3: 122404284-122404284 on Build GRCh38
rs372323698 Chr.3: 122404262-122404262 on Build GRCh38

View Full Product Details