Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35291600-35291869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83C
Entrez Gene ID
128876
Gene Name
family with sequence similarity 83 member C
Gene Aliases
C20orf128, dJ614O4.7
UniGene
Hs.592149
Gene Chromosome Location
Chr.20: 35285731-35292422 on Build GRCh38
Transcripts
NM_178468.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5516088 c.512C>T p.T171M Chr.20: 35291793-35291793 on Build GRCh38
1734283 c.437C>T p.A146V Chr.20: 35291868-35291868 on Build GRCh38
5440905 c.498C>T p.F166F Chr.20: 35291807-35291807 on Build GRCh38
3545689 c.474C>T p.I158I Chr.20: 35291831-35291831 on Build GRCh38
1026210 c.468G>T p.K156N Chr.20: 35291837-35291837 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752031109 Chr.20: 35291805-35291805 on Build GRCh38
rs776690764 Chr.20: 35291792-35291792 on Build GRCh38
rs975021395 Chr.20: 35291660-35291660 on Build GRCh38
rs962401104 Chr.20: 35291823-35291823 on Build GRCh38
rs141044774 Chr.20: 35291845-35291845 on Build GRCh38
rs939716212 Chr.20: 35291729-35291729 on Build GRCh38
rs567189221 Chr.20: 35291688-35291688 on Build GRCh38
rs755472776 Chr.20: 35291818-35291818 on Build GRCh38
rs371573781 Chr.20: 35291764-35291764 on Build GRCh38
rs748475095 Chr.20: 35291754-35291754 on Build GRCh38
rs772399074 Chr.20: 35291837-35291837 on Build GRCh38
rs1038560844 Chr.20: 35291740-35291740 on Build GRCh38
rs965283432 Chr.20: 35291630-35291630 on Build GRCh38
rs202012703 Chr.20: 35291781-35291781 on Build GRCh38
rs373561979 Chr.20: 35291801-35291801 on Build GRCh38
rs2425050 Chr.20: 35291675-35291675 on Build GRCh38
rs143108902 Chr.20: 35291807-35291807 on Build GRCh38
rs905912367 Chr.20: 35291720-35291720 on Build GRCh38
rs1044378267 Chr.20: 35291708-35291708 on Build GRCh38
rs770912116 Chr.20: 35291784-35291784 on Build GRCh38
rs908208109 Chr.20: 35291718-35291718 on Build GRCh38
rs749884709 Chr.20: 35291817-35291817 on Build GRCh38
rs751317526 Chr.20: 35291785-35291785 on Build GRCh38
rs143939846 Chr.20: 35291797-35291797 on Build GRCh38
rs149089081 Chr.20: 35291793-35291793 on Build GRCh38
rs201800817 Chr.20: 35291815-35291815 on Build GRCh38
rs147288270 Chr.20: 35291824-35291824 on Build GRCh38
rs369568026 Chr.20: 35291842-35291842 on Build GRCh38
rs933571599 Chr.20: 35291686-35291686 on Build GRCh38
rs929490574 Chr.20: 35291661-35291661 on Build GRCh38
rs762818480 Chr.20: 35291798-35291798 on Build GRCh38
rs113832637 Chr.20: 35291705-35291705 on Build GRCh38

View Full Product Details