Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15923773-15924046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4F11
Entrez Gene ID
57834
Gene Name
cytochrome P450 family 4 subfamily F member 11
Gene Aliases
CYPIVF11
UniGene
Hs.187393
Gene Chromosome Location
Chr.19: 15912370-15934866 on Build GRCh38
Transcripts
NM_001128932.1 NM_021187.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
438819 c.745C>G p.L249V Chr.19: 15923985-15923985 on Build GRCh38
3529944 c.714G>A p.Q238Q Chr.19: 15924016-15924016 on Build GRCh38
3284299 c.876G>A p.K292K Chr.19: 15923854-15923854 on Build GRCh38
5686991 c.824G>T p.R275L Chr.19: 15923906-15923906 on Build GRCh38
438820 c.718C>G p.L240V Chr.19: 15924012-15924012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550454454 Chr.19: 15923966-15923966 on Build GRCh38
rs766825745 Chr.19: 15923810-15923810 on Build GRCh38
rs373866774 Chr.19: 15923839-15923839 on Build GRCh38
rs749186181 Chr.19: 15923877-15923877 on Build GRCh38
rs144938795 Chr.19: 15923961-15923961 on Build GRCh38
rs774342613 Chr.19: 15923886-15923886 on Build GRCh38
rs140695163 Chr.19: 15923947-15923947 on Build GRCh38
rs754020334 Chr.19: 15923992-15923992 on Build GRCh38
rs748732844 Chr.19: 15923935-15923935 on Build GRCh38
rs751868322 Chr.19: 15923976-15923976 on Build GRCh38
rs753048609 Chr.19: 15923840-15923840 on Build GRCh38
rs528149508 Chr.19: 15923937-15923937 on Build GRCh38
rs755586727 Chr.19: 15923924-15923924 on Build GRCh38
rs148545369 Chr.19: 15923878-15923878 on Build GRCh38
rs943435492 Chr.19: 15923948-15923948 on Build GRCh38
rs186859935 Chr.19: 15923962-15923962 on Build GRCh38
rs778958992 Chr.19: 15923938-15923938 on Build GRCh38
rs371575077 Chr.19: 15923888-15923888 on Build GRCh38
rs771046979 Chr.19: 15923960-15923960 on Build GRCh38
rs200473423 Chr.19: 15923835-15923835 on Build GRCh38
rs765712621 Chr.19: 15923909-15923909 on Build GRCh38
rs375424381 Chr.19: 15923967-15923967 on Build GRCh38
rs762740891 Chr.19: 15923842-15923842 on Build GRCh38
rs988382415 Chr.19: 15923989-15923989 on Build GRCh38
rs565062964 Chr.19: 15923965-15923965 on Build GRCh38
rs561532748 Chr.19: 15923949-15923949 on Build GRCh38
rs957963259 Chr.19: 15923915-15923915 on Build GRCh38
rs760692119 Chr.19: 15923903-15923903 on Build GRCh38
rs150621723 Chr.19: 15923906-15923906 on Build GRCh38
rs866970559 Chr.19: 15923995-15923995 on Build GRCh38
rs752623841 Chr.19: 15924001-15924001 on Build GRCh38
rs758271246 Chr.19: 15924002-15924002 on Build GRCh38
rs200851381 Chr.19: 15923815-15923815 on Build GRCh38
rs147948717 Chr.19: 15923979-15923979 on Build GRCh38
rs758723938 Chr.19: 15923910-15923910 on Build GRCh38
rs760776031 Chr.19: 15923800-15923800 on Build GRCh38
rs370476476 Chr.19: 15923923-15923923 on Build GRCh38
rs747987380 Chr.19: 15923940-15923940 on Build GRCh38
rs868544582 Chr.19: 15923944-15923944 on Build GRCh38
rs753424184 Chr.19: 15923990-15923990 on Build GRCh38
rs755605931 Chr.19: 15923846-15923846 on Build GRCh38
rs767183522 Chr.19: 15923922-15923922 on Build GRCh38
rs745418038 Chr.19: 15923845-15923845 on Build GRCh38
rs779871043 Chr.19: 15923863-15923863 on Build GRCh38
rs747187053 Chr.19: 15923950-15923950 on Build GRCh38
rs989252138 Chr.19: 15923934-15923934 on Build GRCh38
rs374048633 Chr.19: 15923895-15923895 on Build GRCh38
rs765923965 Chr.19: 15923980-15923980 on Build GRCh38
rs8104361 Chr.19: 15923904-15923904 on Build GRCh38
rs777689769 Chr.19: 15924006-15924006 on Build GRCh38
rs764006246 Chr.19: 15923896-15923896 on Build GRCh38
rs1048015544 Chr.19: 15923803-15923803 on Build GRCh38
rs779678379 Chr.19: 15923927-15923927 on Build GRCh38
rs780793484 Chr.19: 15923843-15923843 on Build GRCh38
rs201985393 Chr.19: 15923907-15923907 on Build GRCh38
rs746758825 Chr.19: 15924021-15924021 on Build GRCh38
rs578118279 Chr.19: 15923834-15923835 on Build GRCh38
rs200177921 Chr.19: 15923994-15923994 on Build GRCh38

View Full Product Details