Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 1745843-1746094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELFN1
Entrez Gene ID
392617
Gene Name
extracellular leucine rich repeat and fibronectin type III domain containing 1
Gene Aliases
PPP1R28
UniGene
Hs.42896
Gene Chromosome Location
Chr.7: 1666075-1747954 on Build GRCh38
Transcripts
NM_001128636.2 XM_006715725.3 XM_006715726.2 XM_006715727.3 XM_011515397.2 XM_011515398.2 XM_011515401.2 XM_017012203.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5522585 c.1275C>T p.C425C Chr.7: 1745871-1745871 on Build GRCh38
4779189 c.1277T>G p.L426R Chr.7: 1745873-1745873 on Build GRCh38
4527500 c.1468G>A p.G490S Chr.7: 1746064-1746064 on Build GRCh38
6547944 c.1322G>T p.R441L Chr.7: 1745918-1745918 on Build GRCh38
5450368 c.1417C>T p.P473S Chr.7: 1746013-1746013 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961425799 Chr.7: 1746018-1746018 on Build GRCh38
rs766766017 Chr.7: 1745918-1745918 on Build GRCh38
rs771148993 Chr.7: 1745968-1745968 on Build GRCh38
rs773642663 Chr.7: 1745961-1745961 on Build GRCh38
rs961820215 Chr.7: 1745890-1745890 on Build GRCh38
rs752814083 Chr.7: 1746022-1746022 on Build GRCh38
rs748845288 Chr.7: 1745938-1745938 on Build GRCh38
rs761197449 Chr.7: 1745967-1745967 on Build GRCh38
rs748975264 Chr.7: 1745898-1745898 on Build GRCh38
rs556316889 Chr.7: 1746056-1746056 on Build GRCh38
rs898819466 Chr.7: 1746072-1746072 on Build GRCh38
rs779931507 Chr.7: 1745937-1745937 on Build GRCh38
rs747802275 Chr.7: 1746075-1746075 on Build GRCh38
rs754102885 Chr.7: 1745870-1745870 on Build GRCh38
rs867918790 Chr.7: 1745997-1745997 on Build GRCh38
rs576307658 Chr.7: 1746066-1746066 on Build GRCh38
rs377622926 Chr.7: 1745881-1745881 on Build GRCh38
rs765207495 Chr.7: 1745926-1745926 on Build GRCh38
rs1030844528 Chr.7: 1745953-1745953 on Build GRCh38
rs868390199 Chr.7: 1745941-1745941 on Build GRCh38
rs191432685 Chr.7: 1746030-1746030 on Build GRCh38
rs530200834 Chr.7: 1745901-1745901 on Build GRCh38
rs779693103 Chr.7: 1746047-1746047 on Build GRCh38
rs768011818 Chr.7: 1745899-1745899 on Build GRCh38
rs988484378 Chr.7: 1746076-1746076 on Build GRCh38
rs773552624 Chr.7: 1745906-1745906 on Build GRCh38
rs561161660 Chr.7: 1745891-1745891 on Build GRCh38
rs905735267 Chr.7: 1745959-1745959 on Build GRCh38
rs566025656 Chr.7: 1745998-1745998 on Build GRCh38
rs540684381 Chr.7: 1745884-1745884 on Build GRCh38
rs938885514 Chr.7: 1745882-1745882 on Build GRCh38
rs771583831 Chr.7: 1745960-1745960 on Build GRCh38
rs563858883 Chr.7: 1745927-1745927 on Build GRCh38
rs779076976 Chr.7: 1745878-1745878 on Build GRCh38
rs1048978485 Chr.7: 1745987-1745987 on Build GRCh38
rs536488472 Chr.7: 1746031-1746031 on Build GRCh38
rs370696462 Chr.7: 1745888-1745888 on Build GRCh38
rs947640236 Chr.7: 1745868-1745868 on Build GRCh38
rs769560119 Chr.7: 1745887-1745887 on Build GRCh38
rs980371448 Chr.7: 1746019-1746019 on Build GRCh38
rs756099844 Chr.7: 1746045-1746045 on Build GRCh38
rs755247795 Chr.7: 1745924-1745924 on Build GRCh38
rs898846750 Chr.7: 1745915-1745915 on Build GRCh38
rs765564129 Chr.7: 1746010-1746010 on Build GRCh38
rs768405607 Chr.7: 1745946-1745946 on Build GRCh38
rs755798651 Chr.7: 1745930-1745930 on Build GRCh38
rs148947783 Chr.7: 1746026-1746026 on Build GRCh38
rs745358632 Chr.7: 1745877-1745877 on Build GRCh38
rs751349916 Chr.7: 1745921-1745923 on Build GRCh38
rs532829046 Chr.7: 1745928-1745928 on Build GRCh38
rs753963566 Chr.7: 1745923-1745923 on Build GRCh38
rs552293846 Chr.7: 1745956-1745956 on Build GRCh38
rs769256341 Chr.7: 1745955-1745955 on Build GRCh38
rs760970757 Chr.7: 1745917-1745917 on Build GRCh38
rs1037881811 Chr.7: 1746057-1746057 on Build GRCh38
rs746891715 Chr.7: 1745880-1745880 on Build GRCh38
rs947403023 Chr.7: 1746005-1746005 on Build GRCh38
rs778478791 Chr.7: 1746070-1746070 on Build GRCh38
rs745817796 Chr.7: 1745929-1745929 on Build GRCh38
rs535141936 Chr.7: 1746009-1746009 on Build GRCh38
rs952778525 Chr.7: 1745879-1745879 on Build GRCh38
rs550324873 Chr.7: 1745902-1745902 on Build GRCh38

View Full Product Details