Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 870994-871267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED16
Entrez Gene ID
10025
Gene Name
mediator complex subunit 16
Gene Aliases
DRIP92, THRAP5, TRAP95
UniGene
Hs.365207
Gene Chromosome Location
Chr.19: 867961-893228 on Build GRCh38
Transcripts
NM_005481.2 XM_017026120.1 XM_017026121.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6028398 c.2372+5G>A p.? Chr.19: 871032-871032 on Build GRCh38
6408537 c.2347A>G p.T783A Chr.19: 871062-871062 on Build GRCh38
6423988 c.2284C>T p.Q762* Chr.19: 871125-871125 on Build GRCh38
4636379 c.2245C>T p.R749C Chr.19: 871107-871107 on Build GRCh38
6563391 c.2367C>G p.L789L Chr.19: 871042-871042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534412257 Chr.19: 871032-871032 on Build GRCh38
rs780386131 Chr.19: 871126-871126 on Build GRCh38
rs528331323 Chr.19: 871198-871198 on Build GRCh38
rs967218806 Chr.19: 871235-871235 on Build GRCh38
rs367851435 Chr.19: 871221-871221 on Build GRCh38
rs928819614 Chr.19: 871034-871034 on Build GRCh38
rs555543361 Chr.19: 871113-871113 on Build GRCh38
rs369632896 Chr.19: 871249-871249 on Build GRCh38
rs1011201076 Chr.19: 871019-871019 on Build GRCh38
rs1003125768 Chr.19: 871234-871234 on Build GRCh38
rs764958390 Chr.19: 871106-871106 on Build GRCh38
rs745837160 Chr.19: 871223-871223 on Build GRCh38
rs532917687 Chr.19: 871142-871142 on Build GRCh38
rs559652645 Chr.19: 871138-871138 on Build GRCh38
rs747128251 Chr.19: 871218-871218 on Build GRCh38
rs772256334 Chr.19: 871133-871133 on Build GRCh38
rs757928053 Chr.19: 871013-871013 on Build GRCh38
rs1020081574 Chr.19: 871143-871143 on Build GRCh38
rs752460156 Chr.19: 871107-871107 on Build GRCh38
rs950243154 Chr.19: 871182-871182 on Build GRCh38
rs774377494 Chr.19: 871064-871064 on Build GRCh38
rs968050502 Chr.19: 871023-871023 on Build GRCh38
rs35361724 Chr.19: 871189-871189 on Build GRCh38
rs1020503561 Chr.19: 871022-871022 on Build GRCh38
rs761169713 Chr.19: 871168-871168 on Build GRCh38
rs963384721 Chr.19: 871021-871021 on Build GRCh38
rs377113997 Chr.19: 871131-871131 on Build GRCh38
rs748405693 Chr.19: 871132-871132 on Build GRCh38
rs776518471 Chr.19: 871091-871091 on Build GRCh38
rs375290182 Chr.19: 871211-871211 on Build GRCh38
rs8100258 Chr.19: 871222-871222 on Build GRCh38
rs1011170078 Chr.19: 871100-871100 on Build GRCh38
rs755355354 Chr.19: 871130-871130 on Build GRCh38
rs767039506 Chr.19: 871169-871169 on Build GRCh38
rs376802874 Chr.19: 871087-871087 on Build GRCh38
rs576500969 Chr.19: 871060-871060 on Build GRCh38
rs35799269 Chr.19: 871141-871142 on Build GRCh38
rs974605296 Chr.19: 871031-871031 on Build GRCh38
rs375447353 Chr.19: 871171-871171 on Build GRCh38
rs374408013 Chr.19: 871074-871074 on Build GRCh38
rs544723249 Chr.19: 871144-871144 on Build GRCh38
rs868381701 Chr.19: 871246-871246 on Build GRCh38
rs376670615 Chr.19: 871225-871225 on Build GRCh38
rs751147997 Chr.19: 871167-871167 on Build GRCh38
rs772753521 Chr.19: 871239-871239 on Build GRCh38
rs370073131 Chr.19: 871050-871050 on Build GRCh38
rs763793613 Chr.19: 871159-871159 on Build GRCh38
rs367666846 Chr.19: 871147-871147 on Build GRCh38
rs751092708 Chr.19: 871121-871121 on Build GRCh38
rs762694444 Chr.19: 871110-871110 on Build GRCh38
rs958765686 Chr.19: 871039-871039 on Build GRCh38
rs200816964 Chr.19: 871088-871088 on Build GRCh38
rs748356481 Chr.19: 871067-871067 on Build GRCh38
rs368744761 Chr.19: 871226-871226 on Build GRCh38
rs991522384 Chr.19: 871154-871154 on Build GRCh38
rs1015893573 Chr.19: 871020-871020 on Build GRCh38
rs371111559 Chr.19: 871149-871149 on Build GRCh38
rs34859566 Chr.19: 871044-871044 on Build GRCh38
rs368424152 Chr.19: 871187-871187 on Build GRCh38
rs372917971 Chr.19: 871093-871093 on Build GRCh38
rs370307890 Chr.19: 871071-871071 on Build GRCh38
rs8100009 Chr.19: 871048-871048 on Build GRCh38
rs201156340 Chr.19: 871042-871042 on Build GRCh38
rs373503269 Chr.19: 871250-871250 on Build GRCh38
rs907761214 Chr.19: 871053-871053 on Build GRCh38
rs539478412 Chr.19: 871047-871047 on Build GRCh38
rs754241655 Chr.19: 871128-871128 on Build GRCh38
rs937343648 Chr.19: 871056-871056 on Build GRCh38
rs756668412 Chr.19: 871125-871125 on Build GRCh38
rs753015504 Chr.19: 871072-871072 on Build GRCh38
rs902883254 Chr.19: 871232-871232 on Build GRCh38
rs373765974 Chr.19: 871092-871092 on Build GRCh38
rs761520853 Chr.19: 871231-871231 on Build GRCh38
rs546895377 Chr.19: 871216-871216 on Build GRCh38
rs759978778 Chr.19: 871248-871248 on Build GRCh38
rs1683569 Chr.19: 871135-871135 on Build GRCh38
rs749568080 Chr.19: 871051-871051 on Build GRCh38
rs957075891 Chr.19: 871016-871016 on Build GRCh38
rs72984029 Chr.19: 871033-871033 on Build GRCh38
rs34110403 Chr.19: 871146-871146 on Build GRCh38
rs763076728 Chr.19: 871238-871238 on Build GRCh38
rs769749572 Chr.19: 871224-871224 on Build GRCh38
rs777412850 Chr.19: 871014-871014 on Build GRCh38
rs376654451 Chr.19: 871230-871230 on Build GRCh38
rs1033087086 Chr.19: 871241-871241 on Build GRCh38
rs1037440613 Chr.19: 871201-871201 on Build GRCh38
rs200579290 Chr.19: 871217-871217 on Build GRCh38
rs373485164 Chr.19: 871227-871227 on Build GRCh38
rs940608481 Chr.19: 871186-871186 on Build GRCh38

View Full Product Details