Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 98868348-98868573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD1
Entrez Gene ID
1105
Gene Name
chromodomain helicase DNA binding protein 1
Gene Aliases
-
UniGene
Hs.643465
Gene Chromosome Location
Chr.5: 98855204-98929014 on Build GRCh38
Transcripts
NM_001270.2 XM_005271866.4 XM_005271867.4 XM_011543112.2 XM_017008991.1 XM_017008992.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
50910 c.4234A>T p.K1412* Chr.5: 98868509-98868509 on Build GRCh38
5939346 c.4210G>T p.E1404* Chr.5: 98868533-98868533 on Build GRCh38
5775201 c.4236G>T p.K1412N Chr.5: 98868507-98868507 on Build GRCh38
4818249 c.4192G>A p.E1398K Chr.5: 98868551-98868551 on Build GRCh38
5563288 c.4231C>T p.Q1411* Chr.5: 98868512-98868512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147154039 Chr.5: 98868499-98868499 on Build GRCh38
rs766493819 Chr.5: 98868484-98868485 on Build GRCh38
rs371008887 Chr.5: 98868528-98868528 on Build GRCh38
rs564010044 Chr.5: 98868501-98868501 on Build GRCh38
rs778934817 Chr.5: 98868448-98868448 on Build GRCh38
rs772172931 Chr.5: 98868462-98868462 on Build GRCh38
rs545709692 Chr.5: 98868489-98868489 on Build GRCh38
rs373027854 Chr.5: 98868457-98868457 on Build GRCh38
rs373556965 Chr.5: 98868546-98868546 on Build GRCh38
rs771286539 Chr.5: 98868484-98868484 on Build GRCh38
rs530973976 Chr.5: 98868504-98868504 on Build GRCh38
rs750415851 Chr.5: 98868469-98868470 on Build GRCh38
rs767527743 Chr.5: 98868465-98868466 on Build GRCh38
rs751153238 Chr.5: 98868520-98868520 on Build GRCh38
rs1016569341 Chr.5: 98868545-98868545 on Build GRCh38
rs977133645 Chr.5: 98868534-98868534 on Build GRCh38
rs763663466 Chr.5: 98868505-98868505 on Build GRCh38
rs59294148 Chr.5: 98868391-98868391 on Build GRCh38
rs906247970 Chr.5: 98868406-98868406 on Build GRCh38
rs1035516217 Chr.5: 98868478-98868478 on Build GRCh38
rs1002742692 Chr.5: 98868471-98868471 on Build GRCh38
rs139716875 Chr.5: 98868483-98868483 on Build GRCh38
rs756211620 Chr.5: 98868479-98868479 on Build GRCh38
rs773425844 Chr.5: 98868466-98868466 on Build GRCh38
rs552987523 Chr.5: 98868442-98868442 on Build GRCh38

View Full Product Details