Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7939420-7939667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTROB
Entrez Gene ID
116840
Gene Name
centrobin, centriole duplication and spindle assembly protein
Gene Aliases
LIP8, PP1221
UniGene
Hs.348012
Gene Chromosome Location
Chr.17: 7932054-7949919 on Build GRCh38
Transcripts
NM_001037144.5 NM_053051.3 XM_005256437.2 XM_005256438.2 XM_005256439.2 XM_017024128.1 XM_017024129.1 XM_017024130.1 XM_017024131.1 XM_017024132.1 XM_017024133.1 XM_017024134.1 XM_017024135.1 XM_017024136.1 XM_017024137.1 XM_017024138.1 XM_017024139.1 XM_017024140.1 XM_017024141.1 XM_017024142.1 XM_017024143.1 XM_017024144.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6235623 c.977G>A p.R326Q Chr.17: 7939562-7939562 on Build GRCh38
4070521 c.943G>T p.A315S Chr.17: 7939528-7939528 on Build GRCh38
7153651 c.933G>C p.E311D Chr.17: 7939518-7939518 on Build GRCh38
6632046 c.992A>C p.Q331P Chr.17: 7939577-7939577 on Build GRCh38
4938854 c.945T>C p.A315A Chr.17: 7939530-7939530 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780007077 Chr.17: 7939505-7939505 on Build GRCh38
rs747961408 Chr.17: 7939619-7939619 on Build GRCh38
rs201649892 Chr.17: 7939640-7939640 on Build GRCh38
rs535226060 Chr.17: 7939550-7939550 on Build GRCh38
rs141146363 Chr.17: 7939639-7939639 on Build GRCh38
rs190429332 Chr.17: 7939504-7939504 on Build GRCh38
rs966098620 Chr.17: 7939478-7939478 on Build GRCh38
rs143067938 Chr.17: 7939553-7939553 on Build GRCh38
rs771196866 Chr.17: 7939515-7939515 on Build GRCh38
rs751501555 Chr.17: 7939603-7939603 on Build GRCh38
rs762146766 Chr.17: 7939585-7939585 on Build GRCh38
rs781076257 Chr.17: 7939607-7939607 on Build GRCh38
rs186228515 Chr.17: 7939460-7939460 on Build GRCh38
rs778232253 Chr.17: 7939493-7939493 on Build GRCh38
rs113662563 Chr.17: 7939500-7939500 on Build GRCh38
rs769518030 Chr.17: 7939622-7939622 on Build GRCh38
rs774493362 Chr.17: 7939517-7939517 on Build GRCh38
rs372689651 Chr.17: 7939503-7939503 on Build GRCh38
rs747404196 Chr.17: 7939539-7939540 on Build GRCh38
rs780226443 Chr.17: 7939538-7939539 on Build GRCh38
rs753451693 Chr.17: 7939477-7939477 on Build GRCh38
rs750079552 Chr.17: 7939592-7939592 on Build GRCh38
rs749452226 Chr.17: 7939513-7939513 on Build GRCh38
rs768519053 Chr.17: 7939512-7939512 on Build GRCh38
rs764378498 Chr.17: 7939570-7939570 on Build GRCh38
rs546781575 Chr.17: 7939548-7939548 on Build GRCh38
rs564339167 Chr.17: 7939491-7939491 on Build GRCh38
rs148206376 Chr.17: 7939562-7939562 on Build GRCh38
rs754216809 Chr.17: 7939577-7939577 on Build GRCh38
rs764942032 Chr.17: 7939586-7939586 on Build GRCh38
rs754969156 Chr.17: 7939605-7939605 on Build GRCh38
rs779173991 Chr.17: 7939634-7939634 on Build GRCh38
rs537832109 Chr.17: 7939453-7939453 on Build GRCh38
rs374479317 Chr.17: 7939518-7939518 on Build GRCh38
rs746990006 Chr.17: 7939510-7939510 on Build GRCh38
rs757898540 Chr.17: 7939593-7939593 on Build GRCh38
rs769248537 Chr.17: 7939636-7939637 on Build GRCh38
rs905933564 Chr.17: 7939598-7939598 on Build GRCh38
rs779730791 Chr.17: 7939597-7939597 on Build GRCh38
rs377613708 Chr.17: 7939561-7939561 on Build GRCh38

View Full Product Details