Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 52314404-52314677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKK1
Entrez Gene ID
22943
Gene Name
dickkopf WNT signaling pathway inhibitor 1
Gene Aliases
DKK-1, SK
UniGene
Hs.40499
Gene Chromosome Location
Chr.10: 52314281-52317657 on Build GRCh38
Transcripts
NM_012242.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
685014 c.120C>A p.N40K Chr.10: 52314554-52314554 on Build GRCh38
3438686 c.152G>A p.G51D Chr.10: 52314586-52314586 on Build GRCh38
4938715 c.227C>A p.T76N Chr.10: 52314661-52314661 on Build GRCh38
337214 c.140C>A p.P47Q Chr.10: 52314574-52314574 on Build GRCh38
685013 c.176C>A p.A59E Chr.10: 52314610-52314610 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756723210 Chr.10: 52314602-52314602 on Build GRCh38
rs145640971 Chr.10: 52314555-52314555 on Build GRCh38
rs770544123 Chr.10: 52314504-52314504 on Build GRCh38
rs753773509 Chr.10: 52314653-52314653 on Build GRCh38
rs773252103 Chr.10: 52314626-52314626 on Build GRCh38
rs199874560 Chr.10: 52314437-52314437 on Build GRCh38
rs200786704 Chr.10: 52314509-52314509 on Build GRCh38
rs545596036 Chr.10: 52314450-52314450 on Build GRCh38
rs949343801 Chr.10: 52314483-52314483 on Build GRCh38
rs925067853 Chr.10: 52314430-52314430 on Build GRCh38
rs756819696 Chr.10: 52314609-52314609 on Build GRCh38
rs765494161 Chr.10: 52314476-52314476 on Build GRCh38
rs200283314 Chr.10: 52314524-52314524 on Build GRCh38
rs759732360 Chr.10: 52314545-52314546 on Build GRCh38
rs756021063 Chr.10: 52314495-52314495 on Build GRCh38
rs760871535 Chr.10: 52314466-52314466 on Build GRCh38
rs760457793 Chr.10: 52314638-52314638 on Build GRCh38
rs748757559 Chr.10: 52314593-52314593 on Build GRCh38
rs776076442 Chr.10: 52314452-52314452 on Build GRCh38
rs146628250 Chr.10: 52314645-52314645 on Build GRCh38
rs368619028 Chr.10: 52314545-52314545 on Build GRCh38
rs140471040 Chr.10: 52314480-52314480 on Build GRCh38
rs756108629 Chr.10: 52314584-52314584 on Build GRCh38
rs778465064 Chr.10: 52314603-52314603 on Build GRCh38
rs766580859 Chr.10: 52314633-52314633 on Build GRCh38
rs781137306 Chr.10: 52314482-52314482 on Build GRCh38
rs1021172322 Chr.10: 52314562-52314562 on Build GRCh38
rs547799488 Chr.10: 52314570-52314570 on Build GRCh38
rs34795541 Chr.10: 52314620-52314620 on Build GRCh38
rs753934097 Chr.10: 52314468-52314468 on Build GRCh38
rs372730677 Chr.10: 52314615-52314615 on Build GRCh38
rs759961935 Chr.10: 52314447-52314447 on Build GRCh38
rs748824039 Chr.10: 52314500-52314500 on Build GRCh38
rs761830028 Chr.10: 52314621-52314621 on Build GRCh38
rs1049002817 Chr.10: 52314459-52314459 on Build GRCh38
rs776643011 Chr.10: 52314540-52314540 on Build GRCh38
rs183906578 Chr.10: 52314512-52314512 on Build GRCh38
rs542440178 Chr.10: 52314510-52314510 on Build GRCh38
rs763200950 Chr.10: 52314628-52314628 on Build GRCh38
rs777749212 Chr.10: 52314589-52314589 on Build GRCh38
rs752455729 Chr.10: 52314573-52314573 on Build GRCh38
rs989330025 Chr.10: 52314556-52314556 on Build GRCh38
rs750617685 Chr.10: 52314558-52314558 on Build GRCh38
rs748027511 Chr.10: 52314493-52314493 on Build GRCh38
rs777185220 Chr.10: 52314497-52314497 on Build GRCh38
rs777138901 Chr.10: 52314618-52314618 on Build GRCh38
rs527963973 Chr.10: 52314539-52314539 on Build GRCh38
rs931520011 Chr.10: 52314449-52314449 on Build GRCh38
rs750566358 Chr.10: 52314463-52314463 on Build GRCh38
rs771830747 Chr.10: 52314604-52314604 on Build GRCh38
rs769101264 Chr.10: 52314617-52314617 on Build GRCh38
rs780608058 Chr.10: 52314606-52314606 on Build GRCh38
rs751167321 Chr.10: 52314479-52314479 on Build GRCh38
rs767347236 Chr.10: 52314572-52314572 on Build GRCh38
rs769862048 Chr.10: 52314623-52314623 on Build GRCh38
rs772528448 Chr.10: 52314520-52314520 on Build GRCh38
rs987770662 Chr.10: 52314465-52314465 on Build GRCh38
rs764223547 Chr.10: 52314467-52314467 on Build GRCh38
rs887336853 Chr.10: 52314608-52314608 on Build GRCh38

View Full Product Details