Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 230172787-230173016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SP110
Entrez Gene ID
3431
Gene Name
SP110 nuclear body protein
Gene Aliases
IFI41, IFI75, IPR1, VODI
UniGene
Hs.145150
Gene Chromosome Location
Chr.2: 230165466-230225729 on Build GRCh38
Transcripts
NM_004509.3 NM_080424.2 XM_005246525.3 XM_006712487.3 XM_006712489.3 XM_011511088.2 XM_011511089.2 XM_011511090.2 XM_011511091.2 XM_011511092.2 XM_017003968.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6574646 c.1606G>A p.E536K Chr.2: 230172944-230172944 on Build GRCh38
3578863 c.1668C>T p.F556F Chr.2: 230172882-230172882 on Build GRCh38
129365 c.1591C>T p.R531W Chr.2: 230172959-230172959 on Build GRCh38
5006254 c.1647C>T p.C549C Chr.2: 230172903-230172903 on Build GRCh38
273289 c.1611C>T p.C537C Chr.2: 230172939-230172939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764475133 Chr.2: 230172986-230172986 on Build GRCh38
rs773059144 Chr.2: 230172902-230172902 on Build GRCh38
rs201049659 Chr.2: 230172985-230172985 on Build GRCh38
rs781587648 Chr.2: 230172863-230172863 on Build GRCh38
rs376116343 Chr.2: 230172911-230172911 on Build GRCh38
rs200523969 Chr.2: 230172954-230172954 on Build GRCh38
rs747551994 Chr.2: 230172844-230172844 on Build GRCh38
rs201856437 Chr.2: 230172969-230172969 on Build GRCh38
rs201034113 Chr.2: 230172840-230172840 on Build GRCh38
rs767364651 Chr.2: 230172808-230172808 on Build GRCh38
rs199516292 Chr.2: 230172951-230172951 on Build GRCh38
rs758513207 Chr.2: 230172923-230172923 on Build GRCh38
rs935474158 Chr.2: 230172874-230172874 on Build GRCh38
rs200317062 Chr.2: 230172965-230172965 on Build GRCh38
rs371526319 Chr.2: 230172905-230172905 on Build GRCh38
rs199745658 Chr.2: 230172889-230172889 on Build GRCh38
rs751675699 Chr.2: 230172995-230172995 on Build GRCh38
rs200099412 Chr.2: 230172959-230172959 on Build GRCh38
rs201738789 Chr.2: 230172958-230172958 on Build GRCh38
rs746284144 Chr.2: 230172878-230172878 on Build GRCh38
rs757775938 Chr.2: 230172939-230172939 on Build GRCh38
rs200378335 Chr.2: 230172819-230172819 on Build GRCh38
rs778358013 Chr.2: 230172841-230172841 on Build GRCh38
rs756439485 Chr.2: 230172948-230172948 on Build GRCh38
rs367869427 Chr.2: 230172942-230172942 on Build GRCh38
rs35495464 Chr.2: 230172900-230172900 on Build GRCh38
rs201204126 Chr.2: 230172820-230172820 on Build GRCh38
rs267599251 Chr.2: 230172882-230172882 on Build GRCh38
rs925484782 Chr.2: 230172849-230172849 on Build GRCh38
rs761714428 Chr.2: 230172901-230172901 on Build GRCh38
rs149682257 Chr.2: 230172938-230172938 on Build GRCh38
rs200620941 Chr.2: 230172916-230172916 on Build GRCh38
rs547427992 Chr.2: 230172949-230172949 on Build GRCh38
rs200267588 Chr.2: 230172825-230172825 on Build GRCh38
rs200559722 Chr.2: 230172890-230172890 on Build GRCh38
rs766118945 Chr.2: 230172976-230172976 on Build GRCh38
rs368862360 Chr.2: 230172827-230172827 on Build GRCh38
rs757517084 Chr.2: 230172857-230172857 on Build GRCh38
rs199943105 Chr.2: 230172977-230172977 on Build GRCh38
rs116626378 Chr.2: 230172903-230172903 on Build GRCh38
rs754021147 Chr.2: 230172910-230172912 on Build GRCh38
rs764293077 Chr.2: 230172933-230172933 on Build GRCh38
rs757297259 Chr.2: 230172996-230172996 on Build GRCh38
rs768181968 Chr.2: 230172897-230172897 on Build GRCh38
rs202101309 Chr.2: 230172968-230172968 on Build GRCh38
rs765446789 Chr.2: 230172906-230172906 on Build GRCh38
rs746185280 Chr.2: 230172944-230172944 on Build GRCh38
rs760451870 Chr.2: 230172973-230172973 on Build GRCh38
rs773822608 Chr.2: 230172963-230172963 on Build GRCh38
rs192671550 Chr.2: 230172824-230172824 on Build GRCh38

View Full Product Details