Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 119808600-119808873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REG4
Entrez Gene ID
83998
Gene Name
regenerating family member 4
Gene Aliases
GISP, REG-IV, RELP
UniGene
Hs.660883
Gene Chromosome Location
Chr.1: 119794018-119811580 on Build GRCh38
Transcripts
NM_001159352.1 NM_001159353.1 NM_032044.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6120842 c.35T>A p.L12Q Chr.1: 119808735-119808735 on Build GRCh38
5024913 c.11G>A p.R4K Chr.1: 119808759-119808759 on Build GRCh38
2179954 c.8C>T p.S3F Chr.1: 119808762-119808762 on Build GRCh38
5025021 c.59T>A p.V20D Chr.1: 119808711-119808711 on Build GRCh38
6120844 c.46G>T p.A16S Chr.1: 119808724-119808724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752990498 Chr.1: 119808689-119808689 on Build GRCh38
rs587601757 Chr.1: 119808837-119808837 on Build GRCh38
rs768175778 Chr.1: 119808726-119808726 on Build GRCh38
rs769419888 Chr.1: 119808751-119808751 on Build GRCh38
rs376551362 Chr.1: 119808652-119808652 on Build GRCh38
rs758770577 Chr.1: 119808698-119808698 on Build GRCh38
rs971846984 Chr.1: 119808776-119808776 on Build GRCh38
rs759644251 Chr.1: 119808774-119808774 on Build GRCh38
rs184114427 Chr.1: 119808843-119808843 on Build GRCh38
rs759733203 Chr.1: 119808687-119808687 on Build GRCh38
rs1048360907 Chr.1: 119808767-119808767 on Build GRCh38
rs766484084 Chr.1: 119808670-119808670 on Build GRCh38
rs781750395 Chr.1: 119808793-119808793 on Build GRCh38
rs767705648 Chr.1: 119808790-119808790 on Build GRCh38
rs757775996 Chr.1: 119808703-119808703 on Build GRCh38
rs747842213 Chr.1: 119808750-119808750 on Build GRCh38
rs746500336 Chr.1: 119808705-119808705 on Build GRCh38
rs191792503 Chr.1: 119808819-119808819 on Build GRCh38
rs1044652177 Chr.1: 119808816-119808816 on Build GRCh38
rs769662675 Chr.1: 119808664-119808664 on Build GRCh38
rs138324426 Chr.1: 119808765-119808765 on Build GRCh38
rs77250186 Chr.1: 119808704-119808704 on Build GRCh38
rs1030527807 Chr.1: 119808787-119808787 on Build GRCh38
rs369004688 Chr.1: 119808824-119808824 on Build GRCh38
rs587772457 Chr.1: 119808633-119808633 on Build GRCh38
rs772956554 Chr.1: 119808666-119808666 on Build GRCh38
rs929074629 Chr.1: 119808766-119808766 on Build GRCh38
rs779921882 Chr.1: 119808783-119808783 on Build GRCh38
rs764286101 Chr.1: 119808810-119808810 on Build GRCh38
rs751688232 Chr.1: 119808675-119808675 on Build GRCh38
rs780783584 Chr.1: 119808738-119808738 on Build GRCh38
rs756363605 Chr.1: 119808700-119808700 on Build GRCh38
rs754209624 Chr.1: 119808702-119808702 on Build GRCh38
rs373430078 Chr.1: 119808792-119808792 on Build GRCh38
rs1023537323 Chr.1: 119808640-119808640 on Build GRCh38
rs774483417 Chr.1: 119808772-119808772 on Build GRCh38
rs587768640 Chr.1: 119808688-119808688 on Build GRCh38
rs587675215 Chr.1: 119808635-119808635 on Build GRCh38
rs765683974 Chr.1: 119808821-119808821 on Build GRCh38
rs368686120 Chr.1: 119808701-119808701 on Build GRCh38

View Full Product Details