Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218621403-218621668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLCD4
Entrez Gene ID
84812
Gene Name
phospholipase C delta 4
Gene Aliases
-
UniGene
Hs.632528
Gene Chromosome Location
Chr.2: 218606873-218637186 on Build GRCh38
Transcripts
NM_032726.3 XM_005246913.2 XM_011512012.1 XM_017005114.1 XM_017005115.1 XM_017005116.1 XM_017005117.1 XM_017005118.1 XM_017005119.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1016399 c.517G>T p.E173* Chr.2: 218621576-218621576 on Build GRCh38
5579331 c.421G>A p.D141N Chr.2: 218621480-218621480 on Build GRCh38
6528735 c.416T>C p.L139P Chr.2: 218621475-218621475 on Build GRCh38
7336471 c.472G>C p.E158Q Chr.2: 218621531-218621531 on Build GRCh38
1016397 c.505G>A p.E169K Chr.2: 218621564-218621564 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751590769 Chr.2: 218621524-218621524 on Build GRCh38
rs369778322 Chr.2: 218621626-218621626 on Build GRCh38
rs927200236 Chr.2: 218621612-218621612 on Build GRCh38
rs777463472 Chr.2: 218621439-218621439 on Build GRCh38
rs373037584 Chr.2: 218621637-218621637 on Build GRCh38
rs983170050 Chr.2: 218621469-218621469 on Build GRCh38
rs144457458 Chr.2: 218621428-218621431 on Build GRCh38
rs770475168 Chr.2: 218621580-218621580 on Build GRCh38
rs754714559 Chr.2: 218621541-218621541 on Build GRCh38
rs200369357 Chr.2: 218621437-218621437 on Build GRCh38
rs775884496 Chr.2: 218621619-218621619 on Build GRCh38
rs866284356 Chr.2: 218621531-218621531 on Build GRCh38
rs374046683 Chr.2: 218621529-218621529 on Build GRCh38
rs904039904 Chr.2: 218621573-218621573 on Build GRCh38
rs376944300 Chr.2: 218621560-218621560 on Build GRCh38
rs774974886 Chr.2: 218621509-218621509 on Build GRCh38
rs763205099 Chr.2: 218621512-218621512 on Build GRCh38
rs766163610 Chr.2: 218621536-218621536 on Build GRCh38
rs1031292053 Chr.2: 218621621-218621621 on Build GRCh38
rs868795724 Chr.2: 218621513-218621513 on Build GRCh38
rs575368715 Chr.2: 218621466-218621466 on Build GRCh38
rs894251482 Chr.2: 218621539-218621539 on Build GRCh38
rs771702152 Chr.2: 218621423-218621423 on Build GRCh38
rs752364295 Chr.2: 218621547-218621547 on Build GRCh38
rs778917703 Chr.2: 218621546-218621546 on Build GRCh38
rs774561457 Chr.2: 218621633-218621633 on Build GRCh38
rs564116874 Chr.2: 218621540-218621540 on Build GRCh38
rs906859909 Chr.2: 218621493-218621493 on Build GRCh38
rs758171195 Chr.2: 218621549-218621549 on Build GRCh38
rs746457248 Chr.2: 218621611-218621611 on Build GRCh38
rs748127056 Chr.2: 218621479-218621479 on Build GRCh38
rs938394736 Chr.2: 218621496-218621496 on Build GRCh38
rs528518728 Chr.2: 218621613-218621613 on Build GRCh38
rs763443371 Chr.2: 218621422-218621422 on Build GRCh38
rs770256179 Chr.2: 218621482-218621482 on Build GRCh38
rs200222852 Chr.2: 218621480-218621480 on Build GRCh38
rs375920010 Chr.2: 218621446-218621446 on Build GRCh38
rs1000148666 Chr.2: 218621604-218621604 on Build GRCh38
rs919776160 Chr.2: 218621452-218621452 on Build GRCh38
rs545914229 Chr.2: 218621492-218621492 on Build GRCh38
rs771290198 Chr.2: 218621457-218621457 on Build GRCh38
rs746459213 Chr.2: 218621567-218621567 on Build GRCh38
rs951782139 Chr.2: 218621451-218621451 on Build GRCh38
rs781541758 Chr.2: 218621608-218621608 on Build GRCh38

View Full Product Details