Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 100675953-100676210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYTL4
Entrez Gene ID
94121
Gene Name
synaptotagmin like 4
Gene Aliases
SLP4
UniGene
Hs.592224
Gene Chromosome Location
Chr.X: 100671964-100732148 on Build GRCh38
Transcripts
NM_001129896.2 NM_001174068.1 NM_080737.2 XM_005262229.2 XM_011531068.2 XM_017029966.1 XM_017029967.1 XM_017029968.1 XM_017029969.1 XM_017029970.1 XM_017029971.1

SNP Detail

SNP ID Chromosome Location
rs745368437 Chr.X: 100676063-100676063 on Build GRCh38
rs749283202 Chr.X: 100676095-100676095 on Build GRCh38
rs776842282 Chr.X: 100676141-100676141 on Build GRCh38
rs367971616 Chr.X: 100676039-100676039 on Build GRCh38
rs887527150 Chr.X: 100676180-100676180 on Build GRCh38
rs773413485 Chr.X: 100676136-100676136 on Build GRCh38
rs139707843 Chr.X: 100676062-100676062 on Build GRCh38
rs745402438 Chr.X: 100676076-100676076 on Build GRCh38
rs760160020 Chr.X: 100676171-100676171 on Build GRCh38
rs779981518 Chr.X: 100676091-100676091 on Build GRCh38
rs754334762 Chr.X: 100676027-100676027 on Build GRCh38
rs186949124 Chr.X: 100675994-100675994 on Build GRCh38
rs767752854 Chr.X: 100675987-100675987 on Build GRCh38
rs780848178 Chr.X: 100676068-100676068 on Build GRCh38
rs148558767 Chr.X: 100676139-100676139 on Build GRCh38
rs765817223 Chr.X: 100676175-100676175 on Build GRCh38
rs200526563 Chr.X: 100675995-100675995 on Build GRCh38
rs937097691 Chr.X: 100676081-100676081 on Build GRCh38
rs766996024 Chr.X: 100676002-100676002 on Build GRCh38
rs765763966 Chr.X: 100676058-100676058 on Build GRCh38
rs1049930392 Chr.X: 100675999-100675999 on Build GRCh38
rs755586303 Chr.X: 100676077-100676077 on Build GRCh38
rs765447537 Chr.X: 100675986-100675986 on Build GRCh38
rs1040530389 Chr.X: 100676079-100676079 on Build GRCh38
rs868291409 Chr.X: 100676134-100676134 on Build GRCh38
rs755524501 Chr.X: 100676053-100676053 on Build GRCh38
rs768580707 Chr.X: 100676100-100676100 on Build GRCh38
rs143469082 Chr.X: 100676106-100676106 on Build GRCh38
rs775187605 Chr.X: 100676140-100676140 on Build GRCh38
rs774175887 Chr.X: 100676103-100676103 on Build GRCh38
rs747955441 Chr.X: 100676104-100676104 on Build GRCh38
rs926812087 Chr.X: 100676071-100676071 on Build GRCh38

View Full Product Details