Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 210188870-210189129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADL
Entrez Gene ID
33
Gene Name
acyl-CoA dehydrogenase, long chain
Gene Aliases
ACAD4, LCAD
UniGene
Hs.471277
Gene Chromosome Location
Chr.2: 210187990-210225491 on Build GRCh38
Transcripts
NM_001608.3 XM_005246517.4 XM_017003955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1015834 c.1268C>T p.A423V Chr.2: 210188986-210188986 on Build GRCh38
5984706 c.1281C>G p.V427V Chr.2: 210188973-210188973 on Build GRCh38
5647351 c.1246G>T p.E416* Chr.2: 210189008-210189008 on Build GRCh38
1721424 c.1254G>A p.M418I Chr.2: 210189000-210189000 on Build GRCh38
6591669 c.1292A>C p.*431S Chr.2: 210188962-210188962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370932848 Chr.2: 210188920-210188920 on Build GRCh38
rs765041497 Chr.2: 210188949-210188949 on Build GRCh38
rs751128035 Chr.2: 210188980-210188980 on Build GRCh38
rs763588960 Chr.2: 210188970-210188970 on Build GRCh38
rs779939047 Chr.2: 210189093-210189094 on Build GRCh38
rs542568647 Chr.2: 210188946-210188946 on Build GRCh38
rs767579812 Chr.2: 210188941-210188941 on Build GRCh38
rs899011907 Chr.2: 210189076-210189076 on Build GRCh38
rs761007593 Chr.2: 210189090-210189090 on Build GRCh38
rs756382365 Chr.2: 210189033-210189033 on Build GRCh38
rs778518179 Chr.2: 210188921-210188937 on Build GRCh38
rs775497317 Chr.2: 210189026-210189026 on Build GRCh38
rs763960636 Chr.2: 210189096-210189096 on Build GRCh38
rs372809258 Chr.2: 210188903-210188903 on Build GRCh38
rs562293303 Chr.2: 210189094-210189094 on Build GRCh38
rs200346431 Chr.2: 210188977-210188977 on Build GRCh38
rs764988272 Chr.2: 210189091-210189091 on Build GRCh38
rs771868717 Chr.2: 210189066-210189067 on Build GRCh38
rs745563670 Chr.2: 210189021-210189021 on Build GRCh38
rs780641408 Chr.2: 210189020-210189020 on Build GRCh38
rs768829914 Chr.2: 210188910-210188910 on Build GRCh38
rs758373054 Chr.2: 210188963-210188963 on Build GRCh38
rs748113818 Chr.2: 210189066-210189066 on Build GRCh38
rs769348791 Chr.2: 210189048-210189048 on Build GRCh38
rs773259187 Chr.2: 210189089-210189089 on Build GRCh38
rs756803658 Chr.2: 210188988-210188988 on Build GRCh38
rs774642882 Chr.2: 210188921-210188921 on Build GRCh38
rs747028647 Chr.2: 210189086-210189086 on Build GRCh38
rs780610346 Chr.2: 210189040-210189040 on Build GRCh38
rs752609044 Chr.2: 210188960-210188960 on Build GRCh38
rs769884532 Chr.2: 210188934-210188934 on Build GRCh38
rs370174318 Chr.2: 210188981-210188981 on Build GRCh38
rs745700699 Chr.2: 210188932-210188933 on Build GRCh38
rs909370695 Chr.2: 210188979-210188979 on Build GRCh38
rs775016696 Chr.2: 210189058-210189058 on Build GRCh38
rs749778106 Chr.2: 210189046-210189046 on Build GRCh38
rs772228742 Chr.2: 210189084-210189084 on Build GRCh38

View Full Product Details