Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8366041-8366276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL4
Entrez Gene ID
51129
Gene Name
angiopoietin like 4
Gene Aliases
ARP4, FIAF, HARP, HFARP, NL2, PGAR, TGQTL, UNQ171, pp1158
UniGene
Hs.9613
Gene Chromosome Location
Chr.19: 8364127-8374375 on Build GRCh38
Transcripts
NM_001039667.2 NM_139314.2 NR_104213.1 XM_005272484.3 XM_005272485.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
227632 c.407G>A p.R136Q Chr.19: 8366042-8366042 on Build GRCh38
3681119 c.429G>A p.Q143Q Chr.19: 8366064-8366064 on Build GRCh38
3681120 c.429G>C p.Q143H Chr.19: 8366064-8366064 on Build GRCh38
3541587 c.442G>A p.D148N Chr.19: 8366214-8366214 on Build GRCh38
4749594 c.469G>A p.A157T Chr.19: 8366241-8366241 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs3210979 Chr.19: 8366203-8366203 on Build GRCh38
rs765669768 Chr.19: 8366222-8366222 on Build GRCh38
rs201770562 Chr.19: 8366253-8366253 on Build GRCh38
rs747685487 Chr.19: 8366237-8366237 on Build GRCh38
rs186372928 Chr.19: 8366112-8366112 on Build GRCh38
rs73925320 Chr.19: 8366143-8366143 on Build GRCh38
rs951801645 Chr.19: 8366142-8366142 on Build GRCh38
rs1030677144 Chr.19: 8366230-8366230 on Build GRCh38
rs751370400 Chr.19: 8366113-8366113 on Build GRCh38
rs898783345 Chr.19: 8366094-8366094 on Build GRCh38
rs772487616 Chr.19: 8366164-8366164 on Build GRCh38
rs777171576 Chr.19: 8366074-8366074 on Build GRCh38
rs751194272 Chr.19: 8366240-8366240 on Build GRCh38
rs955089763 Chr.19: 8366250-8366250 on Build GRCh38
rs775839013 Chr.19: 8366184-8366184 on Build GRCh38
rs370365380 Chr.19: 8366189-8366189 on Build GRCh38
rs749881519 Chr.19: 8366085-8366085 on Build GRCh38
rs758054099 Chr.19: 8366093-8366093 on Build GRCh38
rs911396368 Chr.19: 8366079-8366079 on Build GRCh38
rs762440462 Chr.19: 8366234-8366234 on Build GRCh38
rs754546802 Chr.19: 8366241-8366241 on Build GRCh38
rs373597368 Chr.19: 8366115-8366115 on Build GRCh38
rs368376759 Chr.19: 8366246-8366246 on Build GRCh38
rs771453273 Chr.19: 8366210-8366210 on Build GRCh38
rs942918502 Chr.19: 8366090-8366090 on Build GRCh38
rs755956389 Chr.19: 8366153-8366153 on Build GRCh38
rs145223309 Chr.19: 8366116-8366116 on Build GRCh38
rs374733561 Chr.19: 8366190-8366190 on Build GRCh38
rs756125612 Chr.19: 8366248-8366248 on Build GRCh38
rs772778640 Chr.19: 8366233-8366233 on Build GRCh38
rs745874949 Chr.19: 8366156-8366156 on Build GRCh38
rs762399326 Chr.19: 8366211-8366211 on Build GRCh38
rs181710838 Chr.19: 8366075-8366075 on Build GRCh38
rs894977200 Chr.19: 8366160-8366160 on Build GRCh38
rs776816866 Chr.19: 8366206-8366206 on Build GRCh38
rs547981454 Chr.19: 8366247-8366247 on Build GRCh38
rs368039480 Chr.19: 8366162-8366162 on Build GRCh38
rs562366524 Chr.19: 8366120-8366120 on Build GRCh38
rs766070708 Chr.19: 8366238-8366238 on Build GRCh38
rs764754051 Chr.19: 8366076-8366076 on Build GRCh38
rs200197036 Chr.19: 8366197-8366197 on Build GRCh38
rs780461101 Chr.19: 8366254-8366254 on Build GRCh38
rs926515575 Chr.19: 8366168-8366168 on Build GRCh38
rs929954989 Chr.19: 8366107-8366107 on Build GRCh38
rs1012120885 Chr.19: 8366182-8366182 on Build GRCh38

View Full Product Details