Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 23633863-23634090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CST3
Entrez Gene ID
1471
Gene Name
cystatin C
Gene Aliases
ARMD11, HEL-S-2
UniGene
Hs.304682
Gene Chromosome Location
Chr.20: 23627897-23638048 on Build GRCh38
Transcripts
NM_000099.3 NM_001288614.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7268685 c.361G>T p.A121S Chr.20: 23633996-23633996 on Build GRCh38
5119007 c.381C>A p.I127I Chr.20: 23633976-23633976 on Build GRCh38
4843537 c.362C>A p.A121E Chr.20: 23633995-23633995 on Build GRCh38
7131404 c.375C>T p.F125F Chr.20: 23633982-23633982 on Build GRCh38
1025235 c.399G>T p.Q133H Chr.20: 23633958-23633958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879219458 Chr.20: 23634059-23634059 on Build GRCh38
rs747240679 Chr.20: 23634023-23634023 on Build GRCh38
rs752255286 Chr.20: 23633889-23633889 on Build GRCh38
rs137970982 Chr.20: 23633973-23633973 on Build GRCh38
rs765197140 Chr.20: 23633948-23633948 on Build GRCh38
rs149051742 Chr.20: 23633989-23633989 on Build GRCh38
rs1135147 Chr.20: 23633976-23633976 on Build GRCh38
rs371538575 Chr.20: 23633905-23633905 on Build GRCh38
rs779110225 Chr.20: 23633915-23633915 on Build GRCh38
rs759445850 Chr.20: 23633941-23633941 on Build GRCh38
rs755513850 Chr.20: 23633995-23633995 on Build GRCh38
rs748626736 Chr.20: 23634015-23634015 on Build GRCh38
rs770237341 Chr.20: 23633913-23633913 on Build GRCh38
rs753716901 Chr.20: 23633884-23633884 on Build GRCh38
rs764714963 Chr.20: 23633888-23633888 on Build GRCh38
rs894350263 Chr.20: 23633977-23633977 on Build GRCh38
rs748275854 Chr.20: 23633921-23633921 on Build GRCh38
rs770693621 Chr.20: 23633935-23633935 on Build GRCh38
rs933693136 Chr.20: 23633902-23633902 on Build GRCh38
rs755596435 Chr.20: 23633913-23633913 on Build GRCh38
rs773433814 Chr.20: 23633931-23633931 on Build GRCh38
rs201422446 Chr.20: 23633922-23633922 on Build GRCh38
rs199895375 Chr.20: 23634040-23634040 on Build GRCh38
rs141643699 Chr.20: 23633933-23633933 on Build GRCh38
rs931941687 Chr.20: 23634046-23634046 on Build GRCh38
rs745620150 Chr.20: 23634042-23634042 on Build GRCh38
rs758446804 Chr.20: 23634017-23634017 on Build GRCh38
rs777687786 Chr.20: 23633914-23633914 on Build GRCh38
rs763523220 Chr.20: 23633960-23633960 on Build GRCh38
rs77000936 Chr.20: 23633940-23633940 on Build GRCh38
rs763764400 Chr.20: 23633903-23633903 on Build GRCh38
rs769597486 Chr.20: 23634048-23634048 on Build GRCh38
rs779524029 Chr.20: 23634008-23634008 on Build GRCh38
rs777720033 Chr.20: 23634019-23634019 on Build GRCh38
rs754306266 Chr.20: 23633986-23633986 on Build GRCh38
rs762473017 Chr.20: 23633957-23633957 on Build GRCh38
rs770663365 Chr.20: 23633951-23633951 on Build GRCh38
rs200979591 Chr.20: 23633904-23633904 on Build GRCh38
rs374979316 Chr.20: 23634038-23634038 on Build GRCh38
rs75719749 Chr.20: 23634053-23634053 on Build GRCh38
rs145812457 Chr.20: 23633994-23633994 on Build GRCh38
rs371605207 Chr.20: 23633972-23633972 on Build GRCh38
rs878941112 Chr.20: 23634002-23634002 on Build GRCh38
rs766840907 Chr.20: 23633979-23633979 on Build GRCh38
rs757959037 Chr.20: 23633895-23633895 on Build GRCh38
rs773943139 Chr.20: 23634018-23634018 on Build GRCh38
rs749804609 Chr.20: 23633906-23633906 on Build GRCh38
rs940780639 Chr.20: 23634065-23634065 on Build GRCh38

View Full Product Details