Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67645180-67645433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACY3
Entrez Gene ID
91703
Gene Name
aminoacylase 3
Gene Aliases
ACY-3, HCBP1
UniGene
Hs.126265
Gene Chromosome Location
Chr.11: 67642551-67650829 on Build GRCh38
Transcripts
NM_080658.1 XM_017018549.1 XM_017018550.1 XM_017018551.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5999559 c.526+7C>T p.? Chr.11: 67645280-67645280 on Build GRCh38
4146212 c.471C>T p.Y157Y Chr.11: 67645342-67645342 on Build GRCh38
7225891 c.526G>A p.G176S Chr.11: 67645287-67645287 on Build GRCh38
5783849 c.479C>G p.S160C Chr.11: 67645334-67645334 on Build GRCh38
1356465 c.456C>A p.C152* Chr.11: 67645357-67645357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs994633959 Chr.11: 67645238-67645238 on Build GRCh38
rs981585327 Chr.11: 67645405-67645405 on Build GRCh38
rs767566095 Chr.11: 67645347-67645347 on Build GRCh38
rs780702044 Chr.11: 67645236-67645236 on Build GRCh38
rs892994345 Chr.11: 67645408-67645408 on Build GRCh38
rs139407748 Chr.11: 67645306-67645306 on Build GRCh38
rs778527598 Chr.11: 67645256-67645256 on Build GRCh38
rs747600781 Chr.11: 67645322-67645322 on Build GRCh38
rs771569983 Chr.11: 67645259-67645259 on Build GRCh38
rs746720770 Chr.11: 67645338-67645338 on Build GRCh38
rs528679369 Chr.11: 67645282-67645282 on Build GRCh38
rs771683337 Chr.11: 67645337-67645337 on Build GRCh38
rs76819752 Chr.11: 67645368-67645368 on Build GRCh38
rs756412896 Chr.11: 67645304-67645304 on Build GRCh38
rs17148450 Chr.11: 67645300-67645300 on Build GRCh38
rs759417260 Chr.11: 67645276-67645276 on Build GRCh38
rs542851264 Chr.11: 67645378-67645378 on Build GRCh38
rs377442739 Chr.11: 67645309-67645309 on Build GRCh38
rs765186959 Chr.11: 67645279-67645279 on Build GRCh38
rs201907157 Chr.11: 67645258-67645258 on Build GRCh38
rs1024821134 Chr.11: 67645212-67645212 on Build GRCh38
rs539422088 Chr.11: 67645369-67645369 on Build GRCh38
rs567885766 Chr.11: 67645264-67645264 on Build GRCh38
rs1029529054 Chr.11: 67645239-67645239 on Build GRCh38
rs779579306 Chr.11: 67645312-67645312 on Build GRCh38
rs972047768 Chr.11: 67645250-67645250 on Build GRCh38
rs909162479 Chr.11: 67645235-67645235 on Build GRCh38
rs766714932 Chr.11: 67645380-67645380 on Build GRCh38
rs969655566 Chr.11: 67645217-67645217 on Build GRCh38
rs751570351 Chr.11: 67645280-67645280 on Build GRCh38
rs757820368 Chr.11: 67645403-67645403 on Build GRCh38
rs980613386 Chr.11: 67645219-67645219 on Build GRCh38
rs750723413 Chr.11: 67645303-67645303 on Build GRCh38
rs957241929 Chr.11: 67645301-67645301 on Build GRCh38
rs770588590 Chr.11: 67645346-67645346 on Build GRCh38
rs370834010 Chr.11: 67645358-67645358 on Build GRCh38
rs756559492 Chr.11: 67645204-67645204 on Build GRCh38
rs772979621 Chr.11: 67645262-67645262 on Build GRCh38
rs750693428 Chr.11: 67645201-67645201 on Build GRCh38
rs536780241 Chr.11: 67645389-67645389 on Build GRCh38
rs371104378 Chr.11: 67645255-67645255 on Build GRCh38
rs749859855 Chr.11: 67645248-67645248 on Build GRCh38
rs753291142 Chr.11: 67645401-67645401 on Build GRCh38
rs372847889 Chr.11: 67645387-67645387 on Build GRCh38
rs776368388 Chr.11: 67645355-67645355 on Build GRCh38
rs933428670 Chr.11: 67645294-67645295 on Build GRCh38
rs1043221915 Chr.11: 67645330-67645330 on Build GRCh38
rs770796193 Chr.11: 67645263-67645263 on Build GRCh38
rs755453889 Chr.11: 67645388-67645388 on Build GRCh38
rs750492034 Chr.11: 67645376-67645376 on Build GRCh38

View Full Product Details