Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150720020-150720277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIMAP1
Entrez Gene ID
170575
Gene Name
GTPase, IMAP family member 1
Gene Aliases
HIMAP1, IAN2, IMAP1, IMAP38
UniGene
Hs.647079
Gene Chromosome Location
Chr.7: 150716557-150724280 on Build GRCh38
Transcripts
NM_130759.3 NM_001199577.1 NM_001303630.1

Gene Symbol
GIMAP1-GIMAP5
Entrez Gene ID
100527949
Gene Name
GIMAP1-GIMAP5 readthrough
Gene Aliases
-
UniGene
Hs.647079
Gene Chromosome Location
Chr.7: 150716557-150743649 on Build GRCh38
Transcripts
NM_130759.3 NM_001199577.1 NM_001303630.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
225616 c.152G>A p.R51K Chr.7: 150720156-150720156 on Build GRCh38
5455993 c.123C>T p.S41S Chr.7: 150720127-150720127 on Build GRCh38
232403 c.235G>A p.E79K Chr.7: 150720239-150720239 on Build GRCh38
5630420 c.96T>A p.L32L Chr.7: 150720100-150720100 on Build GRCh38
746009 c.83G>A p.R28Q Chr.7: 150720087-150720087 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142046758 Chr.7: 150720195-150720195 on Build GRCh38
rs772350694 Chr.7: 150720079-150720079 on Build GRCh38
rs866130773 Chr.7: 150720080-150720080 on Build GRCh38
rs145107401 Chr.7: 150720085-150720085 on Build GRCh38
rs373301494 Chr.7: 150720136-150720136 on Build GRCh38
rs867001533 Chr.7: 150720087-150720087 on Build GRCh38
rs779713323 Chr.7: 150720135-150720135 on Build GRCh38
rs144313030 Chr.7: 150720139-150720139 on Build GRCh38
rs964759206 Chr.7: 150720204-150720204 on Build GRCh38
rs774171968 Chr.7: 150720186-150720186 on Build GRCh38
rs987918385 Chr.7: 150720062-150720062 on Build GRCh38
rs756928324 Chr.7: 150720209-150720210 on Build GRCh38
rs751170712 Chr.7: 150720056-150720056 on Build GRCh38
rs756859671 Chr.7: 150720126-150720126 on Build GRCh38
rs754787299 Chr.7: 150720256-150720256 on Build GRCh38
rs867616370 Chr.7: 150720179-150720179 on Build GRCh38
rs527414207 Chr.7: 150720231-150720231 on Build GRCh38
rs781321493 Chr.7: 150720060-150720060 on Build GRCh38
rs551296159 Chr.7: 150720072-150720072 on Build GRCh38
rs777141777 Chr.7: 150720086-150720086 on Build GRCh38
rs769499729 Chr.7: 150720063-150720063 on Build GRCh38
rs751279373 Chr.7: 150720148-150720148 on Build GRCh38
rs149617630 Chr.7: 150720057-150720057 on Build GRCh38
rs937763624 Chr.7: 150720254-150720254 on Build GRCh38
rs757857616 Chr.7: 150720051-150720051 on Build GRCh38
rs200708956 Chr.7: 150720183-150720183 on Build GRCh38
rs946354751 Chr.7: 150720208-150720208 on Build GRCh38
rs1028389231 Chr.7: 150720117-150720117 on Build GRCh38
rs765654115 Chr.7: 150720073-150720073 on Build GRCh38
rs771007789 Chr.7: 150720151-150720151 on Build GRCh38
rs61749583 Chr.7: 150720100-150720100 on Build GRCh38
rs144542417 Chr.7: 150720061-150720061 on Build GRCh38
rs751227415 Chr.7: 150720208-150720209 on Build GRCh38
rs746356038 Chr.7: 150720154-150720154 on Build GRCh38
rs377535856 Chr.7: 150720211-150720211 on Build GRCh38
rs199704494 Chr.7: 150720250-150720250 on Build GRCh38
rs75609333 Chr.7: 150720084-150720084 on Build GRCh38
rs755874999 Chr.7: 150720132-150720132 on Build GRCh38
rs1043834348 Chr.7: 150720077-150720077 on Build GRCh38
rs988076574 Chr.7: 150720191-150720191 on Build GRCh38
rs201408566 Chr.7: 150720053-150720053 on Build GRCh38
rs763886483 Chr.7: 150720114-150720114 on Build GRCh38
rs1029606178 Chr.7: 150720235-150720235 on Build GRCh38
rs751080744 Chr.7: 150720118-150720118 on Build GRCh38
rs759883669 Chr.7: 150720091-150720091 on Build GRCh38
rs765521233 Chr.7: 150720099-150720099 on Build GRCh38
rs750341190 Chr.7: 150720128-150720128 on Build GRCh38
rs770389381 Chr.7: 150720157-150720157 on Build GRCh38
rs201983958 Chr.7: 150720245-150720245 on Build GRCh38
rs151144214 Chr.7: 150720222-150720222 on Build GRCh38
rs779139111 Chr.7: 150720071-150720071 on Build GRCh38
rs995495314 Chr.7: 150720103-150720103 on Build GRCh38
rs767014155 Chr.7: 150720214-150720214 on Build GRCh38
rs755995652 Chr.7: 150720232-150720232 on Build GRCh38
rs562054826 Chr.7: 150720127-150720127 on Build GRCh38
rs763450656 Chr.7: 150720178-150720178 on Build GRCh38
rs775693608 Chr.7: 150720158-150720158 on Build GRCh38

View Full Product Details