Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 248592952-248593223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR2T10
Entrez Gene ID
127069
Gene Name
olfactory receptor family 2 subfamily T member 10
Gene Aliases
OR1-64
UniGene
Hs.553585
Gene Chromosome Location
Chr.1: 248483404-248645469 on Build GRCh38
Transcripts
NM_001004693.1 XM_017000273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
302808 c.562T>C p.S188P Chr.1: 248593207-248593207 on Build GRCh38
6509061 c.558G>T p.K186N Chr.1: 248593211-248593211 on Build GRCh38
907361 c.605G>A p.C202Y Chr.1: 248593164-248593164 on Build GRCh38
907360 c.682A>G p.M228V Chr.1: 248593087-248593087 on Build GRCh38
907359 c.816G>T p.M272I Chr.1: 248592953-248592953 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866048007 Chr.1: 248593012-248593012 on Build GRCh38
rs758843051 Chr.1: 248593013-248593013 on Build GRCh38
rs745595536 Chr.1: 248593068-248593068 on Build GRCh38
rs757801389 Chr.1: 248592991-248592991 on Build GRCh38
rs763197158 Chr.1: 248593049-248593051 on Build GRCh38
rs752537672 Chr.1: 248593152-248593154 on Build GRCh38
rs753441046 Chr.1: 248592989-248592989 on Build GRCh38
rs748166599 Chr.1: 248593141-248593141 on Build GRCh38
rs560031490 Chr.1: 248593140-248593140 on Build GRCh38
rs774284253 Chr.1: 248593050-248593050 on Build GRCh38
rs755113821 Chr.1: 248593134-248593134 on Build GRCh38
rs756066204 Chr.1: 248593159-248593159 on Build GRCh38
rs369930196 Chr.1: 248593092-248593092 on Build GRCh38
rs775110216 Chr.1: 248593168-248593168 on Build GRCh38
rs781099217 Chr.1: 248593039-248593039 on Build GRCh38
rs760215769 Chr.1: 248593084-248593084 on Build GRCh38
rs767271900 Chr.1: 248593075-248593076 on Build GRCh38
rs201402953 Chr.1: 248593077-248593077 on Build GRCh38
rs755274322 Chr.1: 248593035-248593035 on Build GRCh38
rs369070111 Chr.1: 248592980-248592980 on Build GRCh38
rs561086727 Chr.1: 248593014-248593014 on Build GRCh38
rs370684993 Chr.1: 248593194-248593194 on Build GRCh38
rs777888163 Chr.1: 248593159-248593159 on Build GRCh38
rs143228008 Chr.1: 248593004-248593004 on Build GRCh38
rs201461753 Chr.1: 248593044-248593044 on Build GRCh38
rs201268729 Chr.1: 248593169-248593169 on Build GRCh38
rs28733753 Chr.1: 248593110-248593110 on Build GRCh38
rs765870174 Chr.1: 248592998-248592998 on Build GRCh38
rs41269353 Chr.1: 248593201-248593201 on Build GRCh38
rs576268124 Chr.1: 248593024-248593024 on Build GRCh38
rs149210040 Chr.1: 248593156-248593156 on Build GRCh38
rs776431345 Chr.1: 248593175-248593175 on Build GRCh38
rs765776577 Chr.1: 248593120-248593120 on Build GRCh38
rs750596936 Chr.1: 248592989-248592989 on Build GRCh38
rs758998533 Chr.1: 248593132-248593132 on Build GRCh38
rs140690496 Chr.1: 248593049-248593049 on Build GRCh38
rs771751855 Chr.1: 248593069-248593069 on Build GRCh38
rs61997185 Chr.1: 248593133-248593133 on Build GRCh38
rs761629982 Chr.1: 248593176-248593176 on Build GRCh38
rs763490781 Chr.1: 248592988-248592988 on Build GRCh38
rs745783658 Chr.1: 248593165-248593165 on Build GRCh38
rs747181762 Chr.1: 248593022-248593022 on Build GRCh38
rs753722914 Chr.1: 248593182-248593182 on Build GRCh38
rs28664620 Chr.1: 248593173-248593173 on Build GRCh38
rs750957100 Chr.1: 248593130-248593130 on Build GRCh38
rs772087986 Chr.1: 248593166-248593166 on Build GRCh38
rs267598512 Chr.1: 248593011-248593011 on Build GRCh38

View Full Product Details