Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 63593332-63593573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGM1
Entrez Gene ID
5236
Gene Name
phosphoglucomutase 1
Gene Aliases
CDG1T, GSD14
UniGene
Hs.1869
Gene Chromosome Location
Chr.1: 63593276-63660245 on Build GRCh38
Transcripts
NM_002633.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5491131 c.39G>T p.Q13H Chr.1: 63593527-63593527 on Build GRCh38
3491685 c.65T>C p.L22P Chr.1: 63593553-63593553 on Build GRCh38
4563997 c.9G>A p.K3K Chr.1: 63593497-63593497 on Build GRCh38
2238733 c.63G>A p.G21G Chr.1: 63593551-63593551 on Build GRCh38
7169493 c.68G>A p.R23Q Chr.1: 63593556-63593556 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973751597 Chr.1: 63593385-63593386 on Build GRCh38
rs752536129 Chr.1: 63593529-63593529 on Build GRCh38
rs371275043 Chr.1: 63593526-63593526 on Build GRCh38
rs920499043 Chr.1: 63593400-63593400 on Build GRCh38
rs538679699 Chr.1: 63593415-63593415 on Build GRCh38
rs112550974 Chr.1: 63593379-63593379 on Build GRCh38
rs774343488 Chr.1: 63593447-63593447 on Build GRCh38
rs773189460 Chr.1: 63593516-63593516 on Build GRCh38
rs758344892 Chr.1: 63593465-63593465 on Build GRCh38
rs368108235 Chr.1: 63593477-63593477 on Build GRCh38
rs767556750 Chr.1: 63593539-63593539 on Build GRCh38
rs756875667 Chr.1: 63593472-63593472 on Build GRCh38
rs368246894 Chr.1: 63593410-63593410 on Build GRCh38
rs551310804 Chr.1: 63593440-63593440 on Build GRCh38
rs111246749 Chr.1: 63593506-63593506 on Build GRCh38
rs764594914 Chr.1: 63593445-63593445 on Build GRCh38
rs938360358 Chr.1: 63593409-63593409 on Build GRCh38
rs912988819 Chr.1: 63593412-63593412 on Build GRCh38
rs374061676 Chr.1: 63593453-63593453 on Build GRCh38
rs775746481 Chr.1: 63593441-63593441 on Build GRCh38
rs752554415 Chr.1: 63593461-63593461 on Build GRCh38
rs370035290 Chr.1: 63593450-63593450 on Build GRCh38
rs935255839 Chr.1: 63593391-63593391 on Build GRCh38
rs900050010 Chr.1: 63593452-63593452 on Build GRCh38
rs1041813760 Chr.1: 63593422-63593422 on Build GRCh38
rs775862619 Chr.1: 63593500-63593500 on Build GRCh38
rs888567436 Chr.1: 63593468-63593468 on Build GRCh38
rs578037734 Chr.1: 63593539-63593539 on Build GRCh38
rs969979081 Chr.1: 63593354-63593354 on Build GRCh38
rs756324880 Chr.1: 63593454-63593454 on Build GRCh38
rs374891070 Chr.1: 63593462-63593462 on Build GRCh38
rs746194486 Chr.1: 63593439-63593439 on Build GRCh38
rs890027917 Chr.1: 63593430-63593430 on Build GRCh38
rs759220275 Chr.1: 63593513-63593522 on Build GRCh38
rs947534673 Chr.1: 63593413-63593413 on Build GRCh38
rs749653003 Chr.1: 63593505-63593505 on Build GRCh38
rs547294819 Chr.1: 63593436-63593436 on Build GRCh38
rs937170258 Chr.1: 63593402-63593402 on Build GRCh38
rs764100819 Chr.1: 63593545-63593545 on Build GRCh38
rs1015038005 Chr.1: 63593376-63593376 on Build GRCh38
rs925255178 Chr.1: 63593385-63593385 on Build GRCh38
rs961723819 Chr.1: 63593381-63593381 on Build GRCh38
rs531186191 Chr.1: 63593353-63593353 on Build GRCh38
rs763399590 Chr.1: 63593444-63593444 on Build GRCh38
rs991285512 Chr.1: 63593405-63593406 on Build GRCh38
rs980159187 Chr.1: 63593368-63593368 on Build GRCh38
rs201805940 Chr.1: 63593448-63593448 on Build GRCh38
rs760894601 Chr.1: 63593519-63593519 on Build GRCh38
rs765034541 Chr.1: 63593456-63593456 on Build GRCh38
rs928299083 Chr.1: 63593387-63593387 on Build GRCh38
rs767597430 Chr.1: 63593514-63593514 on Build GRCh38
rs751586075 Chr.1: 63593554-63593554 on Build GRCh38
rs1007164791 Chr.1: 63593438-63593438 on Build GRCh38
rs200633484 Chr.1: 63593490-63593490 on Build GRCh38
rs374954710 Chr.1: 63593510-63593510 on Build GRCh38
rs549505712 Chr.1: 63593395-63593395 on Build GRCh38
rs780676049 Chr.1: 63593474-63593474 on Build GRCh38
rs556559217 Chr.1: 63593509-63593509 on Build GRCh38

View Full Product Details