Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 43576560-43576813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO10
Entrez Gene ID
55129
Gene Name
anoctamin 10
Gene Aliases
SCAR10, TMEM16K
UniGene
Hs.656657
Gene Chromosome Location
Chr.3: 43366324-43691594 on Build GRCh38
Transcripts
NM_001204831.1 NM_001204832.1 NM_001204833.1 NM_001204834.1 NM_018075.3 XM_011533885.2 XM_011533887.2 XM_011533889.2 XM_011533890.2 XM_017006716.1 XM_017006717.1 XM_017006718.1 XM_017006719.1 XM_017006720.1 XM_017006721.1 XM_017006722.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6008866 c.1104_1105delTG p.V369fs*2 Chr.3: 43576749-43576750 on Build GRCh38
3365183 c.1090A>G p.I364V Chr.3: 43576764-43576764 on Build GRCh38
6601003 c.1132C>T p.R378* Chr.3: 43576722-43576722 on Build GRCh38
7284948 c.1051G>A p.G351R Chr.3: 43576803-43576803 on Build GRCh38
69667 c.1156T>G p.S386A Chr.3: 43576698-43576698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs193181511 Chr.3: 43576667-43576667 on Build GRCh38
rs745434303 Chr.3: 43576677-43576677 on Build GRCh38
rs763801421 Chr.3: 43576638-43576638 on Build GRCh38
rs537457363 Chr.3: 43576731-43576731 on Build GRCh38
rs370018426 Chr.3: 43576770-43576770 on Build GRCh38
rs794726680 Chr.3: 43576703-43576704 on Build GRCh38
rs760973973 Chr.3: 43576755-43576755 on Build GRCh38
rs775049285 Chr.3: 43576685-43576685 on Build GRCh38
rs890815374 Chr.3: 43576708-43576708 on Build GRCh38
rs746710432 Chr.3: 43576688-43576688 on Build GRCh38
rs755048076 Chr.3: 43576645-43576645 on Build GRCh38
rs373948580 Chr.3: 43576773-43576773 on Build GRCh38
rs1054611332 Chr.3: 43576621-43576621 on Build GRCh38
rs1055034872 Chr.3: 43576733-43576733 on Build GRCh38
rs779618110 Chr.3: 43576787-43576787 on Build GRCh38
rs144272231 Chr.3: 43576710-43576710 on Build GRCh38
rs773121159 Chr.3: 43576722-43576722 on Build GRCh38
rs767603679 Chr.3: 43576743-43576743 on Build GRCh38
rs768148483 Chr.3: 43576691-43576691 on Build GRCh38
rs187964933 Chr.3: 43576666-43576666 on Build GRCh38
rs61732728 Chr.3: 43576721-43576721 on Build GRCh38
rs936887940 Chr.3: 43576604-43576604 on Build GRCh38
rs551675434 Chr.3: 43576680-43576680 on Build GRCh38
rs371487223 Chr.3: 43576668-43576668 on Build GRCh38
rs778230480 Chr.3: 43576660-43576660 on Build GRCh38
rs926487903 Chr.3: 43576757-43576757 on Build GRCh38
rs752941925 Chr.3: 43576653-43576653 on Build GRCh38
rs752679400 Chr.3: 43576751-43576751 on Build GRCh38
rs761532493 Chr.3: 43576719-43576719 on Build GRCh38
rs757689043 Chr.3: 43576775-43576775 on Build GRCh38
rs56389778 Chr.3: 43576788-43576788 on Build GRCh38
rs372441963 Chr.3: 43576656-43576656 on Build GRCh38

View Full Product Details