Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73619142-73619367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT6
Entrez Gene ID
641372
Gene Name
acyl-CoA thioesterase 6
Gene Aliases
C14orf42, c14_5530
UniGene
Hs.122038
Gene Chromosome Location
Chr.14: 73612276-73619929 on Build GRCh38
Transcripts
NM_001037162.1 XM_011537076.2 XM_017021289.1

Gene Symbol
HEATR4
Entrez Gene ID
399671
Gene Name
HEAT repeat containing 4
Gene Aliases
-
UniGene
Hs.660774
Gene Chromosome Location
Chr.14: 73478484-73634418 on Build GRCh38
Transcripts
NM_001037162.1 XM_011537076.2 XM_017021289.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6592294 c.86C>A p.S29Y Chr.14: 73619301-73619301 on Build GRCh38
6554515 c.97G>T p.G33C Chr.14: 73619312-73619312 on Build GRCh38
5436365 c.53C>G p.S18C Chr.14: 73619268-73619268 on Build GRCh38
957663 c.64G>C p.D22H Chr.14: 73619279-73619279 on Build GRCh38
2137265 c.73C>T p.L25F Chr.14: 73619288-73619288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139773974 Chr.14: 73619178-73619178 on Build GRCh38
rs780570957 Chr.14: 73619282-73619282 on Build GRCh38
rs762916051 Chr.14: 73619326-73619326 on Build GRCh38
rs781662746 Chr.14: 73619275-73619275 on Build GRCh38
rs199762917 Chr.14: 73619235-73619235 on Build GRCh38
rs771534894 Chr.14: 73619193-73619193 on Build GRCh38
rs770478789 Chr.14: 73619311-73619311 on Build GRCh38
rs994917842 Chr.14: 73619248-73619248 on Build GRCh38
rs775622053 Chr.14: 73619326-73619327 on Build GRCh38
rs781341647 Chr.14: 73619184-73619184 on Build GRCh38
rs760188991 Chr.14: 73619199-73619199 on Build GRCh38
rs1035362048 Chr.14: 73619317-73619317 on Build GRCh38
rs150641071 Chr.14: 73619295-73619295 on Build GRCh38
rs142921619 Chr.14: 73619253-73619253 on Build GRCh38
rs749246127 Chr.14: 73619329-73619329 on Build GRCh38
rs764299779 Chr.14: 73619337-73619337 on Build GRCh38
rs924871186 Chr.14: 73619333-73619333 on Build GRCh38
rs190021634 Chr.14: 73619181-73619181 on Build GRCh38
rs74715947 Chr.14: 73619316-73619316 on Build GRCh38
rs566353450 Chr.14: 73619241-73619241 on Build GRCh38
rs1012322676 Chr.14: 73619334-73619334 on Build GRCh38
rs376187544 Chr.14: 73619324-73619324 on Build GRCh38
rs773902478 Chr.14: 73619190-73619190 on Build GRCh38
rs749746870 Chr.14: 73619290-73619290 on Build GRCh38
rs777656695 Chr.14: 73619252-73619252 on Build GRCh38
rs374698220 Chr.14: 73619214-73619214 on Build GRCh38
rs759432051 Chr.14: 73619318-73619318 on Build GRCh38
rs369953292 Chr.14: 73619194-73619194 on Build GRCh38
rs200727737 Chr.14: 73619323-73619323 on Build GRCh38
rs771629818 Chr.14: 73619294-73619294 on Build GRCh38
rs541857806 Chr.14: 73619246-73619246 on Build GRCh38
rs766201769 Chr.14: 73619211-73619211 on Build GRCh38
rs924780872 Chr.14: 73619175-73619175 on Build GRCh38
rs746745416 Chr.14: 73619308-73619308 on Build GRCh38
rs765233466 Chr.14: 73619236-73619236 on Build GRCh38
rs201661122 Chr.14: 73619277-73619277 on Build GRCh38
rs529773787 Chr.14: 73619189-73619189 on Build GRCh38
rs753573150 Chr.14: 73619233-73619233 on Build GRCh38
rs757414993 Chr.14: 73619254-73619254 on Build GRCh38
rs772286233 Chr.14: 73619293-73619293 on Build GRCh38
rs761285209 Chr.14: 73619192-73619192 on Build GRCh38

View Full Product Details