Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4370390-4370615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF23
Entrez Gene ID
8074
Gene Name
fibroblast growth factor 23
Gene Aliases
ADHR, FGFN, HPDR2, HYPF, PHPTC
UniGene
Hs.287370
Gene Chromosome Location
Chr.12: 4368227-4379728 on Build GRCh38
Transcripts
NM_020638.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
230244 c.610G>A p.A204T Chr.12: 4370489-4370489 on Build GRCh38
7148573 c.537G>T p.R179R Chr.12: 4370562-4370562 on Build GRCh38
5375162 c.486C>T p.N162N Chr.12: 4370613-4370613 on Build GRCh38
244514 c.653C>T p.P218L Chr.12: 4370446-4370446 on Build GRCh38
7368201 c.608C>A p.P203Q Chr.12: 4370491-4370491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772772136 Chr.12: 4370555-4370555 on Build GRCh38
rs756683570 Chr.12: 4370460-4370460 on Build GRCh38
rs758018599 Chr.12: 4370433-4370433 on Build GRCh38
rs749949273 Chr.12: 4370496-4370496 on Build GRCh38
rs149058834 Chr.12: 4370570-4370570 on Build GRCh38
rs750507167 Chr.12: 4370591-4370591 on Build GRCh38
rs28937882 Chr.12: 4370564-4370564 on Build GRCh38
rs963172843 Chr.12: 4370442-4370442 on Build GRCh38
rs754201217 Chr.12: 4370573-4370573 on Build GRCh38
rs762634143 Chr.12: 4370497-4370497 on Build GRCh38
rs764669949 Chr.12: 4370424-4370424 on Build GRCh38
rs749770315 Chr.12: 4370463-4370463 on Build GRCh38
rs768568647 Chr.12: 4370492-4370492 on Build GRCh38
rs753345321 Chr.12: 4370411-4370411 on Build GRCh38
rs374830674 Chr.12: 4370487-4370487 on Build GRCh38
rs746049992 Chr.12: 4370467-4370467 on Build GRCh38
rs1008848282 Chr.12: 4370577-4370577 on Build GRCh38
rs934951717 Chr.12: 4370512-4370512 on Build GRCh38
rs533026808 Chr.12: 4370494-4370494 on Build GRCh38
rs371244880 Chr.12: 4370462-4370462 on Build GRCh38
rs754327706 Chr.12: 4370539-4370539 on Build GRCh38
rs753097960 Chr.12: 4370507-4370507 on Build GRCh38
rs886042161 Chr.12: 4370526-4370526 on Build GRCh38
rs778886291 Chr.12: 4370466-4370466 on Build GRCh38
rs764061118 Chr.12: 4370408-4370408 on Build GRCh38
rs115283398 Chr.12: 4370544-4370544 on Build GRCh38
rs769657664 Chr.12: 4370552-4370552 on Build GRCh38
rs199530638 Chr.12: 4370501-4370501 on Build GRCh38
rs193922702 Chr.12: 4370563-4370563 on Build GRCh38
rs987715580 Chr.12: 4370475-4370475 on Build GRCh38
rs868121222 Chr.12: 4370491-4370491 on Build GRCh38
rs766201128 Chr.12: 4370500-4370500 on Build GRCh38
rs768373820 Chr.12: 4370549-4370549 on Build GRCh38
rs143050343 Chr.12: 4370586-4370586 on Build GRCh38
rs369808475 Chr.12: 4370550-4370550 on Build GRCh38
rs144925325 Chr.12: 4370548-4370548 on Build GRCh38
rs573322878 Chr.12: 4370584-4370584 on Build GRCh38
rs104894347 Chr.12: 4370572-4370572 on Build GRCh38
rs775236283 Chr.12: 4370472-4370472 on Build GRCh38
rs369035509 Chr.12: 4370445-4370445 on Build GRCh38
rs756850883 Chr.12: 4370423-4370423 on Build GRCh38
rs759288956 Chr.12: 4370568-4370568 on Build GRCh38
rs1033823201 Chr.12: 4370413-4370413 on Build GRCh38
rs772236398 Chr.12: 4370471-4370471 on Build GRCh38
rs866041340 Chr.12: 4370489-4370489 on Build GRCh38
rs950401479 Chr.12: 4370473-4370473 on Build GRCh38
rs376521670 Chr.12: 4370407-4370407 on Build GRCh38
rs767171941 Chr.12: 4370459-4370459 on Build GRCh38
rs387907425 Chr.12: 4370476-4370476 on Build GRCh38
rs780315628 Chr.12: 4370545-4370545 on Build GRCh38
rs770974049 Chr.12: 4370464-4370464 on Build GRCh38
rs190841442 Chr.12: 4370540-4370540 on Build GRCh38
rs13312793 Chr.12: 4370516-4370516 on Build GRCh38
rs750025882 Chr.12: 4370426-4370426 on Build GRCh38
rs764766537 Chr.12: 4370495-4370495 on Build GRCh38

View Full Product Details