Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127825188-127825461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENG
Entrez Gene ID
2022
Gene Name
endoglin
Gene Aliases
END, HHT1, ORW1
UniGene
Hs.76753
Gene Chromosome Location
Chr.9: 127815012-127854773 on Build GRCh38
Transcripts
NM_000118.3 NM_001114753.2 NM_001278138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3654517 c.764G>A p.G255D Chr.9: 127825283-127825283 on Build GRCh38
117771 c.752T>C p.L251P Chr.9: 127825295-127825295 on Build GRCh38
244254 c.733G>A p.G245R Chr.9: 127825314-127825314 on Build GRCh38
6437208 c.747C>G p.A249A Chr.9: 127825300-127825300 on Build GRCh38
6437209 c.747C>G p.A249A Chr.9: 127825300-127825300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916649472 Chr.9: 127825437-127825437 on Build GRCh38
rs748849680 Chr.9: 127825365-127825365 on Build GRCh38
rs762621671 Chr.9: 127825361-127825361 on Build GRCh38
rs775160533 Chr.9: 127825305-127825305 on Build GRCh38
rs778084631 Chr.9: 127825392-127825392 on Build GRCh38
rs765377503 Chr.9: 127825332-127825333 on Build GRCh38
rs769339049 Chr.9: 127825356-127825356 on Build GRCh38
rs759102482 Chr.9: 127825299-127825299 on Build GRCh38
rs758252623 Chr.9: 127825254-127825254 on Build GRCh38
rs765936911 Chr.9: 127825223-127825223 on Build GRCh38
rs201457707 Chr.9: 127825306-127825306 on Build GRCh38
rs373475136 Chr.9: 127825405-127825405 on Build GRCh38
rs537154767 Chr.9: 127825273-127825273 on Build GRCh38
rs756252418 Chr.9: 127825277-127825277 on Build GRCh38
rs886041548 Chr.9: 127825335-127825335 on Build GRCh38
rs757090839 Chr.9: 127825272-127825272 on Build GRCh38
rs139767471 Chr.9: 127825349-127825349 on Build GRCh38
rs111471193 Chr.9: 127825230-127825230 on Build GRCh38
rs143779963 Chr.9: 127825281-127825281 on Build GRCh38
rs766598911 Chr.9: 127825380-127825380 on Build GRCh38
rs759191907 Chr.9: 127825225-127825225 on Build GRCh38
rs750477084 Chr.9: 127825275-127825275 on Build GRCh38
rs41446048 Chr.9: 127825422-127825422 on Build GRCh38
rs1046451583 Chr.9: 127825436-127825436 on Build GRCh38
rs374628465 Chr.9: 127825362-127825362 on Build GRCh38
rs774964688 Chr.9: 127825217-127825217 on Build GRCh38
rs545465750 Chr.9: 127825321-127825321 on Build GRCh38
rs369047677 Chr.9: 127825416-127825416 on Build GRCh38
rs377377549 Chr.9: 127825293-127825293 on Build GRCh38
rs776218326 Chr.9: 127825296-127825296 on Build GRCh38
rs747532721 Chr.9: 127825393-127825393 on Build GRCh38
rs763763510 Chr.9: 127825255-127825255 on Build GRCh38
rs990951496 Chr.9: 127825434-127825434 on Build GRCh38
rs895393705 Chr.9: 127825271-127825271 on Build GRCh38
rs547196302 Chr.9: 127825430-127825431 on Build GRCh38
rs779098810 Chr.9: 127825282-127825282 on Build GRCh38
rs557108998 Chr.9: 127825376-127825376 on Build GRCh38
rs1037343267 Chr.9: 127825366-127825366 on Build GRCh38
rs906491967 Chr.9: 127825439-127825439 on Build GRCh38
rs376779134 Chr.9: 127825375-127825375 on Build GRCh38
rs752958277 Chr.9: 127825391-127825391 on Build GRCh38
rs375559067 Chr.9: 127825373-127825373 on Build GRCh38
rs948301971 Chr.9: 127825426-127825426 on Build GRCh38
rs376188437 Chr.9: 127825376-127825376 on Build GRCh38
rs542459678 Chr.9: 127825388-127825388 on Build GRCh38
rs978045920 Chr.9: 127825311-127825311 on Build GRCh38
rs753904326 Chr.9: 127825381-127825381 on Build GRCh38
rs200372420 Chr.9: 127825353-127825353 on Build GRCh38
rs991063378 Chr.9: 127825319-127825319 on Build GRCh38
rs369681397 Chr.9: 127825211-127825211 on Build GRCh38
rs755239753 Chr.9: 127825384-127825384 on Build GRCh38
rs886586243 Chr.9: 127825220-127825220 on Build GRCh38
rs779339363 Chr.9: 127825348-127825348 on Build GRCh38
rs780493782 Chr.9: 127825279-127825279 on Build GRCh38
rs925382657 Chr.9: 127825312-127825312 on Build GRCh38
rs752331196 Chr.9: 127825241-127825241 on Build GRCh38
rs764206593 Chr.9: 127825393-127825393 on Build GRCh38
rs115316938 Chr.9: 127825219-127825219 on Build GRCh38
rs886041326 Chr.9: 127825231-127825231 on Build GRCh38
rs375757277 Chr.9: 127825210-127825210 on Build GRCh38
rs199525659 Chr.9: 127825374-127825374 on Build GRCh38
rs755116051 Chr.9: 127825342-127825342 on Build GRCh38
rs752983157 Chr.9: 127825371-127825379 on Build GRCh38
rs772467926 Chr.9: 127825352-127825352 on Build GRCh38
rs761827492 Chr.9: 127825320-127825320 on Build GRCh38
rs756303592 Chr.9: 127825329-127825329 on Build GRCh38
rs780260845 Chr.9: 127825339-127825339 on Build GRCh38
rs764809807 Chr.9: 127825238-127825238 on Build GRCh38
rs112262663 Chr.9: 127825315-127825315 on Build GRCh38
rs557060922 Chr.9: 127825300-127825300 on Build GRCh38
rs760640573 Chr.9: 127825377-127825377 on Build GRCh38
rs368800416 Chr.9: 127825261-127825261 on Build GRCh38
rs754136153 Chr.9: 127825341-127825341 on Build GRCh38
rs762569294 Chr.9: 127825314-127825314 on Build GRCh38
rs748721879 Chr.9: 127825290-127825290 on Build GRCh38

View Full Product Details