Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75524991-75525264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BATF
Entrez Gene ID
10538
Gene Name
basic leucine zipper ATF-like transcription factor
Gene Aliases
B-ATF, BATF1, SFA-2, SFA2
UniGene
Hs.509964
Gene Chromosome Location
Chr.14: 75522441-75546992 on Build GRCh38
Transcripts
NM_006399.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4450500 c.64-1G>A p.? Chr.14: 75525083-75525083 on Build GRCh38
5676457 c.75T>C p.D25D Chr.14: 75525095-75525095 on Build GRCh38
170604 c.109C>T p.R37C Chr.14: 75525129-75525129 on Build GRCh38
4796522 c.129C>T p.S43S Chr.14: 75525149-75525149 on Build GRCh38
7235114 c.91C>T p.Q31* Chr.14: 75525111-75525111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769335061 Chr.14: 75525069-75525069 on Build GRCh38
rs942975051 Chr.14: 75525201-75525201 on Build GRCh38
rs755608362 Chr.14: 75525196-75525196 on Build GRCh38
rs776402996 Chr.14: 75525174-75525174 on Build GRCh38
rs759437369 Chr.14: 75525096-75525096 on Build GRCh38
rs2540109 Chr.14: 75525209-75525209 on Build GRCh38
rs756280976 Chr.14: 75525041-75525041 on Build GRCh38
rs200342853 Chr.14: 75525104-75525104 on Build GRCh38
rs923675860 Chr.14: 75525090-75525090 on Build GRCh38
rs1019008078 Chr.14: 75525052-75525052 on Build GRCh38
rs758723494 Chr.14: 75525217-75525217 on Build GRCh38
rs765064309 Chr.14: 75525149-75525149 on Build GRCh38
rs1029169171 Chr.14: 75525220-75525220 on Build GRCh38
rs775387933 Chr.14: 75525086-75525086 on Build GRCh38
rs754383692 Chr.14: 75525173-75525173 on Build GRCh38
rs1034923158 Chr.14: 75525042-75525042 on Build GRCh38
rs767726640 Chr.14: 75525137-75525137 on Build GRCh38
rs554606478 Chr.14: 75525057-75525057 on Build GRCh38
rs779291402 Chr.14: 75525205-75525205 on Build GRCh38
rs151038299 Chr.14: 75525068-75525068 on Build GRCh38
rs935511259 Chr.14: 75525122-75525122 on Build GRCh38
rs201306731 Chr.14: 75525056-75525056 on Build GRCh38
rs753058214 Chr.14: 75525216-75525216 on Build GRCh38
rs756227854 Chr.14: 75525142-75525142 on Build GRCh38
rs867753230 Chr.14: 75525146-75525146 on Build GRCh38
rs367895906 Chr.14: 75525240-75525240 on Build GRCh38
rs761224896 Chr.14: 75525239-75525239 on Build GRCh38
rs762940793 Chr.14: 75525091-75525091 on Build GRCh38
rs766614796 Chr.14: 75525161-75525161 on Build GRCh38
rs957648679 Chr.14: 75525222-75525222 on Build GRCh38
rs2540108 Chr.14: 75525208-75525208 on Build GRCh38
rs777206759 Chr.14: 75525223-75525223 on Build GRCh38
rs746635958 Chr.14: 75525063-75525063 on Build GRCh38
rs769099151 Chr.14: 75525189-75525190 on Build GRCh38
rs545498676 Chr.14: 75525243-75525243 on Build GRCh38
rs770560875 Chr.14: 75525066-75525066 on Build GRCh38
rs780663511 Chr.14: 75525045-75525045 on Build GRCh38
rs750637092 Chr.14: 75525140-75525140 on Build GRCh38
rs761523570 Chr.14: 75525134-75525134 on Build GRCh38
rs886974835 Chr.14: 75525012-75525012 on Build GRCh38
rs2540110 Chr.14: 75525215-75525215 on Build GRCh38
rs867226385 Chr.14: 75525178-75525178 on Build GRCh38
rs749851742 Chr.14: 75525046-75525046 on Build GRCh38
rs780866853 Chr.14: 75525229-75525229 on Build GRCh38
rs185424675 Chr.14: 75525195-75525195 on Build GRCh38
rs373715508 Chr.14: 75525036-75525036 on Build GRCh38

View Full Product Details