Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 230249069-230249336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT2
Entrez Gene ID
2590
Gene Name
polypeptide N-acetylgalactosaminyltransferase 2
Gene Aliases
GalNAc-T2
UniGene
Hs.743964
Gene Chromosome Location
Chr.1: 230057789-230282130 on Build GRCh38
Transcripts
NM_001291866.1 NM_004481.4 XM_017000963.1 XM_017000964.1 XM_017000965.1 XM_017000966.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
425555 c.873G>A p.R291R Chr.1: 230249239-230249239 on Build GRCh38
6669079 c.851A>G p.Y284C Chr.1: 230249217-230249217 on Build GRCh38
905343 c.857C>T p.T286M Chr.1: 230249223-230249223 on Build GRCh38
3750981 c.825T>C p.D275D Chr.1: 230249191-230249191 on Build GRCh38
3750982 c.858G>A p.T286T Chr.1: 230249224-230249224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773167377 Chr.1: 230249133-230249133 on Build GRCh38
rs753118010 Chr.1: 230249185-230249185 on Build GRCh38
rs773042550 Chr.1: 230249243-230249243 on Build GRCh38
rs764078856 Chr.1: 230249251-230249252 on Build GRCh38
rs945752069 Chr.1: 230249161-230249161 on Build GRCh38
rs866586671 Chr.1: 230249129-230249129 on Build GRCh38
rs878878591 Chr.1: 230249232-230249232 on Build GRCh38
rs756448084 Chr.1: 230249187-230249187 on Build GRCh38
rs754008774 Chr.1: 230249257-230249257 on Build GRCh38
rs774479602 Chr.1: 230249274-230249274 on Build GRCh38
rs377020573 Chr.1: 230249309-230249309 on Build GRCh38
rs747993447 Chr.1: 230249236-230249236 on Build GRCh38
rs754307633 Chr.1: 230249313-230249313 on Build GRCh38
rs201131857 Chr.1: 230249244-230249244 on Build GRCh38
rs768484528 Chr.1: 230249228-230249228 on Build GRCh38
rs749754600 Chr.1: 230249199-230249199 on Build GRCh38
rs776404910 Chr.1: 230249234-230249234 on Build GRCh38
rs766430695 Chr.1: 230249144-230249144 on Build GRCh38
rs751740822 Chr.1: 230249152-230249152 on Build GRCh38
rs200483633 Chr.1: 230249180-230249180 on Build GRCh38
rs779551095 Chr.1: 230249218-230249218 on Build GRCh38
rs765401508 Chr.1: 230249263-230249270 on Build GRCh38
rs928397505 Chr.1: 230249209-230249209 on Build GRCh38
rs933894333 Chr.1: 230249112-230249112 on Build GRCh38
rs796077715 Chr.1: 230249096-230249096 on Build GRCh38
rs767776362 Chr.1: 230249299-230249299 on Build GRCh38
rs752876828 Chr.1: 230249300-230249300 on Build GRCh38
rs199925000 Chr.1: 230249261-230249261 on Build GRCh38
rs757388468 Chr.1: 230249259-230249260 on Build GRCh38
rs1923950 Chr.1: 230249224-230249224 on Build GRCh38
rs569064893 Chr.1: 230249097-230249097 on Build GRCh38
rs563669465 Chr.1: 230249140-230249140 on Build GRCh38
rs750665719 Chr.1: 230249273-230249281 on Build GRCh38
rs3748008 Chr.1: 230249306-230249306 on Build GRCh38
rs757862876 Chr.1: 230249202-230249202 on Build GRCh38
rs769587065 Chr.1: 230249241-230249241 on Build GRCh38
rs367944876 Chr.1: 230249284-230249284 on Build GRCh38
rs755125585 Chr.1: 230249163-230249163 on Build GRCh38
rs3748006 Chr.1: 230249191-230249191 on Build GRCh38

View Full Product Details