Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 42865041-42865310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACKR2
Entrez Gene ID
1238
Gene Name
atypical chemokine receptor 2
Gene Aliases
CCBP2, CCR10, CCR9, CMKBR9, D6, hD6
UniGene
Hs.146346
Gene Chromosome Location
Chr.3: 42809472-42867283 on Build GRCh38
Transcripts
NM_001296.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5994547 c.636C>A p.F212L Chr.3: 42865138-42865138 on Build GRCh38
5864541 c.739C>T p.R247W Chr.3: 42865241-42865241 on Build GRCh38
336195 c.627C>T p.F209F Chr.3: 42865129-42865129 on Build GRCh38
1044794 c.588C>T p.H196H Chr.3: 42865090-42865090 on Build GRCh38
6192685 c.727G>A p.A243T Chr.3: 42865229-42865229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555778861 Chr.3: 42865262-42865262 on Build GRCh38
rs773498123 Chr.3: 42865179-42865182 on Build GRCh38
rs140892912 Chr.3: 42865106-42865106 on Build GRCh38
rs754635770 Chr.3: 42865090-42865090 on Build GRCh38
rs377711584 Chr.3: 42865279-42865279 on Build GRCh38
rs139094169 Chr.3: 42865286-42865286 on Build GRCh38
rs765210048 Chr.3: 42865275-42865275 on Build GRCh38
rs140255308 Chr.3: 42865100-42865100 on Build GRCh38
rs754502272 Chr.3: 42865244-42865244 on Build GRCh38
rs367690990 Chr.3: 42865073-42865073 on Build GRCh38
rs149364873 Chr.3: 42865137-42865137 on Build GRCh38
rs146641213 Chr.3: 42865133-42865133 on Build GRCh38
rs749698117 Chr.3: 42865182-42865184 on Build GRCh38
rs752550350 Chr.3: 42865151-42865151 on Build GRCh38
rs773787036 Chr.3: 42865241-42865241 on Build GRCh38
rs147684121 Chr.3: 42865155-42865155 on Build GRCh38
rs762302008 Chr.3: 42865071-42865071 on Build GRCh38
rs192611692 Chr.3: 42865212-42865212 on Build GRCh38
rs375679675 Chr.3: 42865196-42865196 on Build GRCh38
rs375376109 Chr.3: 42865102-42865102 on Build GRCh38
rs780038565 Chr.3: 42865190-42865190 on Build GRCh38
rs569371195 Chr.3: 42865091-42865091 on Build GRCh38
rs972813788 Chr.3: 42865202-42865202 on Build GRCh38
rs780349611 Chr.3: 42865104-42865104 on Build GRCh38
rs142132088 Chr.3: 42865207-42865207 on Build GRCh38
rs368719352 Chr.3: 42865271-42865271 on Build GRCh38
rs748499771 Chr.3: 42865120-42865120 on Build GRCh38
rs775246041 Chr.3: 42865256-42865256 on Build GRCh38
rs764033388 Chr.3: 42865263-42865263 on Build GRCh38
rs773667338 Chr.3: 42865131-42865131 on Build GRCh38
rs777049535 Chr.3: 42865115-42865115 on Build GRCh38
rs3732859 Chr.3: 42865108-42865108 on Build GRCh38
rs781335034 Chr.3: 42865197-42865197 on Build GRCh38
rs747107773 Chr.3: 42865194-42865194 on Build GRCh38
rs755009181 Chr.3: 42865280-42865280 on Build GRCh38
rs201310570 Chr.3: 42865200-42865200 on Build GRCh38
rs2228469 Chr.3: 42865245-42865245 on Build GRCh38
rs372734430 Chr.3: 42865254-42865254 on Build GRCh38
rs770152822 Chr.3: 42865073-42865074 on Build GRCh38
rs765855167 Chr.3: 42865132-42865132 on Build GRCh38
rs368758367 Chr.3: 42865103-42865103 on Build GRCh38
rs993789686 Chr.3: 42865101-42865101 on Build GRCh38
rs767262136 Chr.3: 42865149-42865149 on Build GRCh38
rs771462120 Chr.3: 42865230-42865230 on Build GRCh38
rs369814928 Chr.3: 42865238-42865238 on Build GRCh38
rs960929452 Chr.3: 42865076-42865076 on Build GRCh38
rs753637859 Chr.3: 42865265-42865265 on Build GRCh38
rs773542847 Chr.3: 42865210-42865210 on Build GRCh38
rs146146689 Chr.3: 42865169-42865169 on Build GRCh38
rs376898493 Chr.3: 42865261-42865261 on Build GRCh38
rs751123143 Chr.3: 42865084-42865084 on Build GRCh38
rs770288240 Chr.3: 42865128-42865128 on Build GRCh38

View Full Product Details