Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177366873-177367128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RGS14
Entrez Gene ID
10636
Gene Name
regulator of G-protein signaling 14
Gene Aliases
-
UniGene
Hs.9347
Gene Chromosome Location
Chr.5: 177357671-177372601 on Build GRCh38
Transcripts
NM_006480.4 XM_005265794.4 XM_005265795.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5551587 c.409G>T p.D137Y Chr.5: 177366960-177366960 on Build GRCh38
6765846 c.446_450delAGCCC p.E149fs*55 Chr.5: 177366997-177367001 on Build GRCh38
3661735 c.412C>G p.R138G Chr.5: 177366963-177366963 on Build GRCh38
1436301 c.390G>A p.A130A Chr.5: 177366941-177366941 on Build GRCh38
4333156 c.468G>A p.R156R Chr.5: 177367019-177367019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763342865 Chr.5: 177366930-177366930 on Build GRCh38
rs929265687 Chr.5: 177366995-177366995 on Build GRCh38
rs780292648 Chr.5: 177366893-177366893 on Build GRCh38
rs777672925 Chr.5: 177367082-177367082 on Build GRCh38
rs768803674 Chr.5: 177366990-177366990 on Build GRCh38
rs752196625 Chr.5: 177366953-177366953 on Build GRCh38
rs188681772 Chr.5: 177366910-177366910 on Build GRCh38
rs762316004 Chr.5: 177366914-177366914 on Build GRCh38
rs180829045 Chr.5: 177367059-177367059 on Build GRCh38
rs755254716 Chr.5: 177367066-177367066 on Build GRCh38
rs773511198 Chr.5: 177366923-177366923 on Build GRCh38
rs759274665 Chr.5: 177367018-177367018 on Build GRCh38
rs778526422 Chr.5: 177366979-177366979 on Build GRCh38
rs760231858 Chr.5: 177367048-177367048 on Build GRCh38
rs956714512 Chr.5: 177367083-177367083 on Build GRCh38
rs765993641 Chr.5: 177366940-177366940 on Build GRCh38
rs200942382 Chr.5: 177366941-177366941 on Build GRCh38
rs766162868 Chr.5: 177367017-177367017 on Build GRCh38
rs759223328 Chr.5: 177366943-177366943 on Build GRCh38
rs747289630 Chr.5: 177366988-177366988 on Build GRCh38
rs767139409 Chr.5: 177366951-177366951 on Build GRCh38
rs199937137 Chr.5: 177367084-177367084 on Build GRCh38
rs768748398 Chr.5: 177366906-177366906 on Build GRCh38
rs1038967986 Chr.5: 177367043-177367043 on Build GRCh38
rs748272716 Chr.5: 177367077-177367077 on Build GRCh38
rs781268208 Chr.5: 177366994-177366994 on Build GRCh38
rs775207129 Chr.5: 177367041-177367041 on Build GRCh38
rs926621848 Chr.5: 177366931-177366931 on Build GRCh38
rs765087807 Chr.5: 177367054-177367054 on Build GRCh38
rs757105254 Chr.5: 177367053-177367053 on Build GRCh38
rs781357280 Chr.5: 177367067-177367067 on Build GRCh38
rs879578844 Chr.5: 177366977-177366977 on Build GRCh38
rs565630488 Chr.5: 177367103-177367103 on Build GRCh38
rs757055540 Chr.5: 177366974-177366974 on Build GRCh38
rs568641565 Chr.5: 177367049-177367049 on Build GRCh38
rs773744437 Chr.5: 177367008-177367008 on Build GRCh38
rs376359696 Chr.5: 177366980-177366980 on Build GRCh38
rs1007840305 Chr.5: 177367039-177367039 on Build GRCh38
rs756178898 Chr.5: 177366964-177366964 on Build GRCh38
rs756205094 Chr.5: 177367080-177367080 on Build GRCh38
rs550386385 Chr.5: 177367002-177367002 on Build GRCh38
rs753838091 Chr.5: 177367052-177367052 on Build GRCh38
rs929091059 Chr.5: 177366972-177366972 on Build GRCh38
rs767265876 Chr.5: 177367021-177367021 on Build GRCh38
rs771283979 Chr.5: 177367086-177367086 on Build GRCh38
rs545846792 Chr.5: 177366898-177366898 on Build GRCh38
rs771356131 Chr.5: 177367012-177367012 on Build GRCh38
rs369592386 Chr.5: 177366922-177366922 on Build GRCh38
rs372902128 Chr.5: 177366969-177366969 on Build GRCh38

View Full Product Details