Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 93132945-93133206 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAMD9L
Entrez Gene ID
219285
Gene Name
sterile alpha motif domain containing 9 like
Gene Aliases
C7orf6, DRIF2, UEF1
UniGene
Hs.489118
Gene Chromosome Location
Chr.7: 93130054-93148401 on Build GRCh38
Transcripts
NM_001303496.1 NM_001303497.1 NM_001303498.1 NM_001303500.1 NM_152703.3 XM_005250193.3 XM_006715890.1 XM_011515903.2 XM_017011822.1 XM_017011823.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6179022 c.2855G>C p.S952T Chr.7: 93133117-93133117 on Build GRCh38
5873380 c.2827G>A p.E943K Chr.7: 93133145-93133145 on Build GRCh38
5873379 c.2900A>T p.Y967F Chr.7: 93133072-93133072 on Build GRCh38
5408270 c.2768C>T p.S923F Chr.7: 93133204-93133204 on Build GRCh38
384971 c.2864G>T p.W955L Chr.7: 93133108-93133108 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010536613 Chr.7: 93133037-93133037 on Build GRCh38
rs765627772 Chr.7: 93133117-93133117 on Build GRCh38
rs536966231 Chr.7: 93133120-93133120 on Build GRCh38
rs755283987 Chr.7: 93133119-93133120 on Build GRCh38
rs370300473 Chr.7: 93133184-93133184 on Build GRCh38
rs776375139 Chr.7: 93133009-93133009 on Build GRCh38
rs960927778 Chr.7: 93133180-93133180 on Build GRCh38
rs772940160 Chr.7: 93133026-93133026 on Build GRCh38
rs978634830 Chr.7: 93133171-93133171 on Build GRCh38
rs746480205 Chr.7: 93133173-93133173 on Build GRCh38
rs78528622 Chr.7: 93133125-93133125 on Build GRCh38
rs267601635 Chr.7: 93132977-93132977 on Build GRCh38
rs935878482 Chr.7: 93133003-93133003 on Build GRCh38
rs757476135 Chr.7: 93133116-93133116 on Build GRCh38
rs367669568 Chr.7: 93133179-93133179 on Build GRCh38
rs764374618 Chr.7: 93133086-93133086 on Build GRCh38
rs925286289 Chr.7: 93132986-93132986 on Build GRCh38
rs754063645 Chr.7: 93133111-93133111 on Build GRCh38
rs374124481 Chr.7: 93133122-93133122 on Build GRCh38
rs766424103 Chr.7: 93133052-93133052 on Build GRCh38
rs760863414 Chr.7: 93133080-93133080 on Build GRCh38
rs142489925 Chr.7: 93133047-93133047 on Build GRCh38
rs747794349 Chr.7: 93133014-93133014 on Build GRCh38
rs765998314 Chr.7: 93132974-93132974 on Build GRCh38
rs900211001 Chr.7: 93133036-93133036 on Build GRCh38
rs768248968 Chr.7: 93132993-93132993 on Build GRCh38
rs567728437 Chr.7: 93133040-93133040 on Build GRCh38
rs754453650 Chr.7: 93133175-93133175 on Build GRCh38
rs775657123 Chr.7: 93133071-93133071 on Build GRCh38
rs1014081460 Chr.7: 93133073-93133073 on Build GRCh38
rs780560641 Chr.7: 93133147-93133147 on Build GRCh38
rs769611275 Chr.7: 93133015-93133015 on Build GRCh38

View Full Product Details