Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52229715-52229976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TWF2
Entrez Gene ID
11344
Gene Name
twinfilin actin binding protein 2
Gene Aliases
A6RP, A6r, MSTP011, PTK9L
UniGene
Hs.436439
Gene Chromosome Location
Chr.3: 52228610-52239167 on Build GRCh38
Transcripts
NM_007284.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4881310 c.746C>A p.P249H Chr.3: 52229934-52229934 on Build GRCh38
1046841 c.805C>T p.R269* Chr.3: 52229738-52229738 on Build GRCh38
3696181 c.741C>T p.G247G Chr.3: 52229939-52229939 on Build GRCh38
5937122 c.778C>T p.P260S Chr.3: 52229765-52229765 on Build GRCh38
3760161 c.729C>T p.H243H Chr.3: 52229951-52229951 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746396924 Chr.3: 52229871-52229871 on Build GRCh38
rs201549689 Chr.3: 52229904-52229904 on Build GRCh38
rs973481835 Chr.3: 52229807-52229807 on Build GRCh38
rs767283869 Chr.3: 52229741-52229741 on Build GRCh38
rs765236477 Chr.3: 52229770-52229770 on Build GRCh38
rs377621507 Chr.3: 52229887-52229887 on Build GRCh38
rs922757042 Chr.3: 52229758-52229758 on Build GRCh38
rs563863286 Chr.3: 52229837-52229837 on Build GRCh38
rs918521558 Chr.3: 52229838-52229838 on Build GRCh38
rs771749590 Chr.3: 52229876-52229876 on Build GRCh38
rs750584587 Chr.3: 52229787-52229788 on Build GRCh38
rs374109549 Chr.3: 52229886-52229886 on Build GRCh38
rs756643412 Chr.3: 52229939-52229939 on Build GRCh38
rs890732288 Chr.3: 52229842-52229842 on Build GRCh38
rs34038856 Chr.3: 52229951-52229951 on Build GRCh38
rs367943989 Chr.3: 52229913-52229913 on Build GRCh38
rs892844985 Chr.3: 52229893-52229893 on Build GRCh38
rs752893678 Chr.3: 52229894-52229894 on Build GRCh38
rs764462777 Chr.3: 52229925-52229925 on Build GRCh38
rs761464663 Chr.3: 52229738-52229738 on Build GRCh38
rs760572180 Chr.3: 52229751-52229751 on Build GRCh38
rs758753199 Chr.3: 52229793-52229793 on Build GRCh38
rs757685379 Chr.3: 52229810-52229810 on Build GRCh38
rs755045431 Chr.3: 52229765-52229765 on Build GRCh38
rs766387538 Chr.3: 52229760-52229760 on Build GRCh38
rs773055657 Chr.3: 52229747-52229747 on Build GRCh38
rs1011134234 Chr.3: 52229783-52229783 on Build GRCh38
rs370212205 Chr.3: 52229788-52229788 on Build GRCh38
rs114956852 Chr.3: 52229932-52229932 on Build GRCh38
rs565212275 Chr.3: 52229927-52229927 on Build GRCh38
rs1038727248 Chr.3: 52229870-52229870 on Build GRCh38
rs141598715 Chr.3: 52229843-52229843 on Build GRCh38
rs542373367 Chr.3: 52229867-52229867 on Build GRCh38
rs547826086 Chr.3: 52229938-52229938 on Build GRCh38
rs985943479 Chr.3: 52229777-52229777 on Build GRCh38
rs753740098 Chr.3: 52229764-52229764 on Build GRCh38
rs778280155 Chr.3: 52229797-52229797 on Build GRCh38
rs752013242 Chr.3: 52229802-52229802 on Build GRCh38
rs929854227 Chr.3: 52229839-52229839 on Build GRCh38
rs199663186 Chr.3: 52229880-52229880 on Build GRCh38
rs765347323 Chr.3: 52229889-52229889 on Build GRCh38
rs747791708 Chr.3: 52229875-52229875 on Build GRCh38
rs201505921 Chr.3: 52229835-52229835 on Build GRCh38
rs781419320 Chr.3: 52229935-52229935 on Build GRCh38
rs374922294 Chr.3: 52229818-52229818 on Build GRCh38
rs772967626 Chr.3: 52229877-52229877 on Build GRCh38
rs759406321 Chr.3: 52229888-52229888 on Build GRCh38
rs778448361 Chr.3: 52229872-52229872 on Build GRCh38
rs763239264 Chr.3: 52229922-52229922 on Build GRCh38

View Full Product Details