Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151520299-151520538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CGN
Entrez Gene ID
57530
Gene Name
cingulin
Gene Aliases
-
UniGene
Hs.591464
Gene Chromosome Location
Chr.1: 151510859-151538692 on Build GRCh38
Transcripts
NM_020770.2 XM_005245365.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4770848 c.1029G>A p.K343K Chr.1: 151520468-151520468 on Build GRCh38
1134700 c.1001G>A p.R334K Chr.1: 151520440-151520440 on Build GRCh38
6894850 c.982G>A p.E328K Chr.1: 151520421-151520421 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752065051 Chr.1: 151520454-151520454 on Build GRCh38
rs778192060 Chr.1: 151520511-151520511 on Build GRCh38
rs761385304 Chr.1: 151520388-151520388 on Build GRCh38
rs758810075 Chr.1: 151520420-151520420 on Build GRCh38
rs745630003 Chr.1: 151520513-151520513 on Build GRCh38
rs763614980 Chr.1: 151520364-151520364 on Build GRCh38
rs748158258 Chr.1: 151520493-151520495 on Build GRCh38
rs1045434981 Chr.1: 151520444-151520444 on Build GRCh38
rs905224025 Chr.1: 151520480-151520480 on Build GRCh38
rs780085429 Chr.1: 151520448-151520448 on Build GRCh38
rs756652429 Chr.1: 151520399-151520399 on Build GRCh38
rs755461546 Chr.1: 151520456-151520456 on Build GRCh38
rs748242779 Chr.1: 151520494-151520494 on Build GRCh38
rs1052529522 Chr.1: 151520352-151520352 on Build GRCh38
rs78845974 Chr.1: 151520514-151520514 on Build GRCh38
rs138880280 Chr.1: 151520478-151520478 on Build GRCh38
rs963995855 Chr.1: 151520457-151520457 on Build GRCh38
rs770130033 Chr.1: 151520496-151520496 on Build GRCh38
rs775175619 Chr.1: 151520518-151520518 on Build GRCh38
rs750770104 Chr.1: 151520409-151520409 on Build GRCh38
rs764932242 Chr.1: 151520407-151520407 on Build GRCh38
rs935509234 Chr.1: 151520348-151520348 on Build GRCh38
rs753305062 Chr.1: 151520369-151520369 on Build GRCh38
rs182890079 Chr.1: 151520342-151520342 on Build GRCh38
rs781404399 Chr.1: 151520438-151520440 on Build GRCh38
rs115192275 Chr.1: 151520436-151520436 on Build GRCh38

View Full Product Details