Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 19308103-19308356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR7A2
Entrez Gene ID
8574
Gene Name
aldo-keto reductase family 7 member A2
Gene Aliases
AFAR, AFAR1, AFB1-AR1, AKR7
UniGene
Hs.571886
Gene Chromosome Location
Chr.1: 19302708-19312146 on Build GRCh38
Transcripts
NM_001320979.1 NM_003689.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4026488 c.514A>G p.N172D Chr.1: 19308235-19308235 on Build GRCh38
6060136 c.591+1G>T p.? Chr.1: 19308157-19308157 on Build GRCh38
3863954 c.567C>T p.G189G Chr.1: 19308182-19308182 on Build GRCh38
1491921 c.523A>G p.S175G Chr.1: 19308226-19308226 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774097352 Chr.1: 19308195-19308195 on Build GRCh38
rs764419671 Chr.1: 19308268-19308268 on Build GRCh38
rs895372975 Chr.1: 19308319-19308319 on Build GRCh38
rs373108163 Chr.1: 19308275-19308275 on Build GRCh38
rs749041413 Chr.1: 19308185-19308185 on Build GRCh38
rs546461253 Chr.1: 19308123-19308123 on Build GRCh38
rs777459748 Chr.1: 19308294-19308294 on Build GRCh38
rs778253368 Chr.1: 19308132-19308132 on Build GRCh38
rs779252486 Chr.1: 19308140-19308140 on Build GRCh38
rs758848168 Chr.1: 19308280-19308280 on Build GRCh38
rs765791064 Chr.1: 19308225-19308225 on Build GRCh38
rs548010540 Chr.1: 19308231-19308231 on Build GRCh38
rs113687169 Chr.1: 19308278-19308278 on Build GRCh38
rs371034673 Chr.1: 19308277-19308277 on Build GRCh38
rs745763571 Chr.1: 19308258-19308258 on Build GRCh38
rs747316988 Chr.1: 19308330-19308330 on Build GRCh38
rs893157364 Chr.1: 19308202-19308202 on Build GRCh38
rs549972531 Chr.1: 19308158-19308158 on Build GRCh38
rs1012232144 Chr.1: 19308131-19308131 on Build GRCh38
rs2231199 Chr.1: 19308148-19308148 on Build GRCh38
rs201886268 Chr.1: 19308199-19308199 on Build GRCh38
rs914330868 Chr.1: 19308261-19308261 on Build GRCh38
rs778551174 Chr.1: 19308312-19308312 on Build GRCh38
rs777339767 Chr.1: 19308250-19308250 on Build GRCh38
rs775896270 Chr.1: 19308201-19308201 on Build GRCh38
rs757117160 Chr.1: 19308311-19308311 on Build GRCh38
rs2231197 Chr.1: 19308273-19308273 on Build GRCh38
rs199820959 Chr.1: 19308243-19308243 on Build GRCh38
rs859209 Chr.1: 19308289-19308289 on Build GRCh38
rs77672886 Chr.1: 19308212-19308212 on Build GRCh38
rs762468852 Chr.1: 19308145-19308145 on Build GRCh38
rs767492017 Chr.1: 19308200-19308200 on Build GRCh38
rs773008746 Chr.1: 19308178-19308178 on Build GRCh38
rs754446849 Chr.1: 19308136-19308136 on Build GRCh38
rs776938292 Chr.1: 19308272-19308272 on Build GRCh38
rs757810257 Chr.1: 19308138-19308138 on Build GRCh38
rs372566686 Chr.1: 19308265-19308265 on Build GRCh38
rs143442378 Chr.1: 19308252-19308252 on Build GRCh38
rs144757440 Chr.1: 19308172-19308172 on Build GRCh38
rs760801737 Chr.1: 19308267-19308267 on Build GRCh38
rs756614465 Chr.1: 19308122-19308122 on Build GRCh38
rs377217513 Chr.1: 19308254-19308254 on Build GRCh38
rs747765181 Chr.1: 19308165-19308165 on Build GRCh38
rs755562654 Chr.1: 19308290-19308290 on Build GRCh38
rs757500829 Chr.1: 19308218-19308218 on Build GRCh38
rs750883524 Chr.1: 19308226-19308226 on Build GRCh38
rs575968287 Chr.1: 19308136-19308136 on Build GRCh38
rs765880489 Chr.1: 19308223-19308224 on Build GRCh38
rs531745035 Chr.1: 19308149-19308149 on Build GRCh38
rs959343673 Chr.1: 19308129-19308129 on Build GRCh38
rs111682618 Chr.1: 19308253-19308253 on Build GRCh38
rs199593810 Chr.1: 19308227-19308227 on Build GRCh38
rs754611824 Chr.1: 19308233-19308234 on Build GRCh38
rs751138262 Chr.1: 19308227-19308227 on Build GRCh38
rs753425844 Chr.1: 19308296-19308296 on Build GRCh38
rs1054020533 Chr.1: 19308260-19308260 on Build GRCh38
rs770904476 Chr.1: 19308186-19308186 on Build GRCh38
rs370412117 Chr.1: 19308263-19308263 on Build GRCh38
rs940846735 Chr.1: 19308146-19308146 on Build GRCh38
rs859210 Chr.1: 19308211-19308211 on Build GRCh38

View Full Product Details