Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 142748095-142748346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPC1
Entrez Gene ID
7220
Gene Name
transient receptor potential cation channel subfamily C member 1
Gene Aliases
HTRP-1, TRP1
UniGene
Hs.250687
Gene Chromosome Location
Chr.3: 142724032-142807888 on Build GRCh38
Transcripts
NM_001251845.1 NM_003304.4 XM_005247738.3 XM_005247739.2 XM_017007121.1 XM_017007122.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1230492 c.341G>A p.R114Q Chr.3: 142748271-142748271 on Build GRCh38
4319656 c.443G>A p.R148Q Chr.3: 142748271-142748271 on Build GRCh38
6379965 c.506G>A p.R169H Chr.3: 142748334-142748334 on Build GRCh38
6379964 c.404G>A p.R135H Chr.3: 142748334-142748334 on Build GRCh38
1670619 c.373G>T p.D125Y Chr.3: 142748303-142748303 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs928238243 Chr.3: 142748182-142748182 on Build GRCh38
rs898789221 Chr.3: 142748300-142748300 on Build GRCh38
rs945539944 Chr.3: 142748139-142748139 on Build GRCh38
rs746525030 Chr.3: 142748258-142748258 on Build GRCh38
rs546908763 Chr.3: 142748214-142748214 on Build GRCh38
rs776572650 Chr.3: 142748262-142748262 on Build GRCh38
rs766526012 Chr.3: 142748306-142748306 on Build GRCh38
rs982353364 Chr.3: 142748175-142748175 on Build GRCh38
rs995478238 Chr.3: 142748205-142748206 on Build GRCh38
rs759226939 Chr.3: 142748281-142748281 on Build GRCh38
rs917289891 Chr.3: 142748286-142748286 on Build GRCh38
rs914113334 Chr.3: 142748133-142748133 on Build GRCh38
rs772948472 Chr.3: 142748295-142748295 on Build GRCh38
rs768539925 Chr.3: 142748261-142748261 on Build GRCh38
rs963638947 Chr.3: 142748203-142748203 on Build GRCh38
rs571634574 Chr.3: 142748236-142748236 on Build GRCh38
rs778035798 Chr.3: 142748217-142748217 on Build GRCh38
rs761788159 Chr.3: 142748270-142748270 on Build GRCh38
rs763225534 Chr.3: 142748303-142748303 on Build GRCh38
rs113733932 Chr.3: 142748179-142748179 on Build GRCh38
rs539204792 Chr.3: 142748271-142748271 on Build GRCh38
rs373336987 Chr.3: 142748254-142748254 on Build GRCh38
rs745526976 Chr.3: 142748232-142748232 on Build GRCh38

View Full Product Details