Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144311013-144311240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSF1
Entrez Gene ID
3297
Gene Name
heat shock transcription factor 1
Gene Aliases
HSTF1
UniGene
Hs.530227
Gene Chromosome Location
Chr.8: 144291588-144314722 on Build GRCh38
Transcripts
NM_005526.2 XM_005272315.2 XM_005272316.2 XM_005272317.1 XM_011517004.2 XM_017013377.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4498606 c.523C>T p.L175F Chr.8: 144311208-144311208 on Build GRCh38
6217271 c.518C>G p.A173G Chr.8: 144311203-144311203 on Build GRCh38
4807610 c.543G>A p.Q181Q Chr.8: 144311228-144311228 on Build GRCh38
4767324 c.508C>T p.R170W Chr.8: 144311193-144311193 on Build GRCh38
6496628 c.526C>T p.R176W Chr.8: 144311211-144311211 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782208328 Chr.8: 144311185-144311185 on Build GRCh38
rs782192401 Chr.8: 144311151-144311151 on Build GRCh38
rs752756948 Chr.8: 144311048-144311048 on Build GRCh38
rs749492818 Chr.8: 144311088-144311088 on Build GRCh38
rs782615937 Chr.8: 144311177-144311177 on Build GRCh38
rs987428937 Chr.8: 144311160-144311160 on Build GRCh38
rs112403012 Chr.8: 144311119-144311119 on Build GRCh38
rs531255438 Chr.8: 144311075-144311075 on Build GRCh38
rs782386108 Chr.8: 144311187-144311187 on Build GRCh38
rs782513340 Chr.8: 144311144-144311144 on Build GRCh38
rs543995213 Chr.8: 144311091-144311091 on Build GRCh38
rs781974287 Chr.8: 144311200-144311200 on Build GRCh38
rs910843707 Chr.8: 144311180-144311180 on Build GRCh38
rs561881157 Chr.8: 144311065-144311065 on Build GRCh38
rs782423963 Chr.8: 144311208-144311208 on Build GRCh38
rs573761548 Chr.8: 144311103-144311103 on Build GRCh38
rs782675794 Chr.8: 144311146-144311146 on Build GRCh38
rs756285321 Chr.8: 144311049-144311049 on Build GRCh38
rs782152708 Chr.8: 144311203-144311203 on Build GRCh38
rs977893067 Chr.8: 144311094-144311094 on Build GRCh38
rs968853280 Chr.8: 144311086-144311086 on Build GRCh38
rs544022889 Chr.8: 144311114-144311114 on Build GRCh38
rs924660205 Chr.8: 144311112-144311112 on Build GRCh38
rs943618551 Chr.8: 144311212-144311212 on Build GRCh38
rs959247761 Chr.8: 144311052-144311052 on Build GRCh38
rs782306776 Chr.8: 144311159-144311159 on Build GRCh38
rs781942523 Chr.8: 144311211-144311211 on Build GRCh38
rs781875024 Chr.8: 144311171-144311171 on Build GRCh38

View Full Product Details