Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 19964853-19965116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIN2
Entrez Gene ID
54453
Gene Name
Ras and Rab interactor 2
Gene Aliases
MACS, RASSF4
UniGene
Hs.472270
Gene Chromosome Location
Chr.20: 19757610-20002459 on Build GRCh38
Transcripts
NM_001242581.1 NM_018993.3 XM_005260731.2 XM_005260733.2 XM_006723574.3 XM_006723575.3 XM_006723577.2 XM_011529255.2 XM_011529257.1 XM_011529258.2 XM_011529259.2 XM_017027887.1 XM_017027888.1 XM_017027889.1 XM_017027890.1 XM_017027891.1 XM_017027892.1 XM_017027893.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5963642 c.526T>C p.C176R Chr.20: 19965014-19965014 on Build GRCh38
6274810 c.509G>T p.R170L Chr.20: 19964997-19964997 on Build GRCh38
4721788 c.613delT p.S206fs*30 Chr.20: 19964954-19964954 on Build GRCh38
4721789 c.466delT p.S157fs*30 Chr.20: 19964954-19964954 on Build GRCh38
3972453 c.642C>T p.F214F Chr.20: 19964983-19964983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190009357 Chr.20: 19965065-19965065 on Build GRCh38
rs748229896 Chr.20: 19964959-19964959 on Build GRCh38
rs749571406 Chr.20: 19964908-19964908 on Build GRCh38
rs772082705 Chr.20: 19964960-19964960 on Build GRCh38
rs750214530 Chr.20: 19965031-19965031 on Build GRCh38
rs377681843 Chr.20: 19964996-19964996 on Build GRCh38
rs781134385 Chr.20: 19965030-19965030 on Build GRCh38
rs757117401 Chr.20: 19965003-19965003 on Build GRCh38
rs752333750 Chr.20: 19964995-19964995 on Build GRCh38
rs556669989 Chr.20: 19965081-19965081 on Build GRCh38
rs374006351 Chr.20: 19965040-19965040 on Build GRCh38
rs369430457 Chr.20: 19964962-19964962 on Build GRCh38
rs776841443 Chr.20: 19964973-19964973 on Build GRCh38
rs952658505 Chr.20: 19964892-19964892 on Build GRCh38
rs376177658 Chr.20: 19964918-19964918 on Build GRCh38
rs964148897 Chr.20: 19964910-19964910 on Build GRCh38
rs976447899 Chr.20: 19964943-19964943 on Build GRCh38
rs439717 Chr.20: 19964878-19964878 on Build GRCh38
rs536008278 Chr.20: 19965077-19965077 on Build GRCh38
rs764833099 Chr.20: 19964984-19964984 on Build GRCh38
rs752479265 Chr.20: 19964930-19964930 on Build GRCh38
rs544585908 Chr.20: 19965073-19965073 on Build GRCh38
rs74719456 Chr.20: 19964897-19964897 on Build GRCh38
rs779016747 Chr.20: 19965055-19965055 on Build GRCh38
rs927332798 Chr.20: 19964982-19964982 on Build GRCh38
rs762118037 Chr.20: 19965001-19965001 on Build GRCh38
rs369874324 Chr.20: 19964997-19964997 on Build GRCh38
rs759898074 Chr.20: 19964983-19964983 on Build GRCh38
rs777801391 Chr.20: 19965074-19965074 on Build GRCh38
rs947629258 Chr.20: 19964999-19964999 on Build GRCh38
rs374144324 Chr.20: 19964967-19964967 on Build GRCh38
rs767323971 Chr.20: 19964909-19964909 on Build GRCh38
rs770161353 Chr.20: 19965043-19965044 on Build GRCh38

View Full Product Details