Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50505390-50505615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMF2
Entrez Gene ID
91289
Gene Name
lipase maturation factor 2
Gene Aliases
TMEM112B, TMEM153
UniGene
Hs.150540
Gene Chromosome Location
Chr.22: 50502947-50507840 on Build GRCh38
Transcripts
NM_033200.2 XM_005261955.3 XM_006724426.3 XM_006724427.3 XM_017029077.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4105201 c.892G>A p.V298I Chr.22: 50505487-50505487 on Build GRCh38
6701754 c.897C>T p.Y299Y Chr.22: 50505482-50505482 on Build GRCh38
5888482 c.917-6C>T p.? Chr.22: 50505543-50505543 on Build GRCh38
1213482 c.926A>G p.Y309C Chr.22: 50505453-50505453 on Build GRCh38
6292939 c.853G>A p.A285T Chr.22: 50505526-50505526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780870819 Chr.22: 50505413-50505413 on Build GRCh38
rs372876010 Chr.22: 50505554-50505554 on Build GRCh38
rs560502663 Chr.22: 50505586-50505586 on Build GRCh38
rs937431548 Chr.22: 50505545-50505545 on Build GRCh38
rs774817134 Chr.22: 50505460-50505460 on Build GRCh38
rs901778639 Chr.22: 50505569-50505569 on Build GRCh38
rs527860246 Chr.22: 50505589-50505589 on Build GRCh38
rs759012982 Chr.22: 50505565-50505565 on Build GRCh38
rs780731413 Chr.22: 50505440-50505440 on Build GRCh38
rs952985012 Chr.22: 50505584-50505584 on Build GRCh38
rs1028669938 Chr.22: 50505582-50505582 on Build GRCh38
rs986623550 Chr.22: 50505436-50505436 on Build GRCh38
rs566196885 Chr.22: 50505573-50505573 on Build GRCh38
rs776975131 Chr.22: 50505524-50505524 on Build GRCh38
rs777321260 Chr.22: 50505446-50505446 on Build GRCh38
rs780139990 Chr.22: 50505434-50505434 on Build GRCh38
rs1045747247 Chr.22: 50505533-50505533 on Build GRCh38
rs375917497 Chr.22: 50505567-50505567 on Build GRCh38
rs116898513 Chr.22: 50505487-50505487 on Build GRCh38
rs145130809 Chr.22: 50505478-50505478 on Build GRCh38
rs748314717 Chr.22: 50505588-50505588 on Build GRCh38
rs377204899 Chr.22: 50505475-50505475 on Build GRCh38
rs549316793 Chr.22: 50505594-50505594 on Build GRCh38
rs752575906 Chr.22: 50505532-50505532 on Build GRCh38
rs760890488 Chr.22: 50505471-50505471 on Build GRCh38
rs368606508 Chr.22: 50505543-50505543 on Build GRCh38
rs747660191 Chr.22: 50505415-50505415 on Build GRCh38
rs769300731 Chr.22: 50505418-50505418 on Build GRCh38
rs568125029 Chr.22: 50505521-50505521 on Build GRCh38
rs755860692 Chr.22: 50505495-50505495 on Build GRCh38
rs146257188 Chr.22: 50505456-50505456 on Build GRCh38
rs760697194 Chr.22: 50505426-50505426 on Build GRCh38
rs141737208 Chr.22: 50505437-50505437 on Build GRCh38
rs1031456849 Chr.22: 50505505-50505505 on Build GRCh38
rs772437586 Chr.22: 50505466-50505466 on Build GRCh38
rs139703456 Chr.22: 50505507-50505507 on Build GRCh38
rs1028278179 Chr.22: 50505414-50505414 on Build GRCh38
rs972789876 Chr.22: 50505591-50505591 on Build GRCh38
rs150605960 Chr.22: 50505506-50505506 on Build GRCh38
rs533762764 Chr.22: 50505488-50505488 on Build GRCh38
rs763391596 Chr.22: 50505423-50505423 on Build GRCh38
rs772334657 Chr.22: 50505509-50505509 on Build GRCh38
rs759911899 Chr.22: 50505464-50505464 on Build GRCh38
rs765368432 Chr.22: 50505529-50505529 on Build GRCh38
rs966131083 Chr.22: 50505467-50505467 on Build GRCh38
rs749002667 Chr.22: 50505481-50505481 on Build GRCh38
rs768494826 Chr.22: 50505412-50505412 on Build GRCh38
rs777372632 Chr.22: 50505497-50505497 on Build GRCh38
rs778333828 Chr.22: 50505559-50505559 on Build GRCh38
rs752419869 Chr.22: 50505490-50505490 on Build GRCh38
rs758470413 Chr.22: 50505432-50505432 on Build GRCh38
rs771467410 Chr.22: 50505458-50505458 on Build GRCh38
rs764164997 Chr.22: 50505427-50505427 on Build GRCh38
rs769044240 Chr.22: 50505523-50505523 on Build GRCh38
rs950070531 Chr.22: 50505422-50505422 on Build GRCh38
rs369533048 Chr.22: 50505428-50505428 on Build GRCh38
rs370167838 Chr.22: 50505504-50505504 on Build GRCh38
rs752911271 Chr.22: 50505425-50505425 on Build GRCh38
rs367908503 Chr.22: 50505564-50505564 on Build GRCh38
rs149052579 Chr.22: 50505410-50505410 on Build GRCh38
rs372747521 Chr.22: 50505439-50505439 on Build GRCh38
rs747797641 Chr.22: 50505444-50505444 on Build GRCh38
rs762006530 Chr.22: 50505527-50505527 on Build GRCh38
rs755841844 Chr.22: 50505539-50505539 on Build GRCh38
rs773625240 Chr.22: 50505421-50505421 on Build GRCh38
rs918831113 Chr.22: 50505518-50505518 on Build GRCh38
rs769097491 Chr.22: 50505580-50505580 on Build GRCh38
rs769544734 Chr.22: 50505458-50505459 on Build GRCh38
rs975756313 Chr.22: 50505468-50505468 on Build GRCh38
rs778667226 Chr.22: 50505493-50505493 on Build GRCh38
rs745355188 Chr.22: 50505560-50505560 on Build GRCh38
rs375949589 Chr.22: 50505511-50505511 on Build GRCh38
rs138639555 Chr.22: 50505424-50505424 on Build GRCh38
rs557647591 Chr.22: 50505482-50505482 on Build GRCh38
rs769193250 Chr.22: 50505445-50505445 on Build GRCh38
rs775576790 Chr.22: 50505470-50505470 on Build GRCh38
rs751493546 Chr.22: 50505483-50505483 on Build GRCh38
rs759462899 Chr.22: 50505531-50505531 on Build GRCh38
rs1054468850 Chr.22: 50505551-50505551 on Build GRCh38
rs753445391 Chr.22: 50505553-50505553 on Build GRCh38
rs550211846 Chr.22: 50505430-50505430 on Build GRCh38
rs374774405 Chr.22: 50505455-50505455 on Build GRCh38
rs748845188 Chr.22: 50505499-50505499 on Build GRCh38
rs774436301 Chr.22: 50505530-50505530 on Build GRCh38
rs369462014 Chr.22: 50505578-50505578 on Build GRCh38

View Full Product Details