Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 76611288-76611513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100133091
Entrez Gene ID
100133091
Gene Name
uncharacterized LOC100133091
Gene Aliases
-
UniGene
Hs.712837
Gene Chromosome Location
Chr.7: 76549341-76627982 on Build GRCh38
Transcripts
NR_029411.1 NM_012230.3 NM_152992.2

Gene Symbol
POMZP3
Entrez Gene ID
22932
Gene Name
POM121 and ZP3 fusion
Gene Aliases
POM-ZP3, POM121
UniGene
Hs.488877
Gene Chromosome Location
Chr.7: 76609986-76627303 on Build GRCh38
Transcripts
NR_029411.1 NM_012230.3 NM_152992.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4597920 c.548C>G p.T183R Chr.7: 76611481-76611481 on Build GRCh38
3995703 c.545C>A p.S182Y Chr.7: 76611484-76611484 on Build GRCh38
5446155 c.560delT p.L187fs? Chr.7: 76611469-76611469 on Build GRCh38
3942305 c.545C>G p.S182C Chr.7: 76611484-76611484 on Build GRCh38
7254655 c.527G>A p.R176H Chr.7: 76611502-76611502 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755485613 Chr.7: 76611422-76611422 on Build GRCh38
rs934831538 Chr.7: 76611401-76611401 on Build GRCh38
rs115806492 Chr.7: 76611481-76611481 on Build GRCh38
rs781652976 Chr.7: 76611429-76611429 on Build GRCh38
rs377228819 Chr.7: 76611463-76611463 on Build GRCh38
rs572353409 Chr.7: 76611336-76611336 on Build GRCh38
rs903311182 Chr.7: 76611393-76611393 on Build GRCh38
rs148426627 Chr.7: 76611410-76611410 on Build GRCh38
rs188172114 Chr.7: 76611436-76611436 on Build GRCh38
rs369030822 Chr.7: 76611480-76611480 on Build GRCh38
rs761452782 Chr.7: 76611467-76611467 on Build GRCh38
rs756586339 Chr.7: 76611489-76611489 on Build GRCh38
rs947486300 Chr.7: 76611376-76611376 on Build GRCh38
rs754811779 Chr.7: 76611426-76611427 on Build GRCh38
rs551999681 Chr.7: 76611492-76611492 on Build GRCh38
rs899591183 Chr.7: 76611465-76611465 on Build GRCh38
rs766010654 Chr.7: 76611475-76611475 on Build GRCh38
rs778658575 Chr.7: 76611468-76611468 on Build GRCh38
rs372058567 Chr.7: 76611437-76611437 on Build GRCh38
rs530701673 Chr.7: 76611486-76611486 on Build GRCh38
rs928560437 Chr.7: 76611325-76611325 on Build GRCh38
rs1043613797 Chr.7: 76611377-76611377 on Build GRCh38
rs991515947 Chr.7: 76611343-76611343 on Build GRCh38
rs915984281 Chr.7: 76611349-76611349 on Build GRCh38
rs751852019 Chr.7: 76611416-76611416 on Build GRCh38
rs183695161 Chr.7: 76611383-76611383 on Build GRCh38
rs71819724 Chr.7: 76611469-76611469 on Build GRCh38
rs554051207 Chr.7: 76611320-76611320 on Build GRCh38
rs367893419 Chr.7: 76611414-76611414 on Build GRCh38
rs570484973 Chr.7: 76611495-76611495 on Build GRCh38
rs938634372 Chr.7: 76611342-76611345 on Build GRCh38
rs995230184 Chr.7: 76611484-76611484 on Build GRCh38
rs762549256 Chr.7: 76611474-76611474 on Build GRCh38
rs73705106 Chr.7: 76611445-76611445 on Build GRCh38
rs17419421 Chr.7: 76611497-76611497 on Build GRCh38
rs766131453 Chr.7: 76611468-76611469 on Build GRCh38
rs779719441 Chr.7: 76611461-76611461 on Build GRCh38
rs772733313 Chr.7: 76611472-76611472 on Build GRCh38
rs752839795 Chr.7: 76611430-76611430 on Build GRCh38
rs758393686 Chr.7: 76611470-76611470 on Build GRCh38
rs982680096 Chr.7: 76611313-76611313 on Build GRCh38
rs192562971 Chr.7: 76611462-76611462 on Build GRCh38
rs866847986 Chr.7: 76611341-76611341 on Build GRCh38
rs756153920 Chr.7: 76611433-76611433 on Build GRCh38
rs73705107 Chr.7: 76611456-76611456 on Build GRCh38
rs1799211 Chr.7: 76611360-76611360 on Build GRCh38
rs769168525 Chr.7: 76611469-76611469 on Build GRCh38

View Full Product Details