Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 27621917-27622152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR1D
Entrez Gene ID
51082
Gene Name
RNA polymerase I subunit D
Gene Aliases
AC19, POLR1C, RPA16, RPA9, RPAC2, RPC16, RPO1-3, TCS2
UniGene
Hs.507584
Gene Chromosome Location
Chr.13: 27620743-27667422 on Build GRCh38
Transcripts
NM_001206559.1 NM_015972.3 NM_152705.2 XM_005266412.1 XM_005266414.2 XM_017020623.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6752303 c.4G>C p.E2Q Chr.13: 27621987-27621987 on Build GRCh38
6752302 c.4G>C p.E2Q Chr.13: 27621987-27621987 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs979645460 Chr.13: 27622117-27622117 on Build GRCh38
rs761964229 Chr.13: 27622014-27622014 on Build GRCh38
rs555333149 Chr.13: 27622106-27622106 on Build GRCh38
rs765066005 Chr.13: 27622038-27622038 on Build GRCh38
rs756859762 Chr.13: 27622061-27622061 on Build GRCh38
rs763822157 Chr.13: 27622055-27622055 on Build GRCh38
rs2232682 Chr.13: 27622067-27622067 on Build GRCh38
rs376051642 Chr.13: 27621989-27621989 on Build GRCh38
rs766253130 Chr.13: 27622015-27622015 on Build GRCh38
rs1011974460 Chr.13: 27621948-27621948 on Build GRCh38
rs566596016 Chr.13: 27622075-27622075 on Build GRCh38
rs559557492 Chr.13: 27621954-27621954 on Build GRCh38
rs200891022 Chr.13: 27621957-27621957 on Build GRCh38
rs375008435 Chr.13: 27621944-27621944 on Build GRCh38
rs988352057 Chr.13: 27622051-27622051 on Build GRCh38
rs975464233 Chr.13: 27622086-27622086 on Build GRCh38
rs2232681 Chr.13: 27622027-27622027 on Build GRCh38
rs766149327 Chr.13: 27622057-27622058 on Build GRCh38
rs938032639 Chr.13: 27622121-27622121 on Build GRCh38
rs964310242 Chr.13: 27621971-27621971 on Build GRCh38
rs550615488 Chr.13: 27622035-27622035 on Build GRCh38
rs1016945761 Chr.13: 27622050-27622050 on Build GRCh38
rs764374705 Chr.13: 27621945-27621945 on Build GRCh38
rs376899179 Chr.13: 27621934-27621934 on Build GRCh38
rs968874343 Chr.13: 27622107-27622107 on Build GRCh38
rs914676903 Chr.13: 27622104-27622104 on Build GRCh38
rs898299613 Chr.13: 27621962-27621962 on Build GRCh38
rs749220350 Chr.13: 27621937-27621937 on Build GRCh38
rs539572178 Chr.13: 27622046-27622046 on Build GRCh38
rs768779873 Chr.13: 27621984-27621984 on Build GRCh38
rs535641331 Chr.13: 27622084-27622084 on Build GRCh38
rs746196920 Chr.13: 27621968-27621968 on Build GRCh38
rs750987447 Chr.13: 27622058-27622058 on Build GRCh38
rs963052156 Chr.13: 27622062-27622062 on Build GRCh38
rs575511489 Chr.13: 27622119-27622119 on Build GRCh38
rs375054642 Chr.13: 27622041-27622041 on Build GRCh38
rs752330161 Chr.13: 27621945-27621945 on Build GRCh38
rs775835096 Chr.13: 27621982-27621982 on Build GRCh38
rs754119950 Chr.13: 27622019-27622021 on Build GRCh38
rs537023554 Chr.13: 27622125-27622125 on Build GRCh38
rs778495401 Chr.13: 27621941-27621941 on Build GRCh38
rs896236599 Chr.13: 27621935-27621935 on Build GRCh38
rs754787294 Chr.13: 27621939-27621939 on Build GRCh38
rs763146884 Chr.13: 27621983-27621983 on Build GRCh38
rs545965891 Chr.13: 27621950-27621950 on Build GRCh38
rs2232680 Chr.13: 27621963-27621963 on Build GRCh38
rs762755049 Chr.13: 27622034-27622053 on Build GRCh38

View Full Product Details