Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 53651510-53651767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL31
Entrez Gene ID
401265
Gene Name
kelch like family member 31
Gene Aliases
BKLHD6, KBTBD1, KLHL, bA345L23.2
UniGene
Hs.131064
Gene Chromosome Location
Chr.6: 53647901-53665854 on Build GRCh38
Transcripts
NM_001003760.4 XM_005249107.4 XM_017010858.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4185519 c.1771C>T p.L591F Chr.6: 53651732-53651732 on Build GRCh38
5429253 c.1791C>T p.D597D Chr.6: 53651712-53651712 on Build GRCh38
5519986 c.1789G>T p.D597Y Chr.6: 53651714-53651714 on Build GRCh38
4441824 c.1784C>T p.T595M Chr.6: 53651719-53651719 on Build GRCh38
6696697 c.1854G>A p.V618V Chr.6: 53651649-53651649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766567147 Chr.6: 53651671-53651671 on Build GRCh38
rs753026142 Chr.6: 53651610-53651610 on Build GRCh38
rs768692785 Chr.6: 53651648-53651648 on Build GRCh38
rs759648214 Chr.6: 53651699-53651699 on Build GRCh38
rs374819778 Chr.6: 53651713-53651713 on Build GRCh38
rs142726111 Chr.6: 53651718-53651718 on Build GRCh38
rs767969108 Chr.6: 53651703-53651703 on Build GRCh38
rs904371789 Chr.6: 53651587-53651587 on Build GRCh38
rs756499205 Chr.6: 53651616-53651616 on Build GRCh38
rs202117315 Chr.6: 53651666-53651666 on Build GRCh38
rs761750875 Chr.6: 53651565-53651565 on Build GRCh38
rs142874521 Chr.6: 53651667-53651667 on Build GRCh38
rs750678659 Chr.6: 53651593-53651593 on Build GRCh38
rs769724245 Chr.6: 53651662-53651662 on Build GRCh38
rs1015840568 Chr.6: 53651741-53651741 on Build GRCh38
rs996899188 Chr.6: 53651695-53651695 on Build GRCh38
rs935309913 Chr.6: 53651692-53651692 on Build GRCh38
rs12204882 Chr.6: 53651620-53651620 on Build GRCh38
rs1010958646 Chr.6: 53651720-53651720 on Build GRCh38
rs779622806 Chr.6: 53651719-53651719 on Build GRCh38
rs779614575 Chr.6: 53651640-53651640 on Build GRCh38
rs751632062 Chr.6: 53651696-53651696 on Build GRCh38
rs115408188 Chr.6: 53651645-53651645 on Build GRCh38
rs745549645 Chr.6: 53651630-53651630 on Build GRCh38
rs746498141 Chr.6: 53651726-53651726 on Build GRCh38
rs768315149 Chr.6: 53651730-53651730 on Build GRCh38
rs766483858 Chr.6: 53651597-53651597 on Build GRCh38
rs778491234 Chr.6: 53651625-53651625 on Build GRCh38
rs371428644 Chr.6: 53651552-53651552 on Build GRCh38
rs761662714 Chr.6: 53651654-53651654 on Build GRCh38
rs891740184 Chr.6: 53651629-53651629 on Build GRCh38
rs771420967 Chr.6: 53651633-53651633 on Build GRCh38
rs1044230172 Chr.6: 53651584-53651584 on Build GRCh38
rs927983218 Chr.6: 53651682-53651682 on Build GRCh38
rs776751723 Chr.6: 53651652-53651652 on Build GRCh38
rs751656798 Chr.6: 53651599-53651599 on Build GRCh38
rs952913141 Chr.6: 53651728-53651728 on Build GRCh38
rs12204875 Chr.6: 53651601-53651601 on Build GRCh38
rs200886597 Chr.6: 53651740-53651740 on Build GRCh38
rs12204835 Chr.6: 53651554-53651554 on Build GRCh38
rs199925517 Chr.6: 53651709-53651709 on Build GRCh38
rs369927487 Chr.6: 53651606-53651606 on Build GRCh38
rs139251902 Chr.6: 53651583-53651583 on Build GRCh38

View Full Product Details