Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 4098899-4099142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf201
Entrez Gene ID
404220
Gene Name
chromosome 6 open reading frame 201
Gene Aliases
dJ1013A10.5
UniGene
Hs.716731
Gene Chromosome Location
Chr.6: 4079197-4130788 on Build GRCh38
Transcripts
NM_001085401.2 NR_104463.1 NR_104464.1 XM_005249112.3 XM_011514613.2 XM_017010861.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1079143 c.107G>A p.G36D Chr.6: 4099021-4099021 on Build GRCh38
5667220 c.192A>G p.R64R Chr.6: 4099106-4099106 on Build GRCh38
1079144 c.183G>A p.P61P Chr.6: 4099097-4099097 on Build GRCh38
5906768 c.217G>A p.G73R Chr.6: 4099131-4099131 on Build GRCh38
3627131 c.97G>A p.E33K Chr.6: 4099002-4099002 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371231647 Chr.6: 4099112-4099112 on Build GRCh38
rs754957660 Chr.6: 4099049-4099049 on Build GRCh38
rs916207792 Chr.6: 4099024-4099024 on Build GRCh38
rs147712155 Chr.6: 4099097-4099097 on Build GRCh38
rs766478648 Chr.6: 4099043-4099043 on Build GRCh38
rs770655617 Chr.6: 4099091-4099091 on Build GRCh38
rs995715959 Chr.6: 4098966-4098966 on Build GRCh38
rs780925502 Chr.6: 4099052-4099052 on Build GRCh38
rs200932959 Chr.6: 4099092-4099092 on Build GRCh38
rs773692720 Chr.6: 4098986-4098986 on Build GRCh38
rs770604311 Chr.6: 4098980-4098980 on Build GRCh38
rs765632980 Chr.6: 4099101-4099101 on Build GRCh38
rs756022320 Chr.6: 4099075-4099075 on Build GRCh38
rs551522599 Chr.6: 4098994-4098994 on Build GRCh38
rs762289377 Chr.6: 4099099-4099099 on Build GRCh38
rs374300441 Chr.6: 4099081-4099081 on Build GRCh38
rs990462148 Chr.6: 4099020-4099020 on Build GRCh38
rs370237610 Chr.6: 4098948-4098948 on Build GRCh38
rs145554891 Chr.6: 4099096-4099096 on Build GRCh38
rs946466357 Chr.6: 4099029-4099029 on Build GRCh38
rs78486783 Chr.6: 4099008-4099008 on Build GRCh38
rs755079425 Chr.6: 4099116-4099116 on Build GRCh38
rs751546146 Chr.6: 4099045-4099045 on Build GRCh38
rs573260688 Chr.6: 4099115-4099115 on Build GRCh38
rs748978684 Chr.6: 4099082-4099082 on Build GRCh38
rs763669845 Chr.6: 4099091-4099092 on Build GRCh38
rs748102739 Chr.6: 4099057-4099057 on Build GRCh38
rs570040610 Chr.6: 4099072-4099072 on Build GRCh38
rs773056278 Chr.6: 4098961-4098961 on Build GRCh38
rs201992367 Chr.6: 4099106-4099106 on Build GRCh38
rs750696379 Chr.6: 4099018-4099018 on Build GRCh38
rs764844081 Chr.6: 4098988-4098988 on Build GRCh38
rs182870741 Chr.6: 4098992-4098992 on Build GRCh38
rs762236327 Chr.6: 4099013-4099013 on Build GRCh38
rs752182631 Chr.6: 4099055-4099055 on Build GRCh38
rs773528326 Chr.6: 4099105-4099105 on Build GRCh38
rs763218543 Chr.6: 4099113-4099113 on Build GRCh38
rs745305729 Chr.6: 4099093-4099093 on Build GRCh38
rs762672057 Chr.6: 4098974-4098974 on Build GRCh38
rs116327806 Chr.6: 4098959-4098959 on Build GRCh38
rs370584916 Chr.6: 4099014-4099014 on Build GRCh38
rs537302775 Chr.6: 4099034-4099034 on Build GRCh38
rs1043475849 Chr.6: 4099069-4099069 on Build GRCh38
rs867115771 Chr.6: 4099087-4099087 on Build GRCh38
rs757879850 Chr.6: 4099072-4099072 on Build GRCh38

View Full Product Details