Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52814194-52814419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITIH4
Entrez Gene ID
3700
Gene Name
inter-alpha-trypsin inhibitor heavy chain family member 4
Gene Aliases
GP120, H4P, IHRP, ITI-HC4, ITIHL1, PK-120, PK120
UniGene
Hs.709406
Gene Chromosome Location
Chr.3: 52812990-52830701 on Build GRCh38
Transcripts
NM_001166449.1 NM_002218.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4482137 c.2577C>T p.L859L Chr.3: 52814258-52814258 on Build GRCh38
3916412 c.2539C>T p.L847L Chr.3: 52814296-52814296 on Build GRCh38
6688431 c.2562G>T p.G854G Chr.3: 52814273-52814273 on Build GRCh38
4538350 c.2488G>A p.G830S Chr.3: 52814299-52814299 on Build GRCh38
4482149 c.2530C>T p.L844F Chr.3: 52814257-52814257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775598398 Chr.3: 52814354-52814354 on Build GRCh38
rs529775134 Chr.3: 52814315-52814315 on Build GRCh38
rs201607989 Chr.3: 52814250-52814250 on Build GRCh38
rs139369931 Chr.3: 52814277-52814277 on Build GRCh38
rs545979942 Chr.3: 52814253-52814253 on Build GRCh38
rs150977188 Chr.3: 52814340-52814340 on Build GRCh38
rs746076241 Chr.3: 52814368-52814372 on Build GRCh38
rs187101592 Chr.3: 52814310-52814310 on Build GRCh38
rs762247958 Chr.3: 52814379-52814379 on Build GRCh38
rs779379417 Chr.3: 52814238-52814238 on Build GRCh38
rs753524057 Chr.3: 52814382-52814382 on Build GRCh38
rs368376301 Chr.3: 52814239-52814239 on Build GRCh38
rs200311855 Chr.3: 52814371-52814371 on Build GRCh38
rs144664508 Chr.3: 52814283-52814283 on Build GRCh38
rs780892957 Chr.3: 52814397-52814397 on Build GRCh38
rs751509504 Chr.3: 52814395-52814395 on Build GRCh38
rs774509643 Chr.3: 52814378-52814378 on Build GRCh38
rs748153261 Chr.3: 52814251-52814251 on Build GRCh38
rs758345199 Chr.3: 52814388-52814388 on Build GRCh38
rs147753898 Chr.3: 52814278-52814278 on Build GRCh38
rs752419066 Chr.3: 52814345-52814351 on Build GRCh38
rs764513339 Chr.3: 52814223-52814223 on Build GRCh38
rs141684429 Chr.3: 52814270-52814270 on Build GRCh38
rs754802041 Chr.3: 52814246-52814246 on Build GRCh38
rs148943741 Chr.3: 52814285-52814285 on Build GRCh38
rs754494907 Chr.3: 52814300-52814300 on Build GRCh38
rs769838238 Chr.3: 52814347-52814347 on Build GRCh38
rs778965763 Chr.3: 52814365-52814369 on Build GRCh38
rs540681269 Chr.3: 52814302-52814302 on Build GRCh38
rs202071209 Chr.3: 52814299-52814299 on Build GRCh38
rs759301333 Chr.3: 52814384-52814384 on Build GRCh38
rs781170365 Chr.3: 52814253-52814255 on Build GRCh38
rs758457773 Chr.3: 52814314-52814314 on Build GRCh38
rs766970363 Chr.3: 52814272-52814272 on Build GRCh38
rs567867066 Chr.3: 52814339-52814339 on Build GRCh38
rs911526468 Chr.3: 52814233-52814233 on Build GRCh38
rs557292280 Chr.3: 52814224-52814224 on Build GRCh38
rs766057859 Chr.3: 52814297-52814297 on Build GRCh38
rs758600963 Chr.3: 52814371-52814372 on Build GRCh38
rs755928912 Chr.3: 52814363-52814364 on Build GRCh38
rs746827018 Chr.3: 52814318-52814318 on Build GRCh38
rs755848212 Chr.3: 52814225-52814225 on Build GRCh38
rs757072004 Chr.3: 52814396-52814396 on Build GRCh38
rs781553723 Chr.3: 52814289-52814289 on Build GRCh38
rs1037291826 Chr.3: 52814255-52814255 on Build GRCh38
rs763405177 Chr.3: 52814369-52814369 on Build GRCh38
rs112678571 Chr.3: 52814365-52814365 on Build GRCh38
rs145623288 Chr.3: 52814385-52814385 on Build GRCh38
rs755679969 Chr.3: 52814288-52814288 on Build GRCh38
rs1026926289 Chr.3: 52814274-52814274 on Build GRCh38
rs146210920 Chr.3: 52814262-52814262 on Build GRCh38
rs771217141 Chr.3: 52814263-52814263 on Build GRCh38
rs767826935 Chr.3: 52814380-52814380 on Build GRCh38

View Full Product Details