Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 114397809-114398066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM33
Entrez Gene ID
51592
Gene Name
tripartite motif containing 33
Gene Aliases
ECTO, PTC7, RFG7, TF1G, TIF1G, TIF1GAMMA, TIFGAMMA
UniGene
Hs.26837
Gene Chromosome Location
Chr.1: 114392777-114511160 on Build GRCh38
Transcripts
NM_015906.3 NM_033020.2 XM_005270936.3 XM_005270937.3 XM_011541568.2 XM_017001452.1 XM_017001453.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5016741 c.2442_2443insA p.I815fs*2 Chr.1: 114397950-114397951 on Build GRCh38
6120479 c.3126G>A p.M1042I Chr.1: 114397985-114397985 on Build GRCh38
3801349 c.3190C>T p.Q1064* Chr.1: 114397842-114397842 on Build GRCh38
35460 c.3169A>T p.K1057* Chr.1: 114397942-114397942 on Build GRCh38
4769480 c.3123G>A p.M1041I Chr.1: 114397988-114397988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756034591 Chr.1: 114398010-114398011 on Build GRCh38
rs559109821 Chr.1: 114398011-114398012 on Build GRCh38
rs756712386 Chr.1: 114398026-114398026 on Build GRCh38
rs895436037 Chr.1: 114397919-114397919 on Build GRCh38
rs774852932 Chr.1: 114398001-114398001 on Build GRCh38
rs78184484 Chr.1: 114398010-114398010 on Build GRCh38
rs769444726 Chr.1: 114398023-114398023 on Build GRCh38
rs748272280 Chr.1: 114397839-114397839 on Build GRCh38
rs771490153 Chr.1: 114397955-114397956 on Build GRCh38
rs750990577 Chr.1: 114398021-114398021 on Build GRCh38
rs902232225 Chr.1: 114397982-114397982 on Build GRCh38
rs397981357 Chr.1: 114398022-114398022 on Build GRCh38
rs5777192 Chr.1: 114398011-114398011 on Build GRCh38
rs79284281 Chr.1: 114398025-114398025 on Build GRCh38
rs75496873 Chr.1: 114398022-114398022 on Build GRCh38
rs773262839 Chr.1: 114397889-114397889 on Build GRCh38
rs764409725 Chr.1: 114398011-114398014 on Build GRCh38
rs573450786 Chr.1: 114398012-114398012 on Build GRCh38
rs764887317 Chr.1: 114397988-114397988 on Build GRCh38
rs1037971330 Chr.1: 114398022-114398023 on Build GRCh38
rs1011450420 Chr.1: 114397948-114397948 on Build GRCh38
rs75435059 Chr.1: 114398021-114398022 on Build GRCh38
rs867337307 Chr.1: 114397965-114397965 on Build GRCh38
rs746772540 Chr.1: 114397901-114397901 on Build GRCh38
rs369975579 Chr.1: 114397873-114397873 on Build GRCh38
rs776444701 Chr.1: 114397918-114397918 on Build GRCh38
rs770760015 Chr.1: 114397913-114397913 on Build GRCh38
rs774103641 Chr.1: 114397875-114397876 on Build GRCh38
rs1043907251 Chr.1: 114397981-114397981 on Build GRCh38
rs759124781 Chr.1: 114397934-114397934 on Build GRCh38
rs370849641 Chr.1: 114398016-114398016 on Build GRCh38
rs762591563 Chr.1: 114398003-114398003 on Build GRCh38
rs767358049 Chr.1: 114398011-114398013 on Build GRCh38

View Full Product Details