Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 149909684-149909953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SV2A
Entrez Gene ID
9900
Gene Name
synaptic vesicle glycoprotein 2A
Gene Aliases
SV2
UniGene
Hs.516153
Gene Chromosome Location
Chr.1: 149903318-149917882 on Build GRCh38
Transcripts
NM_014849.4 XM_017002992.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5041879 c.1179+1G>A p.? Chr.1: 149909800-149909800 on Build GRCh38
5040695 c.1179+2T>G p.? Chr.1: 149909799-149909799 on Build GRCh38
267534 c.1168C>T p.R390* Chr.1: 149909812-149909812 on Build GRCh38
6793498 c.1147C>T p.R383* Chr.1: 149909833-149909833 on Build GRCh38
81779 c.1179A>C p.S393S Chr.1: 149909801-149909801 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781885911 Chr.1: 149909922-149909922 on Build GRCh38
rs587652420 Chr.1: 149909720-149909720 on Build GRCh38
rs141726737 Chr.1: 149909846-149909846 on Build GRCh38
rs781813901 Chr.1: 149909811-149909811 on Build GRCh38
rs782808009 Chr.1: 149909803-149909803 on Build GRCh38
rs782478152 Chr.1: 149909812-149909812 on Build GRCh38
rs1057022025 Chr.1: 149909758-149909758 on Build GRCh38
rs782142893 Chr.1: 149909792-149909792 on Build GRCh38
rs782626021 Chr.1: 149909861-149909861 on Build GRCh38
rs782213216 Chr.1: 149909760-149909760 on Build GRCh38
rs781998027 Chr.1: 149909788-149909788 on Build GRCh38
rs587718420 Chr.1: 149909906-149909906 on Build GRCh38
rs146579446 Chr.1: 149909833-149909833 on Build GRCh38
rs782745537 Chr.1: 149909820-149909820 on Build GRCh38
rs782038562 Chr.1: 149909773-149909773 on Build GRCh38
rs782396633 Chr.1: 149909882-149909882 on Build GRCh38
rs782634096 Chr.1: 149909767-149909767 on Build GRCh38
rs367556446 Chr.1: 149909902-149909902 on Build GRCh38
rs782212250 Chr.1: 149909853-149909853 on Build GRCh38
rs782318059 Chr.1: 149909782-149909782 on Build GRCh38
rs782801711 Chr.1: 149909917-149909917 on Build GRCh38
rs782581361 Chr.1: 149909852-149909852 on Build GRCh38
rs201829805 Chr.1: 149909894-149909894 on Build GRCh38
rs587703410 Chr.1: 149909726-149909726 on Build GRCh38
rs782031845 Chr.1: 149909883-149909883 on Build GRCh38
rs782676683 Chr.1: 149909834-149909834 on Build GRCh38
rs781862859 Chr.1: 149909829-149909829 on Build GRCh38
rs927027823 Chr.1: 149909710-149909710 on Build GRCh38
rs782514088 Chr.1: 149909832-149909832 on Build GRCh38
rs782252414 Chr.1: 149909873-149909873 on Build GRCh38
rs1030557307 Chr.1: 149909847-149909847 on Build GRCh38
rs938446130 Chr.1: 149909747-149909747 on Build GRCh38
rs782563008 Chr.1: 149909768-149909768 on Build GRCh38
rs199766170 Chr.1: 149909762-149909762 on Build GRCh38
rs781910016 Chr.1: 149909845-149909845 on Build GRCh38
rs782047886 Chr.1: 149909884-149909884 on Build GRCh38
rs782598833 Chr.1: 149909755-149909755 on Build GRCh38
rs372328650 Chr.1: 149909903-149909903 on Build GRCh38
rs782549392 Chr.1: 149909752-149909752 on Build GRCh38
rs896676867 Chr.1: 149909759-149909759 on Build GRCh38
rs782775615 Chr.1: 149909787-149909787 on Build GRCh38

View Full Product Details