Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66846921-66847150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA7
Entrez Gene ID
766
Gene Name
carbonic anhydrase 7
Gene Aliases
CAVII
UniGene
Hs.37014
Gene Chromosome Location
Chr.16: 66844379-66854149 on Build GRCh38
Transcripts
NM_001014435.1 NM_005182.2 XM_005256136.3 XM_011523311.1 XM_011523312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4464141 c.131C>T p.P44L Chr.16: 66847120-66847120 on Build GRCh38
972224 c.86G>A p.R29H Chr.16: 66847075-66847075 on Build GRCh38
4515146 c.99C>T p.I33I Chr.16: 66847088-66847088 on Build GRCh38
1709270 c.67C>T p.P23S Chr.16: 66847056-66847056 on Build GRCh38
703925 c.110C>A p.S37Y Chr.16: 66847099-66847099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529857078 Chr.16: 66846955-66846955 on Build GRCh38
rs991475032 Chr.16: 66846975-66846975 on Build GRCh38
rs538115679 Chr.16: 66846960-66846960 on Build GRCh38
rs776229316 Chr.16: 66847124-66847124 on Build GRCh38
rs376478290 Chr.16: 66847000-66847000 on Build GRCh38
rs924435928 Chr.16: 66847119-66847119 on Build GRCh38
rs774537213 Chr.16: 66847088-66847088 on Build GRCh38
rs575554312 Chr.16: 66846957-66846957 on Build GRCh38
rs758044639 Chr.16: 66847002-66847002 on Build GRCh38
rs117738342 Chr.16: 66847054-66847054 on Build GRCh38
rs781369944 Chr.16: 66847037-66847037 on Build GRCh38
rs756990569 Chr.16: 66847036-66847036 on Build GRCh38
rs749366504 Chr.16: 66847059-66847059 on Build GRCh38
rs1016958809 Chr.16: 66846971-66846971 on Build GRCh38
rs770143403 Chr.16: 66847052-66847052 on Build GRCh38
rs374514983 Chr.16: 66847021-66847021 on Build GRCh38
rs373632795 Chr.16: 66846995-66846995 on Build GRCh38
rs796459854 Chr.16: 66846972-66846972 on Build GRCh38
rs777536968 Chr.16: 66847015-66847015 on Build GRCh38
rs868323251 Chr.16: 66847074-66847074 on Build GRCh38
rs746250502 Chr.16: 66847049-66847049 on Build GRCh38
rs761977983 Chr.16: 66847107-66847107 on Build GRCh38
rs577842491 Chr.16: 66847085-66847085 on Build GRCh38
rs759322502 Chr.16: 66846987-66846987 on Build GRCh38
rs199547005 Chr.16: 66847118-66847118 on Build GRCh38
rs951476710 Chr.16: 66846950-66846951 on Build GRCh38

View Full Product Details