Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186161524-186161753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKG
Entrez Gene ID
1608
Gene Name
diacylglycerol kinase gamma
Gene Aliases
DAGK3, DGK-GAMMA
UniGene
Hs.683449
Gene Chromosome Location
Chr.3: 186147201-186362234 on Build GRCh38
Transcripts
NM_001080744.1 NM_001080745.1 NM_001346.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1042273 c.2265G>T p.Q755H Chr.3: 186161615-186161615 on Build GRCh38
4795478 c.2242G>A p.V748M Chr.3: 186161638-186161638 on Build GRCh38
1308918 c.2236A>G p.M746V Chr.3: 186161644-186161644 on Build GRCh38
6642993 c.2259G>C p.W753C Chr.3: 186161621-186161621 on Build GRCh38
6836245 c.2217-1G>T p.? Chr.3: 186161664-186161664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375249689 Chr.3: 186161589-186161589 on Build GRCh38
rs758548727 Chr.3: 186161623-186161623 on Build GRCh38
rs375221556 Chr.3: 186161694-186161694 on Build GRCh38
rs772032337 Chr.3: 186161559-186161559 on Build GRCh38
rs780032513 Chr.3: 186161644-186161644 on Build GRCh38
rs747204887 Chr.3: 186161573-186161573 on Build GRCh38
rs773816242 Chr.3: 186161679-186161679 on Build GRCh38
rs374848985 Chr.3: 186161672-186161672 on Build GRCh38
rs768962612 Chr.3: 186161579-186161579 on Build GRCh38
rs756842630 Chr.3: 186161715-186161716 on Build GRCh38
rs758979861 Chr.3: 186161702-186161702 on Build GRCh38
rs893813438 Chr.3: 186161726-186161726 on Build GRCh38
rs763335483 Chr.3: 186161685-186161685 on Build GRCh38
rs368878566 Chr.3: 186161678-186161678 on Build GRCh38
rs747040955 Chr.3: 186161657-186161657 on Build GRCh38
rs775834157 Chr.3: 186161572-186161572 on Build GRCh38
rs199753031 Chr.3: 186161604-186161604 on Build GRCh38
rs367877531 Chr.3: 186161598-186161598 on Build GRCh38
rs372243453 Chr.3: 186161691-186161691 on Build GRCh38
rs149298788 Chr.3: 186161603-186161603 on Build GRCh38
rs747943570 Chr.3: 186161697-186161697 on Build GRCh38
rs150593644 Chr.3: 186161720-186161720 on Build GRCh38
rs755898523 Chr.3: 186161614-186161614 on Build GRCh38
rs761067653 Chr.3: 186161596-186161596 on Build GRCh38
rs1033508044 Chr.3: 186161714-186161714 on Build GRCh38
rs769009556 Chr.3: 186161663-186161663 on Build GRCh38
rs370807288 Chr.3: 186161583-186161583 on Build GRCh38
rs777154847 Chr.3: 186161582-186161582 on Build GRCh38
rs767100068 Chr.3: 186161609-186161609 on Build GRCh38
rs776818637 Chr.3: 186161677-186161677 on Build GRCh38
rs766929157 Chr.3: 186161612-186161612 on Build GRCh38
rs371814072 Chr.3: 186161618-186161618 on Build GRCh38
rs777824962 Chr.3: 186161615-186161615 on Build GRCh38
rs762581170 Chr.3: 186161600-186161600 on Build GRCh38
rs973070214 Chr.3: 186161556-186161556 on Build GRCh38

View Full Product Details