Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136941130-136941389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW5
Entrez Gene ID
54461
Gene Name
F-box and WD repeat domain containing 5
Gene Aliases
Fbw5
UniGene
Hs.522507
Gene Chromosome Location
Chr.9: 136940433-136944774 on Build GRCh38
Transcripts
NM_018998.3 XM_005266089.2 XM_011518789.1 XM_017014812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5732425 c.1459G>A p.G487R Chr.9: 136941170-136941170 on Build GRCh38
6662470 c.1346A>G p.D449G Chr.9: 136941362-136941362 on Build GRCh38
5652700 c.1362G>T p.R454R Chr.9: 136941346-136941346 on Build GRCh38
5626517 c.1449C>T p.F483F Chr.9: 136941259-136941259 on Build GRCh38
6662461 c.1390C>T p.R464C Chr.9: 136941318-136941318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753630793 Chr.9: 136941213-136941213 on Build GRCh38
rs777973791 Chr.9: 136941370-136941370 on Build GRCh38
rs201822750 Chr.9: 136941335-136941335 on Build GRCh38
rs765978255 Chr.9: 136941298-136941298 on Build GRCh38
rs745525167 Chr.9: 136941342-136941342 on Build GRCh38
rs760331772 Chr.9: 136941297-136941297 on Build GRCh38
rs138273782 Chr.9: 136941350-136941350 on Build GRCh38
rs371147879 Chr.9: 136941318-136941318 on Build GRCh38
rs751445990 Chr.9: 136941308-136941308 on Build GRCh38
rs144664027 Chr.9: 136941200-136941200 on Build GRCh38
rs760258773 Chr.9: 136941258-136941258 on Build GRCh38
rs746105556 Chr.9: 136941236-136941236 on Build GRCh38
rs367827758 Chr.9: 136941274-136941274 on Build GRCh38
rs772320395 Chr.9: 136941330-136941330 on Build GRCh38
rs750083255 Chr.9: 136941179-136941179 on Build GRCh38
rs111761422 Chr.9: 136941336-136941336 on Build GRCh38
rs370227114 Chr.9: 136941239-136941239 on Build GRCh38
rs779965747 Chr.9: 136941181-136941181 on Build GRCh38
rs759237517 Chr.9: 136941301-136941301 on Build GRCh38
rs369668241 Chr.9: 136941165-136941165 on Build GRCh38
rs372873658 Chr.9: 136941317-136941317 on Build GRCh38
rs367798532 Chr.9: 136941241-136941241 on Build GRCh38
rs748748915 Chr.9: 136941193-136941194 on Build GRCh38
rs762358306 Chr.9: 136941242-136941242 on Build GRCh38
rs750085450 Chr.9: 136941209-136941209 on Build GRCh38
rs374757784 Chr.9: 136941273-136941273 on Build GRCh38
rs201429106 Chr.9: 136941364-136941364 on Build GRCh38
rs370826295 Chr.9: 136941224-136941224 on Build GRCh38
rs377369299 Chr.9: 136941201-136941201 on Build GRCh38
rs200276837 Chr.9: 136941300-136941300 on Build GRCh38
rs376123602 Chr.9: 136941352-136941352 on Build GRCh38
rs755144335 Chr.9: 136941322-136941322 on Build GRCh38
rs756947005 Chr.9: 136941170-136941170 on Build GRCh38
rs753248943 Chr.9: 136941159-136941159 on Build GRCh38
rs768666071 Chr.9: 136941183-136941183 on Build GRCh38
rs759785463 Chr.9: 136941190-136941190 on Build GRCh38
rs748480629 Chr.9: 136941327-136941327 on Build GRCh38
rs768595180 Chr.9: 136941198-136941199 on Build GRCh38
rs761000831 Chr.9: 136941152-136941152 on Build GRCh38
rs755753376 Chr.9: 136941211-136941211 on Build GRCh38
rs748047155 Chr.9: 136941224-136941240 on Build GRCh38
rs780321428 Chr.9: 136941279-136941279 on Build GRCh38
rs764654690 Chr.9: 136941262-136941262 on Build GRCh38
rs372912376 Chr.9: 136941153-136941153 on Build GRCh38
rs1057018926 Chr.9: 136941250-136941250 on Build GRCh38
rs777063269 Chr.9: 136941155-136941155 on Build GRCh38
rs761706594 Chr.9: 136941353-136941353 on Build GRCh38
rs769950338 Chr.9: 136941237-136941237 on Build GRCh38
rs527442645 Chr.9: 136941157-136941157 on Build GRCh38
rs765104564 Chr.9: 136941266-136941266 on Build GRCh38
rs770610292 Chr.9: 136941341-136941341 on Build GRCh38
rs754882485 Chr.9: 136941264-136941264 on Build GRCh38
rs774208303 Chr.9: 136941223-136941224 on Build GRCh38
rs758849693 Chr.9: 136941368-136941368 on Build GRCh38
rs535554815 Chr.9: 136941204-136941204 on Build GRCh38
rs373520014 Chr.9: 136941171-136941171 on Build GRCh38
rs370273990 Chr.9: 136941205-136941205 on Build GRCh38
rs766063269 Chr.9: 136941259-136941259 on Build GRCh38
rs376036392 Chr.9: 136941158-136941158 on Build GRCh38
rs868572748 Chr.9: 136941168-136941168 on Build GRCh38
rs757637555 Chr.9: 136941316-136941316 on Build GRCh38
rs772731220 Chr.9: 136941246-136941246 on Build GRCh38
rs555399084 Chr.9: 136941208-136941208 on Build GRCh38
rs138768587 Chr.9: 136941307-136941307 on Build GRCh38
rs541956434 Chr.9: 136941344-136941344 on Build GRCh38
rs930596842 Chr.9: 136941222-136941222 on Build GRCh38
rs149358178 Chr.9: 136941197-136941197 on Build GRCh38
rs201352991 Chr.9: 136941184-136941184 on Build GRCh38
rs372563353 Chr.9: 136941289-136941289 on Build GRCh38
rs377761216 Chr.9: 136941182-136941182 on Build GRCh38
rs376136024 Chr.9: 136941355-136941355 on Build GRCh38
rs776963435 Chr.9: 136941286-136941288 on Build GRCh38
rs143554180 Chr.9: 136941288-136941288 on Build GRCh38
rs752620281 Chr.9: 136941269-136941269 on Build GRCh38
rs964886549 Chr.9: 136941265-136941265 on Build GRCh38
rs375678798 Chr.9: 136941225-136941225 on Build GRCh38
rs752483874 Chr.9: 136941303-136941303 on Build GRCh38
rs759649346 Chr.9: 136941302-136941303 on Build GRCh38
rs771754193 Chr.9: 136941252-136941257 on Build GRCh38
rs776777980 Chr.9: 136941186-136941186 on Build GRCh38
rs766968739 Chr.9: 136941245-136941245 on Build GRCh38
rs781297662 Chr.9: 136941232-136941232 on Build GRCh38
rs779572531 Chr.9: 136941180-136941181 on Build GRCh38
rs769055293 Chr.9: 136941283-136941283 on Build GRCh38
rs772603230 Chr.9: 136941292-136941292 on Build GRCh38
rs148314608 Chr.9: 136941326-136941326 on Build GRCh38
rs763621568 Chr.9: 136941166-136941166 on Build GRCh38
rs199873362 Chr.9: 136941363-136941363 on Build GRCh38
rs757333173 Chr.9: 136941231-136941231 on Build GRCh38
rs749499384 Chr.9: 136941280-136941280 on Build GRCh38
rs769425591 Chr.9: 136941348-136941348 on Build GRCh38
rs774739687 Chr.9: 136941285-136941285 on Build GRCh38
rs755917999 Chr.9: 136941180-136941180 on Build GRCh38
rs374042401 Chr.9: 136941207-136941207 on Build GRCh38
rs745899081 Chr.9: 136941275-136941275 on Build GRCh38
rs561808530 Chr.9: 136941347-136941347 on Build GRCh38
rs375317859 Chr.9: 136941240-136941240 on Build GRCh38
rs780017206 Chr.9: 136941212-136941212 on Build GRCh38
rs756329824 Chr.9: 136941278-136941278 on Build GRCh38
rs377703274 Chr.9: 136941358-136941358 on Build GRCh38
rs752769641 Chr.9: 136941196-136941196 on Build GRCh38
rs751018678 Chr.9: 136941169-136941169 on Build GRCh38
rs367852952 Chr.9: 136941217-136941217 on Build GRCh38

View Full Product Details