Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63999583-63999814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MACROD1
Entrez Gene ID
28992
Gene Name
MACRO domain containing 1
Gene Aliases
LRP16
UniGene
Hs.602898
Gene Chromosome Location
Chr.11: 63998554-64166652 on Build GRCh38
Transcripts
NM_014067.3 XM_005273939.4 XM_011544970.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6705781 c.708C>T p.S236S Chr.11: 63999720-63999720 on Build GRCh38
3687464 c.700G>C p.E234Q Chr.11: 63999728-63999728 on Build GRCh38
6705783 c.665-5_665-4insC p.? Chr.11: 63999767-63999768 on Build GRCh38
6419777 c.737G>A p.C246Y Chr.11: 63999691-63999691 on Build GRCh38
4632566 c.696C>T p.Y232Y Chr.11: 63999732-63999732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765810583 Chr.11: 63999645-63999645 on Build GRCh38
rs1013338843 Chr.11: 63999769-63999769 on Build GRCh38
rs757510449 Chr.11: 63999733-63999733 on Build GRCh38
rs375200081 Chr.11: 63999662-63999662 on Build GRCh38
rs777561126 Chr.11: 63999729-63999729 on Build GRCh38
rs202221368 Chr.11: 63999755-63999755 on Build GRCh38
rs527433190 Chr.11: 63999686-63999686 on Build GRCh38
rs751418124 Chr.11: 63999731-63999731 on Build GRCh38
rs951360087 Chr.11: 63999796-63999796 on Build GRCh38
rs759728025 Chr.11: 63999689-63999691 on Build GRCh38
rs1044802551 Chr.11: 63999773-63999773 on Build GRCh38
rs764414428 Chr.11: 63999631-63999631 on Build GRCh38
rs74751881 Chr.11: 63999624-63999624 on Build GRCh38
rs771288945 Chr.11: 63999660-63999660 on Build GRCh38
rs891363476 Chr.11: 63999797-63999797 on Build GRCh38
rs763342969 Chr.11: 63999629-63999629 on Build GRCh38
rs747233402 Chr.11: 63999657-63999657 on Build GRCh38
rs760059860 Chr.11: 63999615-63999615 on Build GRCh38
rs11600062 Chr.11: 63999777-63999777 on Build GRCh38
rs776247294 Chr.11: 63999789-63999789 on Build GRCh38
rs993411343 Chr.11: 63999706-63999708 on Build GRCh38
rs1026633094 Chr.11: 63999790-63999790 on Build GRCh38
rs750063535 Chr.11: 63999635-63999635 on Build GRCh38
rs752669675 Chr.11: 63999720-63999720 on Build GRCh38
rs771123725 Chr.11: 63999606-63999606 on Build GRCh38
rs201232991 Chr.11: 63999607-63999607 on Build GRCh38
rs754863055 Chr.11: 63999649-63999649 on Build GRCh38
rs1026592749 Chr.11: 63999713-63999713 on Build GRCh38
rs709594 Chr.11: 63999714-63999714 on Build GRCh38
rs760263385 Chr.11: 63999705-63999705 on Build GRCh38
rs765793438 Chr.11: 63999618-63999618 on Build GRCh38
rs759056024 Chr.11: 63999706-63999706 on Build GRCh38
rs769058023 Chr.11: 63999768-63999768 on Build GRCh38
rs776798912 Chr.11: 63999661-63999661 on Build GRCh38
rs951212920 Chr.11: 63999637-63999637 on Build GRCh38
rs778942047 Chr.11: 63999650-63999650 on Build GRCh38
rs758339227 Chr.11: 63999728-63999728 on Build GRCh38
rs200636276 Chr.11: 63999753-63999753 on Build GRCh38
rs1007527120 Chr.11: 63999675-63999675 on Build GRCh38
rs368583412 Chr.11: 63999628-63999628 on Build GRCh38
rs769902795 Chr.11: 63999761-63999761 on Build GRCh38
rs571362317 Chr.11: 63999655-63999655 on Build GRCh38
rs774702446 Chr.11: 63999704-63999704 on Build GRCh38
rs755830490 Chr.11: 63999641-63999641 on Build GRCh38
rs764514398 Chr.11: 63999696-63999696 on Build GRCh38
rs749736167 Chr.11: 63999763-63999763 on Build GRCh38
rs753359838 Chr.11: 63999646-63999646 on Build GRCh38
rs199876740 Chr.11: 63999694-63999694 on Build GRCh38
rs765217961 Chr.11: 63999722-63999739 on Build GRCh38
rs752727150 Chr.11: 63999767-63999767 on Build GRCh38
rs758192787 Chr.11: 63999656-63999656 on Build GRCh38
rs769801277 Chr.11: 63999669-63999669 on Build GRCh38
rs752972311 Chr.11: 63999775-63999775 on Build GRCh38
rs555366590 Chr.11: 63999783-63999783 on Build GRCh38

View Full Product Details