Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33391867-33392110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIFC1
Entrez Gene ID
3833
Gene Name
kinesin family member C1
Gene Aliases
HSET, KNSL2
UniGene
Hs.436912
Gene Chromosome Location
Chr.6: 33391524-33410226 on Build GRCh38
Transcripts
NM_002263.3 XM_011514585.1 XM_011514587.2 XM_017010836.1 XM_017010837.1

SNP Detail

SNP ID Chromosome Location
rs547910392 Chr.6: 33391901-33391901 on Build GRCh38
rs756550103 Chr.6: 33391989-33391989 on Build GRCh38
rs774644847 Chr.6: 33391977-33391977 on Build GRCh38
rs763321781 Chr.6: 33391975-33391975 on Build GRCh38
rs746897562 Chr.6: 33391964-33391964 on Build GRCh38
rs965523996 Chr.6: 33392069-33392069 on Build GRCh38
rs1019262606 Chr.6: 33392068-33392068 on Build GRCh38
rs754623697 Chr.6: 33391942-33391942 on Build GRCh38
rs758262313 Chr.6: 33392035-33392035 on Build GRCh38
rs767729115 Chr.6: 33391980-33391980 on Build GRCh38
rs113816169 Chr.6: 33391903-33391903 on Build GRCh38
rs465223 Chr.6: 33391939-33391939 on Build GRCh38
rs372226649 Chr.6: 33391937-33391937 on Build GRCh38
rs764795775 Chr.6: 33391994-33391994 on Build GRCh38
rs375216142 Chr.6: 33391947-33391947 on Build GRCh38
rs777398278 Chr.6: 33392037-33392037 on Build GRCh38
rs773222863 Chr.6: 33391961-33391961 on Build GRCh38
rs369275310 Chr.6: 33391995-33391995 on Build GRCh38
rs762296312 Chr.6: 33391979-33391979 on Build GRCh38
rs754429042 Chr.6: 33392007-33392007 on Build GRCh38
rs972472281 Chr.6: 33392074-33392074 on Build GRCh38
rs369942319 Chr.6: 33392040-33392040 on Build GRCh38
rs769955814 Chr.6: 33391967-33391967 on Build GRCh38
rs762704727 Chr.6: 33392010-33392010 on Build GRCh38
rs750992525 Chr.6: 33391986-33391986 on Build GRCh38
rs778738706 Chr.6: 33391944-33391944 on Build GRCh38
rs172435 Chr.6: 33392043-33392043 on Build GRCh38
rs569647725 Chr.6: 33391932-33391932 on Build GRCh38
rs1006937668 Chr.6: 33392055-33392055 on Build GRCh38
rs777586516 Chr.6: 33391935-33391935 on Build GRCh38
rs45521935 Chr.6: 33391959-33391959 on Build GRCh38
rs747951649 Chr.6: 33391958-33391958 on Build GRCh38
rs778542318 Chr.6: 33392008-33392008 on Build GRCh38
rs775219353 Chr.6: 33391944-33391945 on Build GRCh38
rs768070174 Chr.6: 33391936-33391936 on Build GRCh38
rs199721284 Chr.6: 33391927-33391927 on Build GRCh38
rs755675644 Chr.6: 33391938-33391938 on Build GRCh38
rs929900044 Chr.6: 33392073-33392073 on Build GRCh38
rs377116917 Chr.6: 33392048-33392048 on Build GRCh38
rs764421777 Chr.6: 33391976-33391976 on Build GRCh38
rs146617967 Chr.6: 33391895-33391895 on Build GRCh38
rs1029619514 Chr.6: 33392038-33392038 on Build GRCh38
rs918485260 Chr.6: 33392056-33392056 on Build GRCh38
rs200816546 Chr.6: 33391973-33391973 on Build GRCh38
rs747759152 Chr.6: 33392009-33392009 on Build GRCh38

View Full Product Details