Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127818123-127818376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENG
Entrez Gene ID
2022
Gene Name
endoglin
Gene Aliases
END, HHT1, ORW1
UniGene
Hs.76753
Gene Chromosome Location
Chr.9: 127815012-127854773 on Build GRCh38
Transcripts
NM_000118.3 NM_001114753.2 NM_001278138.1 NR_136302.1

Gene Symbol
LOC102723566
Entrez Gene ID
102723566
Gene Name
uncharacterized LOC102723566
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.9: 127816066-127822520 on Build GRCh38
Transcripts
NM_000118.3 NM_001114753.2 NM_001278138.1 NR_136302.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6337502 c.1583C>T p.P528L Chr.9: 127818223-127818223 on Build GRCh38
5422282 c.1528C>T p.R510W Chr.9: 127818278-127818278 on Build GRCh38
5422281 c.1528C>T p.R510W Chr.9: 127818278-127818278 on Build GRCh38
6337503 c.1583C>T p.P528L Chr.9: 127818223-127818223 on Build GRCh38
7418444 c.1466A>G p.Q489R Chr.9: 127818340-127818340 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150456852 Chr.9: 127818351-127818351 on Build GRCh38
rs748966307 Chr.9: 127818145-127818145 on Build GRCh38
rs754208447 Chr.9: 127818187-127818187 on Build GRCh38
rs929266862 Chr.9: 127818241-127818241 on Build GRCh38
rs770013214 Chr.9: 127818313-127818313 on Build GRCh38
rs766531566 Chr.9: 127818185-127818185 on Build GRCh38
rs768491175 Chr.9: 127818151-127818151 on Build GRCh38
rs780884220 Chr.9: 127818214-127818214 on Build GRCh38
rs375870845 Chr.9: 127818191-127818191 on Build GRCh38
rs144595802 Chr.9: 127818280-127818280 on Build GRCh38
rs760682477 Chr.9: 127818234-127818234 on Build GRCh38
rs765564489 Chr.9: 127818268-127818268 on Build GRCh38
rs754619120 Chr.9: 127818287-127818287 on Build GRCh38
rs532853893 Chr.9: 127818196-127818196 on Build GRCh38
rs941578347 Chr.9: 127818324-127818324 on Build GRCh38
rs910711416 Chr.9: 127818182-127818182 on Build GRCh38
rs764626017 Chr.9: 127818275-127818275 on Build GRCh38
rs774162691 Chr.9: 127818225-127818225 on Build GRCh38
rs997074986 Chr.9: 127818353-127818353 on Build GRCh38
rs775718026 Chr.9: 127818326-127818326 on Build GRCh38
rs140760635 Chr.9: 127818273-127818273 on Build GRCh38
rs200224098 Chr.9: 127818274-127818274 on Build GRCh38
rs779916731 Chr.9: 127818223-127818223 on Build GRCh38
rs777756221 Chr.9: 127818297-127818297 on Build GRCh38
rs746937879 Chr.9: 127818299-127818299 on Build GRCh38
rs747919839 Chr.9: 127818290-127818290 on Build GRCh38
rs766445175 Chr.9: 127818238-127818238 on Build GRCh38
rs116330805 Chr.9: 127818296-127818296 on Build GRCh38
rs147428654 Chr.9: 127818249-127818249 on Build GRCh38
rs199629212 Chr.9: 127818277-127818277 on Build GRCh38
rs142896669 Chr.9: 127818173-127818173 on Build GRCh38
rs755367205 Chr.9: 127818188-127818188 on Build GRCh38
rs886041325 Chr.9: 127818251-127818252 on Build GRCh38
rs201517535 Chr.9: 127818278-127818278 on Build GRCh38
rs148537759 Chr.9: 127818333-127818333 on Build GRCh38
rs759785487 Chr.9: 127818260-127818260 on Build GRCh38
rs863223541 Chr.9: 127818255-127818256 on Build GRCh38
rs771948067 Chr.9: 127818231-127818231 on Build GRCh38
rs570296266 Chr.9: 127818195-127818195 on Build GRCh38
rs189572386 Chr.9: 127818222-127818222 on Build GRCh38
rs764536505 Chr.9: 127818343-127818343 on Build GRCh38
rs372072854 Chr.9: 127818305-127818305 on Build GRCh38
rs117510900 Chr.9: 127818174-127818174 on Build GRCh38
rs554788017 Chr.9: 127818308-127818308 on Build GRCh38
rs773294798 Chr.9: 127818233-127818233 on Build GRCh38
rs115450389 Chr.9: 127818354-127818354 on Build GRCh38
rs763475207 Chr.9: 127818337-127818337 on Build GRCh38
rs979666386 Chr.9: 127818165-127818165 on Build GRCh38
rs759712732 Chr.9: 127818312-127818312 on Build GRCh38
rs372453075 Chr.9: 127818169-127818169 on Build GRCh38
rs778594104 Chr.9: 127818289-127818289 on Build GRCh38
rs369183236 Chr.9: 127818224-127818224 on Build GRCh38
rs863223538 Chr.9: 127818220-127818220 on Build GRCh38
rs753472534 Chr.9: 127818285-127818285 on Build GRCh38
rs745316066 Chr.9: 127818221-127818221 on Build GRCh38
rs372900775 Chr.9: 127818230-127818230 on Build GRCh38

View Full Product Details