Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 128846328-128846595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JADE1
Entrez Gene ID
79960
Gene Name
jade family PHD finger 1
Gene Aliases
PHF17
UniGene
Hs.12420
Gene Chromosome Location
Chr.4: 128809623-128875224 on Build GRCh38
Transcripts
NM_001287437.1 NM_001287439.1 NM_001287440.1 NM_001287441.1 NM_001287442.1 NM_001287443.1 NM_024900.4 NM_199320.3 XM_005263232.1 XM_017008626.1 XM_017008627.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3010224 c.260C>A p.P87H Chr.4: 128846496-128846496 on Build GRCh38
6587041 c.206A>T p.Y69F Chr.4: 128846442-128846442 on Build GRCh38
7309964 c.296+7A>T p.? Chr.4: 128846539-128846539 on Build GRCh38
4122215 c.179A>G p.D60G Chr.4: 128846415-128846415 on Build GRCh38
6587042 c.206A>T p.Y69F Chr.4: 128846442-128846442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751973244 Chr.4: 128846432-128846432 on Build GRCh38
rs1020572085 Chr.4: 128846401-128846401 on Build GRCh38
rs903025143 Chr.4: 128846526-128846526 on Build GRCh38
rs756884134 Chr.4: 128846482-128846482 on Build GRCh38
rs200609664 Chr.4: 128846434-128846434 on Build GRCh38
rs753409370 Chr.4: 128846436-128846436 on Build GRCh38
rs746734088 Chr.4: 128846363-128846363 on Build GRCh38
rs376255869 Chr.4: 128846519-128846519 on Build GRCh38
rs761933433 Chr.4: 128846380-128846380 on Build GRCh38
rs768314124 Chr.4: 128846509-128846509 on Build GRCh38
rs773296670 Chr.4: 128846547-128846547 on Build GRCh38
rs774685498 Chr.4: 128846575-128846575 on Build GRCh38
rs748265023 Chr.4: 128846541-128846542 on Build GRCh38
rs201663272 Chr.4: 128846364-128846364 on Build GRCh38
rs369236924 Chr.4: 128846525-128846525 on Build GRCh38
rs369937525 Chr.4: 128846424-128846424 on Build GRCh38
rs756164073 Chr.4: 128846500-128846500 on Build GRCh38
rs776686028 Chr.4: 128846370-128846370 on Build GRCh38
rs770064066 Chr.4: 128846569-128846569 on Build GRCh38
rs1011324195 Chr.4: 128846366-128846366 on Build GRCh38
rs759775725 Chr.4: 128846433-128846433 on Build GRCh38
rs945012862 Chr.4: 128846421-128846421 on Build GRCh38
rs746861939 Chr.4: 128846506-128846506 on Build GRCh38
rs766736396 Chr.4: 128846426-128846426 on Build GRCh38
rs760642234 Chr.4: 128846518-128846518 on Build GRCh38
rs749884285 Chr.4: 128846494-128846494 on Build GRCh38
rs200104707 Chr.4: 128846549-128846549 on Build GRCh38
rs771497213 Chr.4: 128846556-128846556 on Build GRCh38
rs775618201 Chr.4: 128846488-128846488 on Build GRCh38
rs773777832 Chr.4: 128846412-128846412 on Build GRCh38
rs1041995022 Chr.4: 128846425-128846425 on Build GRCh38
rs375719680 Chr.4: 128846410-128846410 on Build GRCh38
rs781538943 Chr.4: 128846511-128846511 on Build GRCh38
rs779817654 Chr.4: 128846505-128846505 on Build GRCh38

View Full Product Details