Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 27622740-27623013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR1D
Entrez Gene ID
51082
Gene Name
RNA polymerase I subunit D
Gene Aliases
AC19, POLR1C, RPA16, RPA9, RPAC2, RPC16, RPO1-3, TCS2
UniGene
Hs.507584
Gene Chromosome Location
Chr.13: 27620743-27667422 on Build GRCh38
Transcripts
NM_001206559.1 NM_015972.3 NM_152705.2 XM_005266412.1 XM_005266414.2 XM_017020623.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5980466 c.76G>A p.A26T Chr.13: 27622924-27622924 on Build GRCh38
1262713 c.115T>A p.C39S Chr.13: 27622963-27622963 on Build GRCh38
432262 c.161C>T p.S54F Chr.13: 27623009-27623009 on Build GRCh38
7082833 c.142G>T p.D48Y Chr.13: 27622990-27622990 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs569336578 Chr.13: 27622844-27622844 on Build GRCh38
rs780353340 Chr.13: 27622851-27622851 on Build GRCh38
rs982913005 Chr.13: 27622831-27622831 on Build GRCh38
rs532914429 Chr.13: 27622804-27622804 on Build GRCh38
rs372189960 Chr.13: 27622926-27622926 on Build GRCh38
rs768698541 Chr.13: 27622857-27622857 on Build GRCh38
rs774653944 Chr.13: 27622858-27622858 on Build GRCh38
rs751267443 Chr.13: 27622945-27622945 on Build GRCh38
rs749664600 Chr.13: 27622856-27622856 on Build GRCh38
rs932578285 Chr.13: 27622911-27622911 on Build GRCh38
rs1013753456 Chr.13: 27622882-27622882 on Build GRCh38
rs1010350773 Chr.13: 27622834-27622834 on Build GRCh38
rs151041219 Chr.13: 27622914-27622914 on Build GRCh38
rs959489852 Chr.13: 27622865-27622865 on Build GRCh38
rs1052076414 Chr.13: 27622819-27622819 on Build GRCh38
rs921178811 Chr.13: 27622895-27622895 on Build GRCh38
rs184084459 Chr.13: 27622839-27622839 on Build GRCh38
rs781757131 Chr.13: 27622833-27622833 on Build GRCh38
rs540248256 Chr.13: 27622818-27622818 on Build GRCh38
rs759527489 Chr.13: 27622909-27622909 on Build GRCh38
rs777412814 Chr.13: 27622922-27622922 on Build GRCh38
rs763616303 Chr.13: 27622927-27622927 on Build GRCh38
rs190982909 Chr.13: 27622802-27622802 on Build GRCh38
rs980403809 Chr.13: 27622775-27622775 on Build GRCh38
rs933248799 Chr.13: 27622805-27622805 on Build GRCh38
rs186081951 Chr.13: 27622783-27622783 on Build GRCh38
rs754908359 Chr.13: 27622836-27622836 on Build GRCh38
rs575836696 Chr.13: 27622774-27622774 on Build GRCh38
rs530941743 Chr.13: 27622867-27622867 on Build GRCh38
rs756808529 Chr.13: 27622954-27622954 on Build GRCh38
rs751506774 Chr.13: 27622832-27622833 on Build GRCh38
rs886050108 Chr.13: 27622972-27622972 on Build GRCh38
rs529013502 Chr.13: 27622902-27622902 on Build GRCh38
rs780656601 Chr.13: 27622864-27622864 on Build GRCh38
rs752153066 Chr.13: 27622890-27622892 on Build GRCh38
rs756538526 Chr.13: 27622849-27622849 on Build GRCh38
rs1053573832 Chr.13: 27622952-27622952 on Build GRCh38
rs952484501 Chr.13: 27622794-27622794 on Build GRCh38
rs765011144 Chr.13: 27622912-27622912 on Build GRCh38
rs776525888 Chr.13: 27622905-27622905 on Build GRCh38

View Full Product Details